Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103128576-103128827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAPEPLD
Entrez Gene ID
222236
Gene Name
N-acyl phosphatidylethanolamine phospholipase D
Gene Aliases
FMP30, NAPE-PLD
UniGene
Hs.324271
Gene Chromosome Location
Chr.7: 103099576-103149560 on Build GRCh38
Transcripts
NM_001122838.1 NM_198990.4 XM_005250214.4 XM_005250215.2 XM_005250218.2 XM_011515933.2 XM_017011854.1 XM_017011855.1 XM_017011856.1 XM_017011857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6478026 c.225T>A p.D75E Chr.7: 103128771-103128771 on Build GRCh38
5871432 c.204-3_204-2insT p.? Chr.7: 103128794-103128795 on Build GRCh38
4483429 c.245C>T p.S82F Chr.7: 103128751-103128751 on Build GRCh38
744360 c.146G>C p.R49T Chr.7: 103128631-103128631 on Build GRCh38
3631491 c.92C>T p.S31F Chr.7: 103128685-103128685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746865644 Chr.7: 103128780-103128780 on Build GRCh38
rs759829176 Chr.7: 103128693-103128693 on Build GRCh38
rs752733400 Chr.7: 103128737-103128737 on Build GRCh38
rs542549561 Chr.7: 103128771-103128771 on Build GRCh38
rs148947157 Chr.7: 103128692-103128692 on Build GRCh38
rs971415194 Chr.7: 103128683-103128683 on Build GRCh38
rs774800636 Chr.7: 103128794-103128794 on Build GRCh38
rs997237712 Chr.7: 103128733-103128733 on Build GRCh38
rs374854751 Chr.7: 103128690-103128690 on Build GRCh38
rs765265919 Chr.7: 103128634-103128645 on Build GRCh38
rs757595572 Chr.7: 103128707-103128707 on Build GRCh38
rs74898469 Chr.7: 103128797-103128797 on Build GRCh38
rs145821009 Chr.7: 103128691-103128691 on Build GRCh38
rs747555715 Chr.7: 103128785-103128785 on Build GRCh38
rs753989417 Chr.7: 103128664-103128664 on Build GRCh38
rs779977953 Chr.7: 103128772-103128772 on Build GRCh38
rs757135925 Chr.7: 103128665-103128665 on Build GRCh38
rs769298504 Chr.7: 103128792-103128792 on Build GRCh38
rs752921659 Chr.7: 103128619-103128619 on Build GRCh38
rs374468226 Chr.7: 103128717-103128717 on Build GRCh38
rs114728464 Chr.7: 103128629-103128629 on Build GRCh38
rs757368623 Chr.7: 103128745-103128745 on Build GRCh38
rs745597246 Chr.7: 103128676-103128676 on Build GRCh38
rs371491119 Chr.7: 103128783-103128783 on Build GRCh38
rs947585444 Chr.7: 103128655-103128655 on Build GRCh38
rs753109607 Chr.7: 103128706-103128706 on Build GRCh38
rs753846829 Chr.7: 103128740-103128740 on Build GRCh38
rs373405050 Chr.7: 103128623-103128623 on Build GRCh38
rs778924087 Chr.7: 103128746-103128746 on Build GRCh38
rs767899867 Chr.7: 103128697-103128697 on Build GRCh38
rs141112494 Chr.7: 103128613-103128613 on Build GRCh38
rs777691619 Chr.7: 103128652-103128652 on Build GRCh38
rs779808607 Chr.7: 103128681-103128681 on Build GRCh38
rs768407647 Chr.7: 103128684-103128684 on Build GRCh38
rs530322396 Chr.7: 103128766-103128766 on Build GRCh38
rs771994194 Chr.7: 103128680-103128680 on Build GRCh38
rs763951240 Chr.7: 103128794-103128795 on Build GRCh38
rs762593966 Chr.7: 103128795-103128795 on Build GRCh38
rs779048805 Chr.7: 103128668-103128668 on Build GRCh38
rs746685656 Chr.7: 103128682-103128682 on Build GRCh38
rs780958580 Chr.7: 103128784-103128784 on Build GRCh38
rs760013967 Chr.7: 103128795-103128795 on Build GRCh38

View Full Product Details