Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 22130918-22131191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAPGEF5
Entrez Gene ID
9771
Gene Name
Rap guanine nucleotide exchange factor 5
Gene Aliases
GFR, MR-GEF, REPAC
UniGene
Hs.174768
Gene Chromosome Location
Chr.7: 22118290-22357133 on Build GRCh38
Transcripts
NM_012294.3 XM_005249914.2 XM_005249916.1 XM_011515652.2 XM_017012837.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4452574 c.1118A>T p.E373V Chr.7: 22131083-22131083 on Build GRCh38
1223156 c.1972C>A p.H658N Chr.7: 22131087-22131087 on Build GRCh38
4970896 c.2022+5G>A p.? Chr.7: 22131032-22131032 on Build GRCh38
1223157 c.1114C>A p.H372N Chr.7: 22131087-22131087 on Build GRCh38
5517864 c.2022+4C>T p.? Chr.7: 22131033-22131033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776131293 Chr.7: 22130996-22130996 on Build GRCh38
rs544564359 Chr.7: 22131026-22131026 on Build GRCh38
rs1019024824 Chr.7: 22131154-22131154 on Build GRCh38
rs555374697 Chr.7: 22130953-22130953 on Build GRCh38
rs375982123 Chr.7: 22131021-22131021 on Build GRCh38
rs765816842 Chr.7: 22131133-22131133 on Build GRCh38
rs533630332 Chr.7: 22131128-22131128 on Build GRCh38
rs367909478 Chr.7: 22131033-22131033 on Build GRCh38
rs779005994 Chr.7: 22131155-22131155 on Build GRCh38
rs768231542 Chr.7: 22130971-22130971 on Build GRCh38
rs771006212 Chr.7: 22130961-22130961 on Build GRCh38
rs745906858 Chr.7: 22130991-22130991 on Build GRCh38
rs148125266 Chr.7: 22130980-22130980 on Build GRCh38
rs1003600243 Chr.7: 22131001-22131001 on Build GRCh38
rs1003410118 Chr.7: 22131020-22131020 on Build GRCh38
rs750013883 Chr.7: 22131119-22131119 on Build GRCh38
rs761551298 Chr.7: 22131059-22131059 on Build GRCh38
rs770494023 Chr.7: 22130992-22130992 on Build GRCh38
rs767044409 Chr.7: 22131086-22131086 on Build GRCh38
rs953215779 Chr.7: 22131061-22131061 on Build GRCh38
rs369371745 Chr.7: 22131030-22131030 on Build GRCh38
rs540519129 Chr.7: 22131147-22131147 on Build GRCh38
rs375339054 Chr.7: 22131018-22131018 on Build GRCh38
rs61758818 Chr.7: 22131100-22131100 on Build GRCh38
rs753776123 Chr.7: 22131136-22131136 on Build GRCh38
rs1027504814 Chr.7: 22131060-22131060 on Build GRCh38

View Full Product Details