Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87536349-87536594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB1
Entrez Gene ID
5243
Gene Name
ATP binding cassette subfamily B member 1
Gene Aliases
ABC20, CD243, CLCS, GP170, MDR1, P-GP, PGY1
UniGene
Hs.489033
Gene Chromosome Location
Chr.7: 87503863-87713323 on Build GRCh38
Transcripts
NM_000927.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4537929 c.2480G>A p.G827E Chr.7: 87536459-87536459 on Build GRCh38
4993025 c.2420C>T p.P807L Chr.7: 87536519-87536519 on Build GRCh38
7089556 c.2420C>A p.P807H Chr.7: 87536519-87536519 on Build GRCh38
5938175 c.2398G>A p.D800N Chr.7: 87536541-87536541 on Build GRCh38
5139801 c.2426delA p.N809fs*6 Chr.7: 87536513-87536513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151169676 Chr.7: 87536392-87536392 on Build GRCh38
rs768893111 Chr.7: 87536453-87536453 on Build GRCh38
rs2235038 Chr.7: 87536552-87536552 on Build GRCh38
rs759267875 Chr.7: 87536485-87536485 on Build GRCh38
rs201744003 Chr.7: 87536501-87536501 on Build GRCh38
rs769383225 Chr.7: 87536402-87536402 on Build GRCh38
rs199573028 Chr.7: 87536423-87536423 on Build GRCh38
rs2235043 Chr.7: 87536381-87536381 on Build GRCh38
rs767286079 Chr.7: 87536491-87536491 on Build GRCh38
rs929075205 Chr.7: 87536393-87536393 on Build GRCh38
rs2235042 Chr.7: 87536385-87536385 on Build GRCh38
rs200754866 Chr.7: 87536484-87536484 on Build GRCh38
rs2235041 Chr.7: 87536418-87536418 on Build GRCh38
rs2235039 Chr.7: 87536538-87536538 on Build GRCh38
rs2235040 Chr.7: 87536434-87536434 on Build GRCh38
rs899656547 Chr.7: 87536403-87536403 on Build GRCh38
rs41305517 Chr.7: 87536541-87536541 on Build GRCh38
rs776548654 Chr.7: 87536454-87536454 on Build GRCh38
rs962100921 Chr.7: 87536506-87536506 on Build GRCh38
rs891617033 Chr.7: 87536547-87536547 on Build GRCh38
rs201142514 Chr.7: 87536539-87536539 on Build GRCh38
rs199856295 Chr.7: 87536513-87536513 on Build GRCh38
rs34350171 Chr.7: 87536450-87536450 on Build GRCh38
rs200155994 Chr.7: 87536473-87536473 on Build GRCh38
rs763717036 Chr.7: 87536546-87536546 on Build GRCh38
rs200422791 Chr.7: 87536438-87536438 on Build GRCh38
rs764597620 Chr.7: 87536425-87536426 on Build GRCh38
rs764059820 Chr.7: 87536505-87536505 on Build GRCh38

View Full Product Details