Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31690424-31690697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD16A
Entrez Gene ID
7920
Gene Name
abhydrolase domain containing 16A
Gene Aliases
BAT5, D6S82E, NG26, PP199
UniGene
Hs.388188
Gene Chromosome Location
Chr.6: 31686949-31703360 on Build GRCh38
Transcripts
NM_001177515.1 NM_021160.2 NR_033488.1 NR_033489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6584611 c.877G>C p.E293Q Chr.6: 31690569-31690569 on Build GRCh38
5880357 c.889G>T p.V297F Chr.6: 31690557-31690557 on Build GRCh38
7214805 c.900C>T p.P300P Chr.6: 31690546-31690546 on Build GRCh38
1716014 c.904G>A p.E302K Chr.6: 31690542-31690542 on Build GRCh38
5870185 c.844-2A>T p.? Chr.6: 31690604-31690604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184095096 Chr.6: 31690606-31690606 on Build GRCh38
rs373535293 Chr.6: 31690668-31690668 on Build GRCh38
rs989117310 Chr.6: 31690647-31690647 on Build GRCh38
rs772162668 Chr.6: 31690629-31690629 on Build GRCh38
rs189527662 Chr.6: 31690509-31690509 on Build GRCh38
rs536796078 Chr.6: 31690492-31690492 on Build GRCh38
rs574673478 Chr.6: 31690630-31690630 on Build GRCh38
rs142905776 Chr.6: 31690550-31690550 on Build GRCh38
rs373253921 Chr.6: 31690576-31690576 on Build GRCh38
rs937646600 Chr.6: 31690641-31690641 on Build GRCh38
rs772480298 Chr.6: 31690618-31690619 on Build GRCh38
rs1017300875 Chr.6: 31690644-31690644 on Build GRCh38
rs779110521 Chr.6: 31690624-31690624 on Build GRCh38
rs141329347 Chr.6: 31690453-31690453 on Build GRCh38
rs545892145 Chr.6: 31690457-31690457 on Build GRCh38
rs749457656 Chr.6: 31690620-31690620 on Build GRCh38
rs769475244 Chr.6: 31690632-31690632 on Build GRCh38
rs780533614 Chr.6: 31690615-31690615 on Build GRCh38
rs750984440 Chr.6: 31690585-31690585 on Build GRCh38
rs751058351 Chr.6: 31690553-31690553 on Build GRCh38
rs565451421 Chr.6: 31690463-31690463 on Build GRCh38
rs759204224 Chr.6: 31690531-31690531 on Build GRCh38
rs143395858 Chr.6: 31690546-31690546 on Build GRCh38
rs780889400 Chr.6: 31690629-31690630 on Build GRCh38
rs762738392 Chr.6: 31690549-31690549 on Build GRCh38
rs748523966 Chr.6: 31690582-31690583 on Build GRCh38
rs576918622 Chr.6: 31690532-31690532 on Build GRCh38
rs748187015 Chr.6: 31690627-31690627 on Build GRCh38
rs755133692 Chr.6: 31690623-31690623 on Build GRCh38
rs752606968 Chr.6: 31690452-31690452 on Build GRCh38
rs756855546 Chr.6: 31690555-31690555 on Build GRCh38
rs770786822 Chr.6: 31690510-31690510 on Build GRCh38
rs776583010 Chr.6: 31690530-31690530 on Build GRCh38
rs775417208 Chr.6: 31690650-31690650 on Build GRCh38

View Full Product Details