Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146600240-146600513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2R2B
Entrez Gene ID
5521
Gene Name
protein phosphatase 2 regulatory subunit Bbeta
Gene Aliases
B55BETA, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55-BETA, PR55BETA, SCA12
UniGene
Hs.627618
Gene Chromosome Location
Chr.5: 146589504-147081520 on Build GRCh38
Transcripts
NM_001271899.1 NM_001271900.1 NM_001271948.1 NM_181674.2 NM_181675.3 NM_181676.2 NM_181677.2 NM_181678.2 NR_073526.1 NR_073527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6754235 c.907C>T p.R303C Chr.5: 146600311-146600311 on Build GRCh38
3852450 c.809A>G p.E270G Chr.5: 146600451-146600451 on Build GRCh38
6102452 c.1021T>G p.S341A Chr.5: 146600404-146600404 on Build GRCh38
3852446 c.863A>C p.K288T Chr.5: 146600388-146600388 on Build GRCh38
3919262 c.943G>A p.E315K Chr.5: 146600317-146600317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772162797 Chr.5: 146600476-146600476 on Build GRCh38
rs867967245 Chr.5: 146600406-146600406 on Build GRCh38
rs759906165 Chr.5: 146600441-146600441 on Build GRCh38
rs761316578 Chr.5: 146600285-146600285 on Build GRCh38
rs540200978 Chr.5: 146600271-146600271 on Build GRCh38
rs531328300 Chr.5: 146600366-146600366 on Build GRCh38
rs905001520 Chr.5: 146600296-146600296 on Build GRCh38
rs781142522 Chr.5: 146600357-146600357 on Build GRCh38
rs774151829 Chr.5: 146600428-146600428 on Build GRCh38
rs528909222 Chr.5: 146600286-146600286 on Build GRCh38
rs372515064 Chr.5: 146600371-146600371 on Build GRCh38
rs776442979 Chr.5: 146600282-146600282 on Build GRCh38
rs754730796 Chr.5: 146600351-146600351 on Build GRCh38
rs375413594 Chr.5: 146600303-146600303 on Build GRCh38
rs147524300 Chr.5: 146600298-146600298 on Build GRCh38
rs764126610 Chr.5: 146600335-146600335 on Build GRCh38
rs746745667 Chr.5: 146600347-146600347 on Build GRCh38
rs764153596 Chr.5: 146600471-146600471 on Build GRCh38
rs757541383 Chr.5: 146600342-146600342 on Build GRCh38
rs551016876 Chr.5: 146600456-146600456 on Build GRCh38
rs748939898 Chr.5: 146600402-146600402 on Build GRCh38
rs772927479 Chr.5: 146600396-146600396 on Build GRCh38
rs375376283 Chr.5: 146600314-146600314 on Build GRCh38
rs147383897 Chr.5: 146600447-146600447 on Build GRCh38
rs140308315 Chr.5: 146600310-146600310 on Build GRCh38
rs560393664 Chr.5: 146600280-146600280 on Build GRCh38
rs111346525 Chr.5: 146600345-146600345 on Build GRCh38
rs1004275629 Chr.5: 146600344-146600344 on Build GRCh38
rs770649547 Chr.5: 146600403-146600403 on Build GRCh38
rs756171345 Chr.5: 146600327-146600327 on Build GRCh38
rs771831241 Chr.5: 146600268-146600268 on Build GRCh38
rs148423117 Chr.5: 146600448-146600448 on Build GRCh38
rs369931023 Chr.5: 146600341-146600341 on Build GRCh38
rs772978732 Chr.5: 146600287-146600287 on Build GRCh38
rs768691920 Chr.5: 146600275-146600275 on Build GRCh38
rs768002675 Chr.5: 146600311-146600311 on Build GRCh38
rs1030537122 Chr.5: 146600467-146600467 on Build GRCh38
rs372129488 Chr.5: 146600269-146600269 on Build GRCh38

View Full Product Details