Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 121059884-121060153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBCEL
Entrez Gene ID
219899
Gene Name
tubulin folding cofactor E like
Gene Aliases
El, LRRC35
UniGene
Hs.504136
Gene Chromosome Location
Chr.11: 121024094-121090776 on Build GRCh38
Transcripts
NM_001130047.1 NM_152715.3 XM_017017326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1351949 c.879T>A p.T293T Chr.11: 121060008-121060008 on Build GRCh38
7285009 c.956+2_956+3insA p.? Chr.11: 121060087-121060088 on Build GRCh38
110365 c.940G>A p.E314K Chr.11: 121060069-121060069 on Build GRCh38
923986 c.844C>T p.P282S Chr.11: 121059973-121059973 on Build GRCh38
1166419 c.955A>G p.R319G Chr.11: 121060084-121060084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746429782 Chr.11: 121060001-121060001 on Build GRCh38
rs367973304 Chr.11: 121060124-121060124 on Build GRCh38
rs374353071 Chr.11: 121059955-121059955 on Build GRCh38
rs761896922 Chr.11: 121059936-121059936 on Build GRCh38
rs765527302 Chr.11: 121060087-121060088 on Build GRCh38
rs1032653069 Chr.11: 121060118-121060118 on Build GRCh38
rs751457280 Chr.11: 121059945-121059945 on Build GRCh38
rs779589116 Chr.11: 121060000-121060000 on Build GRCh38
rs760537257 Chr.11: 121059942-121059942 on Build GRCh38
rs146743853 Chr.11: 121060069-121060069 on Build GRCh38
rs775416567 Chr.11: 121059936-121059936 on Build GRCh38
rs752457707 Chr.11: 121059961-121059961 on Build GRCh38
rs370115170 Chr.11: 121060107-121060107 on Build GRCh38
rs971404322 Chr.11: 121059917-121059917 on Build GRCh38
rs761768726 Chr.11: 121060075-121060078 on Build GRCh38
rs901223646 Chr.11: 121060035-121060035 on Build GRCh38
rs753924237 Chr.11: 121059933-121059933 on Build GRCh38
rs766233344 Chr.11: 121059944-121059944 on Build GRCh38
rs765250285 Chr.11: 121060105-121060105 on Build GRCh38
rs772679731 Chr.11: 121060045-121060045 on Build GRCh38
rs748759546 Chr.11: 121059999-121059999 on Build GRCh38
rs781014409 Chr.11: 121059960-121059960 on Build GRCh38
rs755786235 Chr.11: 121059966-121059966 on Build GRCh38
rs577258957 Chr.11: 121060110-121060110 on Build GRCh38
rs776829720 Chr.11: 121060019-121060020 on Build GRCh38
rs761831946 Chr.11: 121060103-121060103 on Build GRCh38
rs761087746 Chr.11: 121060060-121060060 on Build GRCh38
rs377427847 Chr.11: 121060018-121060018 on Build GRCh38
rs776848591 Chr.11: 121060080-121060080 on Build GRCh38
rs768473506 Chr.11: 121059953-121059954 on Build GRCh38
rs556998901 Chr.11: 121059984-121059984 on Build GRCh38
rs1019792095 Chr.11: 121060008-121060008 on Build GRCh38
rs776067976 Chr.11: 121060054-121060054 on Build GRCh38
rs375293030 Chr.11: 121060115-121060115 on Build GRCh38

View Full Product Details