Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15238244-15238515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
UniGene
Hs.187763
Gene Chromosome Location
Chr.19: 15236836-15332543 on Build GRCh38
Transcripts
NM_058243.2 XM_011527854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
992123 c.4057C>A p.L1353I Chr.19: 15238409-15238409 on Build GRCh38
992122 c.4081C>A p.L1361I Chr.19: 15238385-15238385 on Build GRCh38
4422593 c.4021-8T>A p.? Chr.19: 15238453-15238453 on Build GRCh38
3529578 c.4047C>T p.F1349F Chr.19: 15238419-15238419 on Build GRCh38
6328240 c.4083T>C p.L1361L Chr.19: 15238383-15238383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757945788 Chr.19: 15238492-15238492 on Build GRCh38
rs777668547 Chr.19: 15238495-15238495 on Build GRCh38
rs202020423 Chr.19: 15238363-15238363 on Build GRCh38
rs199829327 Chr.19: 15238437-15238437 on Build GRCh38
rs200421172 Chr.19: 15238375-15238375 on Build GRCh38
rs566960746 Chr.19: 15238406-15238406 on Build GRCh38
rs905566309 Chr.19: 15238491-15238491 on Build GRCh38
rs201316873 Chr.19: 15238453-15238453 on Build GRCh38
rs376522537 Chr.19: 15238409-15238409 on Build GRCh38
rs200365284 Chr.19: 15238355-15238355 on Build GRCh38
rs769702248 Chr.19: 15238428-15238428 on Build GRCh38
rs201616082 Chr.19: 15238361-15238361 on Build GRCh38
rs199524813 Chr.19: 15238466-15238466 on Build GRCh38
rs200092853 Chr.19: 15238301-15238301 on Build GRCh38
rs878965476 Chr.19: 15238484-15238484 on Build GRCh38
rs778489901 Chr.19: 15238360-15238360 on Build GRCh38
rs369068599 Chr.19: 15238374-15238374 on Build GRCh38
rs758284750 Chr.19: 15238475-15238475 on Build GRCh38
rs1035884192 Chr.19: 15238262-15238262 on Build GRCh38
rs767955897 Chr.19: 15238461-15238461 on Build GRCh38
rs201626544 Chr.19: 15238348-15238348 on Build GRCh38
rs764751035 Chr.19: 15238335-15238335 on Build GRCh38
rs201672958 Chr.19: 15238311-15238311 on Build GRCh38
rs777756355 Chr.19: 15238366-15238366 on Build GRCh38
rs964219012 Chr.19: 15238306-15238306 on Build GRCh38
rs200850506 Chr.19: 15238471-15238471 on Build GRCh38
rs763189436 Chr.19: 15238451-15238451 on Build GRCh38
rs201354263 Chr.19: 15238386-15238386 on Build GRCh38
rs146914097 Chr.19: 15238407-15238407 on Build GRCh38
rs200858283 Chr.19: 15238310-15238310 on Build GRCh38
rs373651760 Chr.19: 15238345-15238345 on Build GRCh38
rs751965642 Chr.19: 15238336-15238336 on Build GRCh38
rs530582261 Chr.19: 15238352-15238352 on Build GRCh38
rs763563522 Chr.19: 15238329-15238329 on Build GRCh38
rs200691233 Chr.19: 15238452-15238452 on Build GRCh38
rs528104254 Chr.19: 15238263-15238263 on Build GRCh38
rs752202032 Chr.19: 15238481-15238481 on Build GRCh38
rs541498496 Chr.19: 15238289-15238289 on Build GRCh38
rs912468726 Chr.19: 15238400-15238400 on Build GRCh38
rs199890538 Chr.19: 15238316-15238316 on Build GRCh38
rs928279685 Chr.19: 15238342-15238342 on Build GRCh38
rs200073875 Chr.19: 15238326-15238326 on Build GRCh38
rs202240581 Chr.19: 15238324-15238324 on Build GRCh38
rs775045605 Chr.19: 15238459-15238459 on Build GRCh38
rs753364398 Chr.19: 15238463-15238463 on Build GRCh38
rs187083313 Chr.19: 15238314-15238314 on Build GRCh38
rs868647992 Chr.19: 15238403-15238403 on Build GRCh38
rs765924158 Chr.19: 15238351-15238351 on Build GRCh38
rs200971099 Chr.19: 15238323-15238323 on Build GRCh38
rs764928135 Chr.19: 15238474-15238474 on Build GRCh38
rs758318108 Chr.19: 15238364-15238364 on Build GRCh38
rs200882370 Chr.19: 15238344-15238344 on Build GRCh38
rs201664432 Chr.19: 15238290-15238290 on Build GRCh38

View Full Product Details