Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232458747-232459010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPI
Entrez Gene ID
248
Gene Name
alkaline phosphatase, intestinal
Gene Aliases
IAP
UniGene
Hs.37009
Gene Chromosome Location
Chr.2: 232456112-232460745 on Build GRCh38
Transcripts
NM_001631.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5746062 c.1439T>A p.V480D Chr.2: 232458998-232458998 on Build GRCh38
5344103 c.1365C>T p.D455D Chr.2: 232458924-232458924 on Build GRCh38
1017815 c.1433C>T p.A478V Chr.2: 232458992-232458992 on Build GRCh38
6598740 c.1344C>T p.S448S Chr.2: 232458903-232458903 on Build GRCh38
5862007 c.1301-1G>A p.? Chr.2: 232458859-232458859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147819122 Chr.2: 232458887-232458887 on Build GRCh38
rs761937885 Chr.2: 232458770-232458770 on Build GRCh38
rs377199045 Chr.2: 232458953-232458953 on Build GRCh38
rs978168253 Chr.2: 232458817-232458817 on Build GRCh38
rs754655247 Chr.2: 232458800-232458800 on Build GRCh38
rs200119638 Chr.2: 232458932-232458932 on Build GRCh38
rs955598502 Chr.2: 232458990-232458990 on Build GRCh38
rs747092209 Chr.2: 232458989-232458989 on Build GRCh38
rs772392101 Chr.2: 232458916-232458916 on Build GRCh38
rs369465035 Chr.2: 232458957-232458957 on Build GRCh38
rs973255537 Chr.2: 232458963-232458963 on Build GRCh38
rs765357281 Chr.2: 232458880-232458881 on Build GRCh38
rs760826298 Chr.2: 232458924-232458924 on Build GRCh38
rs746903330 Chr.2: 232458823-232458823 on Build GRCh38
rs146167085 Chr.2: 232458915-232458915 on Build GRCh38
rs375115516 Chr.2: 232458918-232458918 on Build GRCh38
rs756695623 Chr.2: 232458947-232458947 on Build GRCh38
rs867126363 Chr.2: 232458954-232458954 on Build GRCh38
rs745776528 Chr.2: 232458949-232458949 on Build GRCh38
rs754224263 Chr.2: 232458799-232458799 on Build GRCh38
rs773493934 Chr.2: 232458923-232458923 on Build GRCh38
rs761801596 Chr.2: 232458857-232458857 on Build GRCh38
rs781110431 Chr.2: 232458948-232458948 on Build GRCh38
rs961453382 Chr.2: 232458922-232458922 on Build GRCh38
rs764342811 Chr.2: 232458768-232458768 on Build GRCh38
rs1020632792 Chr.2: 232458867-232458867 on Build GRCh38
rs768816700 Chr.2: 232458845-232458845 on Build GRCh38
rs1005350424 Chr.2: 232458910-232458910 on Build GRCh38
rs373643375 Chr.2: 232458960-232458960 on Build GRCh38
rs756577443 Chr.2: 232458790-232458790 on Build GRCh38
rs922607849 Chr.2: 232458967-232458967 on Build GRCh38
rs567870283 Chr.2: 232458809-232458809 on Build GRCh38
rs758104990 Chr.2: 232458940-232458940 on Build GRCh38
rs936187870 Chr.2: 232458807-232458807 on Build GRCh38
rs960716702 Chr.2: 232458846-232458846 on Build GRCh38
rs139666979 Chr.2: 232458933-232458934 on Build GRCh38
rs772481331 Chr.2: 232458965-232458965 on Build GRCh38
rs747627424 Chr.2: 232458813-232458813 on Build GRCh38
rs781092139 Chr.2: 232458899-232458899 on Build GRCh38
rs755882618 Chr.2: 232458952-232458952 on Build GRCh38
rs547893587 Chr.2: 232458787-232458787 on Build GRCh38
rs552251603 Chr.2: 232458863-232458863 on Build GRCh38
rs146536956 Chr.2: 232458889-232458889 on Build GRCh38
rs766989978 Chr.2: 232458871-232458871 on Build GRCh38
rs2678480 Chr.2: 232458815-232458815 on Build GRCh38
rs1016948809 Chr.2: 232458912-232458912 on Build GRCh38
rs780614557 Chr.2: 232458795-232458795 on Build GRCh38
rs1006999120 Chr.2: 232458779-232458779 on Build GRCh38
rs776446666 Chr.2: 232458824-232458824 on Build GRCh38
rs369043469 Chr.2: 232458865-232458865 on Build GRCh38
rs751654749 Chr.2: 232458769-232458769 on Build GRCh38
rs150732356 Chr.2: 232458925-232458925 on Build GRCh38
rs148157889 Chr.2: 232458906-232458906 on Build GRCh38
rs200208371 Chr.2: 232458883-232458883 on Build GRCh38
rs372776745 Chr.2: 232458868-232458868 on Build GRCh38
rs774467960 Chr.2: 232458909-232458909 on Build GRCh38
rs767921465 Chr.2: 232458781-232458781 on Build GRCh38
rs200403345 Chr.2: 232458874-232458874 on Build GRCh38
rs145542166 Chr.2: 232458891-232458891 on Build GRCh38
rs751124232 Chr.2: 232458945-232458945 on Build GRCh38
rs759852158 Chr.2: 232458929-232458929 on Build GRCh38
rs368265920 Chr.2: 232458942-232458942 on Build GRCh38
rs773099520 Chr.2: 232458936-232458937 on Build GRCh38
rs774572512 Chr.2: 232458854-232458854 on Build GRCh38
rs771097063 Chr.2: 232458993-232458993 on Build GRCh38
rs202195901 Chr.2: 232458978-232458978 on Build GRCh38
rs757249017 Chr.2: 232458892-232458892 on Build GRCh38
rs553764942 Chr.2: 232458832-232458832 on Build GRCh38
rs890028990 Chr.2: 232458973-232458973 on Build GRCh38
rs1051315862 Chr.2: 232458939-232458939 on Build GRCh38
rs778436684 Chr.2: 232458885-232458885 on Build GRCh38
rs765592366 Chr.2: 232458930-232458930 on Build GRCh38

View Full Product Details