Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 90954406-90954639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCER1
Entrez Gene ID
196477
Gene Name
coiled-coil glutamate rich protein 1
Gene Aliases
C12orf12
UniGene
Hs.128574
Gene Chromosome Location
Chr.12: 90952215-90955176 on Build GRCh38
Transcripts
NM_152638.3 NR_130711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5704612 c.182G>C p.G61A Chr.12: 90954561-90954561 on Build GRCh38
5697777 c.208C>G p.Q70E Chr.12: 90954535-90954535 on Build GRCh38
944353 c.215G>A p.G72D Chr.12: 90954528-90954528 on Build GRCh38
4878377 c.334T>C p.Y112H Chr.12: 90954409-90954409 on Build GRCh38
3466341 c.199C>T p.P67S Chr.12: 90954544-90954544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768934116 Chr.12: 90954502-90954502 on Build GRCh38
rs748293418 Chr.12: 90954568-90954568 on Build GRCh38
rs375426730 Chr.12: 90954588-90954588 on Build GRCh38
rs925976939 Chr.12: 90954482-90954482 on Build GRCh38
rs780714455 Chr.12: 90954546-90954546 on Build GRCh38
rs200138098 Chr.12: 90954558-90954558 on Build GRCh38
rs1049296318 Chr.12: 90954603-90954603 on Build GRCh38
rs373647939 Chr.12: 90954512-90954512 on Build GRCh38
rs971411632 Chr.12: 90954544-90954544 on Build GRCh38
rs372103112 Chr.12: 90954587-90954587 on Build GRCh38
rs764977047 Chr.12: 90954607-90954607 on Build GRCh38
rs771937641 Chr.12: 90954582-90954582 on Build GRCh38
rs756710971 Chr.12: 90954459-90954459 on Build GRCh38
rs763699453 Chr.12: 90954497-90954498 on Build GRCh38
rs200693488 Chr.12: 90954515-90954515 on Build GRCh38
rs749980284 Chr.12: 90954538-90954538 on Build GRCh38
rs138371183 Chr.12: 90954533-90954533 on Build GRCh38
rs888143688 Chr.12: 90954604-90954604 on Build GRCh38
rs781647721 Chr.12: 90954562-90954562 on Build GRCh38
rs371781245 Chr.12: 90954484-90954484 on Build GRCh38
rs759419365 Chr.12: 90954520-90954520 on Build GRCh38
rs775391790 Chr.12: 90954586-90954586 on Build GRCh38
rs146867450 Chr.12: 90954591-90954591 on Build GRCh38
rs747323020 Chr.12: 90954550-90954550 on Build GRCh38
rs531881142 Chr.12: 90954428-90954428 on Build GRCh38
rs550837479 Chr.12: 90954471-90954471 on Build GRCh38
rs537131134 Chr.12: 90954615-90954615 on Build GRCh38
rs749373110 Chr.12: 90954579-90954579 on Build GRCh38
rs761323171 Chr.12: 90954602-90954602 on Build GRCh38
rs140727551 Chr.12: 90954599-90954599 on Build GRCh38
rs145112834 Chr.12: 90954578-90954578 on Build GRCh38
rs772759314 Chr.12: 90954466-90954466 on Build GRCh38
rs777293162 Chr.12: 90954456-90954456 on Build GRCh38
rs561247270 Chr.12: 90954525-90954525 on Build GRCh38
rs151076167 Chr.12: 90954530-90954530 on Build GRCh38
rs767277765 Chr.12: 90954429-90954429 on Build GRCh38
rs767473855 Chr.12: 90954522-90954522 on Build GRCh38
rs752506930 Chr.12: 90954449-90954449 on Build GRCh38
rs141295104 Chr.12: 90954542-90954542 on Build GRCh38
rs759509093 Chr.12: 90954427-90954427 on Build GRCh38
rs778555550 Chr.12: 90954461-90954461 on Build GRCh38
rs760328847 Chr.12: 90954456-90954457 on Build GRCh38
rs1053636588 Chr.12: 90954509-90954509 on Build GRCh38
rs755291593 Chr.12: 90954552-90954552 on Build GRCh38
rs753429746 Chr.12: 90954532-90954532 on Build GRCh38
rs776702190 Chr.12: 90954503-90954503 on Build GRCh38
rs755720011 Chr.12: 90954453-90954453 on Build GRCh38
rs747410932 Chr.12: 90954489-90954489 on Build GRCh38
rs868391652 Chr.12: 90954491-90954491 on Build GRCh38
rs773235580 Chr.12: 90954573-90954573 on Build GRCh38
rs367748968 Chr.12: 90954569-90954569 on Build GRCh38
rs201979819 Chr.12: 90954457-90954457 on Build GRCh38
rs762160201 Chr.12: 90954513-90954513 on Build GRCh38
rs778264837 Chr.12: 90954535-90954535 on Build GRCh38
rs746350246 Chr.12: 90954465-90954465 on Build GRCh38
rs568255782 Chr.12: 90954597-90954597 on Build GRCh38
rs201194959 Chr.12: 90954506-90954506 on Build GRCh38
rs766046971 Chr.12: 90954621-90954621 on Build GRCh38
rs751866539 Chr.12: 90954434-90954434 on Build GRCh38

View Full Product Details