Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29149084-29149349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K7CL
Entrez Gene ID
56911
Gene Name
MAP3K7 C-terminal like
Gene Aliases
C21orf7, HC21ORF7, TAK1L, TAKL, TAKL-1, TAKL-2, TAKL-4
UniGene
Hs.222802
Gene Chromosome Location
Chr.21: 29077471-29175889 on Build GRCh38
Transcripts
NM_001286617.1 NM_001286618.1 NM_001286619.1 NM_001286620.1 NM_001286622.1 NM_001286624.1 NM_001286634.1 NM_020152.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6446750 c.371-1G>C p.? Chr.21: 29149188-29149188 on Build GRCh38
3735866 c.80C>T p.P27L Chr.21: 29149198-29149198 on Build GRCh38
1029780 c.397C>A p.P133T Chr.21: 29149215-29149215 on Build GRCh38
3550055 c.397C>T p.P133S Chr.21: 29149215-29149215 on Build GRCh38
6446752 c.53-1G>C p.? Chr.21: 29149188-29149188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002100725 Chr.21: 29149256-29149256 on Build GRCh38
rs957838946 Chr.21: 29149300-29149300 on Build GRCh38
rs754231178 Chr.21: 29149245-29149245 on Build GRCh38
rs770043335 Chr.21: 29149265-29149265 on Build GRCh38
rs1032124112 Chr.21: 29149290-29149290 on Build GRCh38
rs78743527 Chr.21: 29149144-29149144 on Build GRCh38
rs1031312176 Chr.21: 29149198-29149198 on Build GRCh38
rs751315194 Chr.21: 29149189-29149189 on Build GRCh38
rs748919964 Chr.21: 29149226-29149226 on Build GRCh38
rs754885042 Chr.21: 29149190-29149190 on Build GRCh38
rs367876875 Chr.21: 29149194-29149194 on Build GRCh38
rs375740460 Chr.21: 29149156-29149156 on Build GRCh38
rs1021417308 Chr.21: 29149306-29149306 on Build GRCh38
rs769277267 Chr.21: 29149248-29149248 on Build GRCh38
rs1037868965 Chr.21: 29149133-29149133 on Build GRCh38
rs774182788 Chr.21: 29149296-29149296 on Build GRCh38
rs377546025 Chr.21: 29149282-29149282 on Build GRCh38
rs750714284 Chr.21: 29149157-29149157 on Build GRCh38
rs777378620 Chr.21: 29149210-29149210 on Build GRCh38
rs773189322 Chr.21: 29149271-29149271 on Build GRCh38
rs144808763 Chr.21: 29149305-29149305 on Build GRCh38
rs573439743 Chr.21: 29149281-29149281 on Build GRCh38
rs777348854 Chr.21: 29149250-29149250 on Build GRCh38
rs762636643 Chr.21: 29149163-29149163 on Build GRCh38
rs1017935712 Chr.21: 29149241-29149241 on Build GRCh38
rs769293543 Chr.21: 29149138-29149138 on Build GRCh38
rs756888459 Chr.21: 29149228-29149228 on Build GRCh38
rs780402725 Chr.21: 29149238-29149238 on Build GRCh38
rs766150163 Chr.21: 29149168-29149168 on Build GRCh38
rs372225924 Chr.21: 29149212-29149212 on Build GRCh38
rs752198255 Chr.21: 29149197-29149197 on Build GRCh38
rs886866649 Chr.21: 29149174-29149174 on Build GRCh38
rs140780164 Chr.21: 29149208-29149208 on Build GRCh38
rs748208449 Chr.21: 29149263-29149263 on Build GRCh38
rs878950350 Chr.21: 29149213-29149213 on Build GRCh38
rs762045309 Chr.21: 29149147-29149147 on Build GRCh38

View Full Product Details