Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31721683-31721936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABALON
Entrez Gene ID
103021294
Gene Name
apoptotic BCL2L1-antisense long non-coding RNA
Gene Aliases
INXS
UniGene
Gene Chromosome Location
Chr.20: 31721507-31723409 on Build GRCh38
Transcripts
NR_131907.1 NM_001191.3 NM_001317919.1 NM_001317920.1 NM_001317921.1 NM_001322239.1 NM_001322240.1 NM_001322242.1 NM_138578.2 NR_134257.1 XM_011528964.2 XM_017027993.1

Gene Symbol
BCL2L1
Entrez Gene ID
598
Gene Name
BCL2 like 1
Gene Aliases
BCL-XL/S, BCL2L, BCLX, Bcl-X, PPP1R52
UniGene
Hs.516966
Gene Chromosome Location
Chr.20: 31664452-31723999 on Build GRCh38
Transcripts
NR_131907.1 NM_001191.3 NM_001317919.1 NM_001317920.1 NM_001317921.1 NM_001322239.1 NM_001322240.1 NM_001322242.1 NM_138578.2 NR_134257.1 XM_011528964.2 XM_017027993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6008546 c.401G>A p.G134E Chr.20: 31721818-31721818 on Build GRCh38
1246044 c.319G>A p.D107N Chr.20: 31721900-31721900 on Build GRCh38
1246043 c.493C>T p.R165W Chr.20: 31721726-31721726 on Build GRCh38
5857600 c.427T>C p.F143L Chr.20: 31721792-31721792 on Build GRCh38
1025598 c.524A>C p.N175T Chr.20: 31721695-31721695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374318445 Chr.20: 31721892-31721892 on Build GRCh38
rs145707771 Chr.20: 31721781-31721781 on Build GRCh38
rs761080342 Chr.20: 31721763-31721763 on Build GRCh38
rs768364028 Chr.20: 31721748-31721748 on Build GRCh38
rs377336613 Chr.20: 31721838-31721838 on Build GRCh38
rs1040490728 Chr.20: 31721738-31721738 on Build GRCh38
rs748329440 Chr.20: 31721756-31721756 on Build GRCh38
rs771436075 Chr.20: 31721872-31721872 on Build GRCh38
rs766818591 Chr.20: 31721765-31721765 on Build GRCh38
rs774178481 Chr.20: 31721869-31721869 on Build GRCh38
rs141475590 Chr.20: 31721856-31721856 on Build GRCh38
rs769242031 Chr.20: 31721715-31721715 on Build GRCh38
rs139457299 Chr.20: 31721896-31721896 on Build GRCh38
rs940931151 Chr.20: 31721734-31721734 on Build GRCh38
rs561461281 Chr.20: 31721793-31721793 on Build GRCh38
rs868063030 Chr.20: 31721914-31721914 on Build GRCh38
rs757071036 Chr.20: 31721803-31721803 on Build GRCh38
rs765234540 Chr.20: 31721775-31721775 on Build GRCh38
rs773815952 Chr.20: 31721853-31721853 on Build GRCh38
rs1011204866 Chr.20: 31721909-31721909 on Build GRCh38
rs751391960 Chr.20: 31721802-31721802 on Build GRCh38
rs773733186 Chr.20: 31721755-31721755 on Build GRCh38
rs756076815 Chr.20: 31721824-31721824 on Build GRCh38
rs748811612 Chr.20: 31721861-31721861 on Build GRCh38
rs780912099 Chr.20: 31721806-31721806 on Build GRCh38
rs776985963 Chr.20: 31721873-31721873 on Build GRCh38
rs199692294 Chr.20: 31721719-31721719 on Build GRCh38
rs201719152 Chr.20: 31721772-31721772 on Build GRCh38
rs372986095 Chr.20: 31721720-31721720 on Build GRCh38
rs768464418 Chr.20: 31721867-31721867 on Build GRCh38
rs745800708 Chr.20: 31721811-31721811 on Build GRCh38
rs751797236 Chr.20: 31721910-31721910 on Build GRCh38
rs111521268 Chr.20: 31721829-31721829 on Build GRCh38
rs369606077 Chr.20: 31721871-31721871 on Build GRCh38
rs758675098 Chr.20: 31721784-31721784 on Build GRCh38
rs764114817 Chr.20: 31721907-31721907 on Build GRCh38
rs1036461217 Chr.20: 31721854-31721854 on Build GRCh38
rs148962546 Chr.20: 31721874-31721874 on Build GRCh38
rs762514744 Chr.20: 31721721-31721721 on Build GRCh38
rs754323913 Chr.20: 31721766-31721766 on Build GRCh38

View Full Product Details