Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53963238-53963487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG8
Entrez Gene ID
59283
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 8
Gene Aliases
-
UniGene
Hs.631595
Gene Chromosome Location
Chr.19: 53963036-53990215 on Build GRCh38
Transcripts
NM_031895.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
713821 c.196C>A p.P66T Chr.19: 53963338-53963338 on Build GRCh38
4132417 c.168C>A p.N56K Chr.19: 53963310-53963310 on Build GRCh38
1172658 c.235C>T p.P79S Chr.19: 53963377-53963377 on Build GRCh38
6618747 c.178G>A p.G60S Chr.19: 53963320-53963320 on Build GRCh38
6618746 c.179G>C p.G60A Chr.19: 53963321-53963321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756805616 Chr.19: 53963465-53963465 on Build GRCh38
rs1034925737 Chr.19: 53963356-53963356 on Build GRCh38
rs746437541 Chr.19: 53963453-53963453 on Build GRCh38
rs1004840584 Chr.19: 53963340-53963340 on Build GRCh38
rs550251807 Chr.19: 53963438-53963438 on Build GRCh38
rs943685270 Chr.19: 53963452-53963452 on Build GRCh38
rs546450727 Chr.19: 53963282-53963282 on Build GRCh38
rs757896358 Chr.19: 53963322-53963322 on Build GRCh38
rs751771188 Chr.19: 53963295-53963295 on Build GRCh38
rs767581716 Chr.19: 53963396-53963396 on Build GRCh38
rs779504131 Chr.19: 53963449-53963449 on Build GRCh38
rs775561047 Chr.19: 53963359-53963359 on Build GRCh38
rs1036844702 Chr.19: 53963364-53963364 on Build GRCh38
rs757374297 Chr.19: 53963303-53963303 on Build GRCh38
rs755389760 Chr.19: 53963443-53963443 on Build GRCh38
rs745596933 Chr.19: 53963349-53963349 on Build GRCh38
rs756488946 Chr.19: 53963316-53963316 on Build GRCh38
rs878927192 Chr.19: 53963306-53963306 on Build GRCh38
rs765329910 Chr.19: 53963347-53963347 on Build GRCh38
rs763107632 Chr.19: 53963285-53963285 on Build GRCh38
rs913561536 Chr.19: 53963444-53963444 on Build GRCh38
rs776350647 Chr.19: 53963336-53963336 on Build GRCh38
rs376239386 Chr.19: 53963363-53963363 on Build GRCh38
rs750723024 Chr.19: 53963315-53963315 on Build GRCh38
rs989054432 Chr.19: 53963440-53963440 on Build GRCh38
rs994569352 Chr.19: 53963323-53963323 on Build GRCh38
rs750702386 Chr.19: 53963399-53963399 on Build GRCh38
rs746550882 Chr.19: 53963332-53963332 on Build GRCh38
rs766770976 Chr.19: 53963436-53963436 on Build GRCh38
rs756466739 Chr.19: 53963429-53963429 on Build GRCh38
rs1023448929 Chr.19: 53963374-53963374 on Build GRCh38
rs774258878 Chr.19: 53963376-53963376 on Build GRCh38
rs770449989 Chr.19: 53963334-53963334 on Build GRCh38
rs761960382 Chr.19: 53963381-53963381 on Build GRCh38
rs1027739937 Chr.19: 53963330-53963331 on Build GRCh38
rs532141444 Chr.19: 53963373-53963373 on Build GRCh38
rs765198697 Chr.19: 53963272-53963272 on Build GRCh38
rs781510400 Chr.19: 53963304-53963304 on Build GRCh38
rs777195304 Chr.19: 53963326-53963326 on Build GRCh38
rs769765479 Chr.19: 53963353-53963353 on Build GRCh38
rs886064233 Chr.19: 53963331-53963331 on Build GRCh38
rs565070833 Chr.19: 53963320-53963320 on Build GRCh38
rs764144621 Chr.19: 53963286-53963286 on Build GRCh38

View Full Product Details