The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127457.2 | NP_001120929.1 | 10 - 11 | 1589 | 123 |
NM_021117.3 | NP_066940.2 | 10 - 11 | 1725 | 123 | |
GenBank mRNA | AB014558.1 | - | 10 - 11 | 1629 | 123 |
AK291260.1 | - | 10 - 11 | 1656 | 123 | |
AK294904.1 | - | 10 - 11 | 1583 | 123 | |
AK295627.1 | - | 6 - 7 | 1125 | 123 | |
AK302865.1 | - | 10 - 11 | 1725 | 123 | |
BC035161.1 | - | 12 - 13 | 1988 | 123 | |
BC041814.1 | - | 10 - 11 | 1653 | 123 | |
EU219622.1 | - | 10 - 11 | 1648 | 123 |