Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70407430-70407938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTTN
Entrez Gene ID
2017
Gene Name
cortactin
Gene Aliases
EMS1
UniGene
Hs.596164
Gene Chromosome Location
Chr.11: 70398506-70436584 on Build GRCh38
Transcripts
NM_001184740.1 NM_005231.3 NM_138565.2 XM_006718447.2 XM_006718448.3 XM_017017312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1741890 c.88-8C>G p.? Chr.11: 70407510-70407510 on Build GRCh38
6315500 c.137G>T p.G46V Chr.11: 70407567-70407567 on Build GRCh38
4036297 c.120T>C p.G40G Chr.11: 70407550-70407550 on Build GRCh38
6315498 c.137G>T p.G46V Chr.11: 70407567-70407567 on Build GRCh38
4036295 c.120T>C p.G40G Chr.11: 70407550-70407550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576866735 Chr.11: 70407888-70407888 on Build GRCh38
rs985520074 Chr.11: 70407668-70407668 on Build GRCh38
rs370832342 Chr.11: 70407473-70407473 on Build GRCh38
rs758315252 Chr.11: 70407476-70407476 on Build GRCh38
rs762856289 Chr.11: 70407599-70407599 on Build GRCh38
rs976041527 Chr.11: 70407728-70407728 on Build GRCh38
rs781179465 Chr.11: 70407479-70407479 on Build GRCh38
rs386754728 Chr.11: 70407480-70407482 on Build GRCh38
rs755166324 Chr.11: 70407616-70407616 on Build GRCh38
rs377265857 Chr.11: 70407618-70407618 on Build GRCh38
rs534467189 Chr.11: 70407868-70407868 on Build GRCh38
rs140774618 Chr.11: 70407805-70407805 on Build GRCh38
rs1056775753 Chr.11: 70407898-70407898 on Build GRCh38
rs914709185 Chr.11: 70407839-70407839 on Build GRCh38
rs1035337954 Chr.11: 70407580-70407580 on Build GRCh38
rs142633573 Chr.11: 70407571-70407571 on Build GRCh38
rs752516344 Chr.11: 70407598-70407598 on Build GRCh38
rs377138627 Chr.11: 70407643-70407643 on Build GRCh38
rs114090250 Chr.11: 70407851-70407851 on Build GRCh38
rs567206365 Chr.11: 70407863-70407863 on Build GRCh38
rs745555974 Chr.11: 70407642-70407642 on Build GRCh38
rs778345095 Chr.11: 70407621-70407621 on Build GRCh38
rs928862625 Chr.11: 70407856-70407856 on Build GRCh38
rs199504305 Chr.11: 70407504-70407504 on Build GRCh38
rs562549352 Chr.11: 70407482-70407482 on Build GRCh38
rs760254247 Chr.11: 70407535-70407535 on Build GRCh38
rs893345265 Chr.11: 70407554-70407554 on Build GRCh38
rs776240454 Chr.11: 70407632-70407632 on Build GRCh38
rs558601511 Chr.11: 70407884-70407884 on Build GRCh38
rs934736305 Chr.11: 70407515-70407515 on Build GRCh38
rs759215358 Chr.11: 70407559-70407559 on Build GRCh38
rs536899030 Chr.11: 70407822-70407822 on Build GRCh38
rs371318858 Chr.11: 70407603-70407603 on Build GRCh38
rs529849470 Chr.11: 70407500-70407500 on Build GRCh38
rs764906374 Chr.11: 70407774-70407774 on Build GRCh38
rs373863481 Chr.11: 70407520-70407520 on Build GRCh38
rs924645688 Chr.11: 70407762-70407762 on Build GRCh38
rs776435275 Chr.11: 70407538-70407538 on Build GRCh38
rs368185387 Chr.11: 70407499-70407499 on Build GRCh38
rs372082242 Chr.11: 70407503-70407503 on Build GRCh38
rs766241826 Chr.11: 70407867-70407867 on Build GRCh38
rs934726747 Chr.11: 70407792-70407793 on Build GRCh38
rs150990706 Chr.11: 70407660-70407660 on Build GRCh38
rs1017493469 Chr.11: 70407702-70407702 on Build GRCh38
rs760640680 Chr.11: 70407675-70407675 on Build GRCh38
rs770497990 Chr.11: 70407536-70407536 on Build GRCh38
rs921757938 Chr.11: 70407659-70407659 on Build GRCh38
rs113659666 Chr.11: 70407592-70407592 on Build GRCh38
rs754094226 Chr.11: 70407612-70407612 on Build GRCh38
rs551015608 Chr.11: 70407724-70407724 on Build GRCh38
rs895563055 Chr.11: 70407907-70407907 on Build GRCh38
rs944052488 Chr.11: 70407704-70407704 on Build GRCh38
rs776159640 Chr.11: 70407600-70407600 on Build GRCh38
rs1027432046 Chr.11: 70407661-70407661 on Build GRCh38
rs201100144 Chr.11: 70407627-70407627 on Build GRCh38
rs966365383 Chr.11: 70407722-70407722 on Build GRCh38
rs770585892 Chr.11: 70407631-70407631 on Build GRCh38
rs569732 Chr.11: 70407480-70407480 on Build GRCh38
rs947653440 Chr.11: 70407844-70407844 on Build GRCh38
rs367719086 Chr.11: 70407522-70407522 on Build GRCh38
rs780063275 Chr.11: 70407611-70407611 on Build GRCh38
rs111344988 Chr.11: 70407593-70407593 on Build GRCh38
rs190831384 Chr.11: 70407471-70407471 on Build GRCh38
rs370048343 Chr.11: 70407617-70407617 on Build GRCh38
rs756397092 Chr.11: 70407481-70407481 on Build GRCh38
rs986386878 Chr.11: 70407684-70407684 on Build GRCh38
rs948893932 Chr.11: 70407845-70407845 on Build GRCh38
rs910524324 Chr.11: 70407477-70407477 on Build GRCh38
rs560054370 Chr.11: 70407601-70407601 on Build GRCh38

View Full Product Details