Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 110430090-110430619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDX1
Entrez Gene ID
2230
Gene Name
ferredoxin 1
Gene Aliases
ADX, FDX, LOH11CR1D
UniGene
Hs.744
Gene Chromosome Location
Chr.11: 110429937-110464884 on Build GRCh38
Transcripts
NM_004109.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799819 c.48C>T p.V16V Chr.11: 110430168-110430168 on Build GRCh38
7213374 c.43_60del18 p.V16_A21delVLGGPA Chr.11: 110430163-110430180 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs997248360 Chr.11: 110430534-110430534 on Build GRCh38
rs985941348 Chr.11: 110430410-110430410 on Build GRCh38
rs762228607 Chr.11: 110430320-110430320 on Build GRCh38
rs1021867914 Chr.11: 110430274-110430274 on Build GRCh38
rs1056622697 Chr.11: 110430529-110430529 on Build GRCh38
rs551316001 Chr.11: 110430409-110430409 on Build GRCh38
rs919270137 Chr.11: 110430242-110430242 on Build GRCh38
rs1044503183 Chr.11: 110430225-110430225 on Build GRCh38
rs902833409 Chr.11: 110430361-110430361 on Build GRCh38
rs1019965086 Chr.11: 110430138-110430138 on Build GRCh38
rs1030017881 Chr.11: 110430538-110430538 on Build GRCh38
rs766060368 Chr.11: 110430580-110430580 on Build GRCh38
rs929262415 Chr.11: 110430177-110430177 on Build GRCh38
rs980036694 Chr.11: 110430595-110430595 on Build GRCh38
rs578015255 Chr.11: 110430144-110430144 on Build GRCh38
rs964399270 Chr.11: 110430527-110430527 on Build GRCh38
rs953235199 Chr.11: 110430180-110430180 on Build GRCh38
rs567432338 Chr.11: 110430526-110430526 on Build GRCh38
rs1010523184 Chr.11: 110430258-110430258 on Build GRCh38
rs944626520 Chr.11: 110430223-110430223 on Build GRCh38
rs930669789 Chr.11: 110430244-110430244 on Build GRCh38
rs529494476 Chr.11: 110430147-110430147 on Build GRCh38
rs781024881 Chr.11: 110430598-110430598 on Build GRCh38
rs756444105 Chr.11: 110430579-110430579 on Build GRCh38
rs867738540 Chr.11: 110430183-110430183 on Build GRCh38
rs577889448 Chr.11: 110430129-110430129 on Build GRCh38
rs1042659644 Chr.11: 110430573-110430573 on Build GRCh38
rs950539092 Chr.11: 110430338-110430338 on Build GRCh38
rs911774728 Chr.11: 110430194-110430194 on Build GRCh38
rs938186966 Chr.11: 110430249-110430249 on Build GRCh38
rs896700985 Chr.11: 110430287-110430287 on Build GRCh38
rs542788383 Chr.11: 110430136-110430136 on Build GRCh38
rs986401811 Chr.11: 110430188-110430188 on Build GRCh38
rs1021245659 Chr.11: 110430326-110430326 on Build GRCh38
rs923191817 Chr.11: 110430172-110430172 on Build GRCh38
rs1042198 Chr.11: 110430109-110430109 on Build GRCh38
rs985232400 Chr.11: 110430482-110430482 on Build GRCh38
rs888846947 Chr.11: 110430585-110430585 on Build GRCh38
rs553811356 Chr.11: 110430131-110430131 on Build GRCh38
rs368668727 Chr.11: 110430536-110430536 on Build GRCh38
rs764114299 Chr.11: 110430123-110430123 on Build GRCh38
rs187021002 Chr.11: 110430439-110430439 on Build GRCh38
rs751541149 Chr.11: 110430528-110430528 on Build GRCh38
rs551237718 Chr.11: 110430443-110430443 on Build GRCh38
rs904135220 Chr.11: 110430574-110430574 on Build GRCh38
rs983354427 Chr.11: 110430340-110430340 on Build GRCh38
rs1002810071 Chr.11: 110430119-110430119 on Build GRCh38
rs544984583 Chr.11: 110430130-110430130 on Build GRCh38
rs75300550 Chr.11: 110430524-110430524 on Build GRCh38
rs1017725993 Chr.11: 110430525-110430525 on Build GRCh38
rs753985474 Chr.11: 110430446-110430446 on Build GRCh38
rs972013428 Chr.11: 110430241-110430241 on Build GRCh38
rs143739712 Chr.11: 110430550-110430550 on Build GRCh38
rs572120843 Chr.11: 110430133-110430133 on Build GRCh38
rs1049128470 Chr.11: 110430246-110430246 on Build GRCh38
rs77165361 Chr.11: 110430432-110430432 on Build GRCh38
rs1042395336 Chr.11: 110430266-110430266 on Build GRCh38
rs942006842 Chr.11: 110430222-110430222 on Build GRCh38
rs545290208 Chr.11: 110430282-110430282 on Build GRCh38
rs1017364626 Chr.11: 110430314-110430314 on Build GRCh38
rs905316327 Chr.11: 110430248-110430248 on Build GRCh38
rs909109282 Chr.11: 110430356-110430356 on Build GRCh38
rs138066706 Chr.11: 110430255-110430255 on Build GRCh38
rs926694021 Chr.11: 110430502-110430502 on Build GRCh38
rs1000912910 Chr.11: 110430304-110430304 on Build GRCh38
rs943232252 Chr.11: 110430169-110430169 on Build GRCh38
rs533688780 Chr.11: 110430400-110430400 on Build GRCh38
rs930660219 Chr.11: 110430470-110430470 on Build GRCh38
rs909245058 Chr.11: 110430187-110430187 on Build GRCh38
rs544813521 Chr.11: 110430171-110430171 on Build GRCh38
rs924214964 Chr.11: 110430293-110430293 on Build GRCh38
rs923141103 Chr.11: 110430329-110430329 on Build GRCh38
rs983218890 Chr.11: 110430571-110430571 on Build GRCh38
rs1018749260 Chr.11: 110430431-110430431 on Build GRCh38
rs758220643 Chr.11: 110430523-110430523 on Build GRCh38
rs866188610 Chr.11: 110430387-110430387 on Build GRCh38

View Full Product Details