Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128905813-128906336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf45
Entrez Gene ID
219833
Gene Name
chromosome 11 open reading frame 45
Gene Aliases
-
UniGene
Hs.689860
Gene Chromosome Location
Chr.11: 128899565-128909520 on Build GRCh38
Transcripts
NM_001256088.1 NR_045767.1 XM_011542654.2 NM_000890.3 XM_011542810.2

Gene Symbol
KCNJ5
Entrez Gene ID
3762
Gene Name
potassium voltage-gated channel subfamily J member 5
Gene Aliases
CIR, GIRK4, KATP1, KIR3.4, LQT13
UniGene
Hs.444595
Gene Chromosome Location
Chr.11: 128891418-128921163 on Build GRCh38
Transcripts
NM_001256088.1 NR_045767.1 XM_011542654.2 NM_000890.3 XM_011542810.2

SNP Detail

SNP ID Chromosome Location
rs888447701 Chr.11: 128906217-128906217 on Build GRCh38
rs892623993 Chr.11: 128905843-128905843 on Build GRCh38
rs74647764 Chr.11: 128906185-128906185 on Build GRCh38
rs1024504642 Chr.11: 128905900-128905900 on Build GRCh38
rs1054437619 Chr.11: 128906120-128906120 on Build GRCh38
rs745769779 Chr.11: 128906000-128906000 on Build GRCh38
rs945374209 Chr.11: 128905990-128905990 on Build GRCh38
rs886327784 Chr.11: 128906148-128906148 on Build GRCh38
rs566859548 Chr.11: 128905951-128905951 on Build GRCh38
rs188709286 Chr.11: 128906292-128906292 on Build GRCh38
rs892684132 Chr.11: 128906130-128906130 on Build GRCh38
rs547111163 Chr.11: 128905918-128905918 on Build GRCh38
rs761841110 Chr.11: 128906310-128906310 on Build GRCh38
rs3867249 Chr.11: 128905868-128905868 on Build GRCh38
rs969815317 Chr.11: 128905910-128905910 on Build GRCh38
rs1020208800 Chr.11: 128905860-128905860 on Build GRCh38
rs945621941 Chr.11: 128906136-128906136 on Build GRCh38
rs539077318 Chr.11: 128905974-128905974 on Build GRCh38
rs911986091 Chr.11: 128906048-128906048 on Build GRCh38
rs760809482 Chr.11: 128906065-128906065 on Build GRCh38
rs1009771058 Chr.11: 128905859-128905859 on Build GRCh38
rs577844905 Chr.11: 128906140-128906140 on Build GRCh38
rs1039310557 Chr.11: 128906056-128906056 on Build GRCh38
rs536247492 Chr.11: 128905915-128905915 on Build GRCh38
rs183832550 Chr.11: 128906189-128906189 on Build GRCh38
rs1001301659 Chr.11: 128906193-128906193 on Build GRCh38
rs1051751870 Chr.11: 128906015-128906015 on Build GRCh38
rs531563811 Chr.11: 128906235-128906235 on Build GRCh38
rs1005586697 Chr.11: 128906260-128906260 on Build GRCh38
rs992642725 Chr.11: 128905980-128905980 on Build GRCh38
rs993174725 Chr.11: 128906305-128906308 on Build GRCh38
rs901218852 Chr.11: 128905862-128905862 on Build GRCh38
rs559620208 Chr.11: 128906113-128906113 on Build GRCh38
rs935706517 Chr.11: 128906085-128906085 on Build GRCh38
rs1023971866 Chr.11: 128906188-128906188 on Build GRCh38
rs1013850918 Chr.11: 128905894-128905894 on Build GRCh38
rs1041193932 Chr.11: 128906165-128906165 on Build GRCh38
rs560055989 Chr.11: 128906271-128906271 on Build GRCh38
rs950389526 Chr.11: 128905872-128905872 on Build GRCh38
rs961281880 Chr.11: 128906297-128906297 on Build GRCh38
rs559218787 Chr.11: 128906052-128906052 on Build GRCh38
rs961496813 Chr.11: 128905978-128905978 on Build GRCh38
rs958415167 Chr.11: 128905939-128905939 on Build GRCh38
rs575872660 Chr.11: 128906069-128906069 on Build GRCh38
rs994590953 Chr.11: 128906119-128906119 on Build GRCh38
rs994705570 Chr.11: 128905839-128905839 on Build GRCh38
rs914133644 Chr.11: 128905952-128905952 on Build GRCh38
rs926803650 Chr.11: 128905931-128905931 on Build GRCh38
rs539840608 Chr.11: 128906212-128906212 on Build GRCh38
rs545456656 Chr.11: 128906312-128906312 on Build GRCh38
rs538573551 Chr.11: 128906161-128906161 on Build GRCh38

View Full Product Details