Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1387699-1388197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNL2
Entrez Gene ID
81669
Gene Name
cyclin L2
Gene Aliases
ANIA-6B, CCNM, CCNS, HCLA-ISO, HLA-ISO, PCEE, SB138
UniGene
Hs.515704
Gene Chromosome Location
Chr.1: 1385711-1399338 on Build GRCh38
Transcripts
NM_001320153.1 NM_001320155.1 NM_030937.4 NR_135154.1 XM_011542216.2 XM_011542219.2 XM_011542221.2 XM_017002420.1 XM_017002421.1 XM_017002422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700475 c.1175C>A p.S392* Chr.1: 1387813-1387813 on Build GRCh38
894965 c.1133G>A p.R378Q Chr.1: 1387855-1387855 on Build GRCh38
2229773 c.1180T>C p.S394P Chr.1: 1387808-1387808 on Build GRCh38
2229778 c.1080G>T p.E360D Chr.1: 1387992-1387992 on Build GRCh38
6378885 c.1156A>G p.S386G Chr.1: 1387832-1387832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759638862 Chr.1: 1388173-1388173 on Build GRCh38
rs1038055838 Chr.1: 1387727-1387727 on Build GRCh38
rs536202089 Chr.1: 1387741-1387741 on Build GRCh38
rs758590729 Chr.1: 1387747-1387747 on Build GRCh38
rs773522715 Chr.1: 1388019-1388019 on Build GRCh38
rs140275617 Chr.1: 1387828-1387828 on Build GRCh38
rs35654872 Chr.1: 1387764-1387765 on Build GRCh38
rs980715854 Chr.1: 1387880-1387880 on Build GRCh38
rs573790257 Chr.1: 1387922-1387922 on Build GRCh38
rs762581855 Chr.1: 1387882-1387882 on Build GRCh38
rs762990432 Chr.1: 1387962-1387962 on Build GRCh38
rs567433441 Chr.1: 1387723-1387723 on Build GRCh38
rs776160735 Chr.1: 1387858-1387858 on Build GRCh38
rs147048179 Chr.1: 1387853-1387853 on Build GRCh38
rs373269685 Chr.1: 1388100-1388100 on Build GRCh38
rs757961175 Chr.1: 1387765-1387766 on Build GRCh38
rs1039515178 Chr.1: 1388115-1388115 on Build GRCh38
rs764289996 Chr.1: 1387829-1387829 on Build GRCh38
rs368076251 Chr.1: 1387779-1387779 on Build GRCh38
rs565206447 Chr.1: 1387983-1387983 on Build GRCh38
rs781311172 Chr.1: 1387782-1387782 on Build GRCh38
rs770263989 Chr.1: 1387865-1387865 on Build GRCh38
rs748453803 Chr.1: 1388012-1388012 on Build GRCh38
rs35912304 Chr.1: 1387919-1387920 on Build GRCh38
rs369616636 Chr.1: 1387925-1387925 on Build GRCh38
rs760069253 Chr.1: 1387811-1387811 on Build GRCh38
rs769342845 Chr.1: 1387993-1387993 on Build GRCh38
rs145080018 Chr.1: 1388013-1388013 on Build GRCh38
rs559094253 Chr.1: 1387862-1387862 on Build GRCh38
rs778753600 Chr.1: 1387845-1387845 on Build GRCh38
rs778612064 Chr.1: 1387766-1387766 on Build GRCh38
rs957488817 Chr.1: 1387789-1387789 on Build GRCh38
rs769352032 Chr.1: 1387931-1387931 on Build GRCh38
rs764087575 Chr.1: 1388059-1388059 on Build GRCh38
rs749832366 Chr.1: 1387937-1387937 on Build GRCh38
rs753450886 Chr.1: 1388085-1388085 on Build GRCh38
rs556747357 Chr.1: 1387914-1387914 on Build GRCh38
rs749259458 Chr.1: 1387762-1387762 on Build GRCh38
rs371968093 Chr.1: 1387846-1387846 on Build GRCh38
rs113128675 Chr.1: 1387765-1387765 on Build GRCh38
rs770887466 Chr.1: 1388042-1388042 on Build GRCh38
rs777076414 Chr.1: 1387803-1387803 on Build GRCh38
rs776571206 Chr.1: 1388112-1388112 on Build GRCh38
rs977067794 Chr.1: 1387896-1387896 on Build GRCh38
rs768298362 Chr.1: 1387891-1387891 on Build GRCh38
rs760993316 Chr.1: 1388036-1388036 on Build GRCh38
rs1047763162 Chr.1: 1388055-1388055 on Build GRCh38
rs141555700 Chr.1: 1387840-1387840 on Build GRCh38
rs200056802 Chr.1: 1387791-1387791 on Build GRCh38
rs750105467 Chr.1: 1387985-1387985 on Build GRCh38
rs867250840 Chr.1: 1387722-1387722 on Build GRCh38
rs976389405 Chr.1: 1387874-1387874 on Build GRCh38
rs775197672 Chr.1: 1387839-1387839 on Build GRCh38
rs765546031 Chr.1: 1387740-1387740 on Build GRCh38
rs372610777 Chr.1: 1388010-1388010 on Build GRCh38
rs546582270 Chr.1: 1388168-1388168 on Build GRCh38
rs544813154 Chr.1: 1387903-1387903 on Build GRCh38
rs375591952 Chr.1: 1387879-1387879 on Build GRCh38
rs764312826 Chr.1: 1387827-1387828 on Build GRCh38
rs780375734 Chr.1: 1387933-1387933 on Build GRCh38
rs757314768 Chr.1: 1387836-1387837 on Build GRCh38
rs779670113 Chr.1: 1387885-1387885 on Build GRCh38
rs757363125 Chr.1: 1387835-1387835 on Build GRCh38
rs143382296 Chr.1: 1387956-1387956 on Build GRCh38
rs774277728 Chr.1: 1388063-1388063 on Build GRCh38
rs539006374 Chr.1: 1387798-1387798 on Build GRCh38
rs754537134 Chr.1: 1388086-1388086 on Build GRCh38
rs530582050 Chr.1: 1387989-1387989 on Build GRCh38
rs758213227 Chr.1: 1387852-1387852 on Build GRCh38
rs960627867 Chr.1: 1388106-1388106 on Build GRCh38
rs763856887 Chr.1: 1387965-1387965 on Build GRCh38
rs765013725 Chr.1: 1387902-1387902 on Build GRCh38
rs751843737 Chr.1: 1387834-1387834 on Build GRCh38
rs371879342 Chr.1: 1387953-1387953 on Build GRCh38
rs751738515 Chr.1: 1387837-1387842 on Build GRCh38
rs894263639 Chr.1: 1387768-1387768 on Build GRCh38
rs751414472 Chr.1: 1387972-1387972 on Build GRCh38
rs941129285 Chr.1: 1387940-1387940 on Build GRCh38
rs756901180 Chr.1: 1387975-1387975 on Build GRCh38
rs771245776 Chr.1: 1388108-1388108 on Build GRCh38
rs567373829 Chr.1: 1388091-1388091 on Build GRCh38
rs781284424 Chr.1: 1388106-1388107 on Build GRCh38
rs369333066 Chr.1: 1388084-1388084 on Build GRCh38
rs1017810145 Chr.1: 1387847-1387847 on Build GRCh38
rs748682723 Chr.1: 1388099-1388099 on Build GRCh38
rs900691250 Chr.1: 1388135-1388135 on Build GRCh38
rs112675079 Chr.1: 1387780-1387780 on Build GRCh38
rs759840166 Chr.1: 1387736-1387736 on Build GRCh38
rs758083981 Chr.1: 1387913-1387913 on Build GRCh38
rs962774551 Chr.1: 1388169-1388169 on Build GRCh38
rs1026296230 Chr.1: 1387830-1387830 on Build GRCh38
rs868278772 Chr.1: 1387835-1387836 on Build GRCh38
rs763161176 Chr.1: 1387872-1387872 on Build GRCh38
rs770392095 Chr.1: 1387857-1387857 on Build GRCh38
rs776823901 Chr.1: 1388048-1388048 on Build GRCh38
rs747179595 Chr.1: 1388101-1388101 on Build GRCh38
rs761538965 Chr.1: 1388072-1388072 on Build GRCh38
rs774904348 Chr.1: 1387942-1387942 on Build GRCh38
rs775919840 Chr.1: 1387866-1387866 on Build GRCh38
rs371077547 Chr.1: 1388114-1388114 on Build GRCh38
rs774673746 Chr.1: 1387878-1387878 on Build GRCh38
rs80254624 Chr.1: 1387841-1387841 on Build GRCh38
rs148355846 Chr.1: 1387986-1387986 on Build GRCh38
rs767330903 Chr.1: 1388075-1388075 on Build GRCh38
rs768869908 Chr.1: 1387941-1387941 on Build GRCh38
rs559430048 Chr.1: 1387928-1387928 on Build GRCh38
rs142455256 Chr.1: 1388027-1388027 on Build GRCh38
rs769240244 Chr.1: 1387792-1387792 on Build GRCh38
rs746690488 Chr.1: 1387855-1387855 on Build GRCh38
rs966939013 Chr.1: 1388165-1388165 on Build GRCh38
rs765606459 Chr.1: 1387819-1387819 on Build GRCh38
rs764541020 Chr.1: 1387877-1387877 on Build GRCh38
rs762110584 Chr.1: 1387943-1387943 on Build GRCh38
rs761046066 Chr.1: 1388136-1388136 on Build GRCh38
rs751648968 Chr.1: 1387754-1387754 on Build GRCh38
rs777724917 Chr.1: 1387763-1387764 on Build GRCh38
rs780081134 Chr.1: 1387760-1387760 on Build GRCh38
rs780669107 Chr.1: 1387927-1387927 on Build GRCh38
rs369827861 Chr.1: 1387987-1387987 on Build GRCh38
rs779653949 Chr.1: 1387988-1387988 on Build GRCh38
rs374147211 Chr.1: 1387774-1387774 on Build GRCh38
rs900051961 Chr.1: 1387732-1387732 on Build GRCh38
rs552949710 Chr.1: 1387748-1387748 on Build GRCh38
rs376876085 Chr.1: 1388083-1388083 on Build GRCh38
rs375538019 Chr.1: 1387961-1387961 on Build GRCh38
rs762788071 Chr.1: 1388052-1388052 on Build GRCh38
rs772487356 Chr.1: 1387951-1387951 on Build GRCh38
rs530177506 Chr.1: 1388144-1388144 on Build GRCh38
rs752698205 Chr.1: 1387906-1387906 on Build GRCh38
rs760610228 Chr.1: 1387955-1387955 on Build GRCh38
rs575779870 Chr.1: 1387887-1387887 on Build GRCh38
rs377592048 Chr.1: 1387756-1387756 on Build GRCh38

View Full Product Details