Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9916512-9917054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC2A
Entrez Gene ID
387836
Gene Name
C-type lectin domain family 2 member A
Gene Aliases
INPE5792, KACL, PILAR, UNQ5792
UniGene
Hs.527665
Gene Chromosome Location
Chr.12: 9882733-9932430 on Build GRCh38
Transcripts
NM_001130711.1 NM_207375.2 XM_011520654.2 XM_011520656.2 XM_011520657.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361494 c.404A>G p.N135S Chr.12: 9916706-9916706 on Build GRCh38
3416365 c.310T>G p.F104V Chr.12: 9916800-9916800 on Build GRCh38
467639 c.407G>A p.G136D Chr.12: 9916703-9916703 on Build GRCh38
4882331 c.400T>A p.F134I Chr.12: 9916710-9916710 on Build GRCh38
5641087 c.370G>A p.D124N Chr.12: 9916740-9916740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138190578 Chr.12: 9916762-9916762 on Build GRCh38
rs575317127 Chr.12: 9916653-9916653 on Build GRCh38
rs34974509 Chr.12: 9916818-9916819 on Build GRCh38
rs1019869655 Chr.12: 9916619-9916619 on Build GRCh38
rs1057220182 Chr.12: 9916550-9916550 on Build GRCh38
rs895948184 Chr.12: 9916560-9916560 on Build GRCh38
rs113041524 Chr.12: 9916786-9916786 on Build GRCh38
rs2166809 Chr.12: 9916987-9916987 on Build GRCh38
rs762786447 Chr.12: 9916657-9916657 on Build GRCh38
rs946626877 Chr.12: 9916687-9916687 on Build GRCh38
rs937600329 Chr.12: 9916756-9916756 on Build GRCh38
rs955480280 Chr.12: 9916647-9916647 on Build GRCh38
rs562093449 Chr.12: 9916870-9916870 on Build GRCh38
rs753662157 Chr.12: 9916929-9916929 on Build GRCh38
rs141736643 Chr.12: 9916598-9916604 on Build GRCh38
rs766541724 Chr.12: 9916704-9916704 on Build GRCh38
rs199613466 Chr.12: 9916658-9916658 on Build GRCh38
rs1038561344 Chr.12: 9916873-9916873 on Build GRCh38
rs190255533 Chr.12: 9916960-9916960 on Build GRCh38
rs769366198 Chr.12: 9916801-9916801 on Build GRCh38
rs763955516 Chr.12: 9916784-9916784 on Build GRCh38
rs995157403 Chr.12: 9916591-9916591 on Build GRCh38
rs1010054363 Chr.12: 9917012-9917012 on Build GRCh38
rs535799701 Chr.12: 9916774-9916774 on Build GRCh38
rs368400669 Chr.12: 9916711-9916711 on Build GRCh38
rs150435467 Chr.12: 9916537-9916537 on Build GRCh38
rs911189194 Chr.12: 9916686-9916686 on Build GRCh38
rs149571714 Chr.12: 9917001-9917001 on Build GRCh38
rs750510144 Chr.12: 9916699-9916699 on Build GRCh38
rs994335295 Chr.12: 9916892-9916892 on Build GRCh38
rs538604121 Chr.12: 9916951-9916951 on Build GRCh38
rs563888001 Chr.12: 9916680-9916680 on Build GRCh38
rs524994 Chr.12: 9916567-9916567 on Build GRCh38
rs367648527 Chr.12: 9916781-9916781 on Build GRCh38
rs1057342933 Chr.12: 9916760-9916760 on Build GRCh38
rs896826563 Chr.12: 9916576-9916576 on Build GRCh38
rs370767849 Chr.12: 9916675-9916675 on Build GRCh38
rs979416371 Chr.12: 9916997-9916997 on Build GRCh38
rs1029724779 Chr.12: 9916628-9916628 on Build GRCh38
rs767613949 Chr.12: 9916688-9916688 on Build GRCh38
rs766760257 Chr.12: 9916684-9916684 on Build GRCh38
rs759248673 Chr.12: 9916584-9916584 on Build GRCh38
rs544001808 Chr.12: 9916678-9916678 on Build GRCh38
rs566443583 Chr.12: 9917024-9917024 on Build GRCh38
rs978727842 Chr.12: 9916742-9916742 on Build GRCh38
rs1029776911 Chr.12: 9916928-9916928 on Build GRCh38
rs200701997 Chr.12: 9916727-9916727 on Build GRCh38
rs370351914 Chr.12: 9916718-9916718 on Build GRCh38
rs12306080 Chr.12: 9917017-9917017 on Build GRCh38
rs968045109 Chr.12: 9916957-9916957 on Build GRCh38
rs1017061407 Chr.12: 9916564-9916564 on Build GRCh38
rs547030492 Chr.12: 9916596-9916596 on Build GRCh38
rs113499307 Chr.12: 9916946-9916946 on Build GRCh38
rs752228885 Chr.12: 9916608-9916608 on Build GRCh38
rs887412391 Chr.12: 9916818-9916818 on Build GRCh38
rs995207078 Chr.12: 9916872-9916872 on Build GRCh38
rs972861405 Chr.12: 9916595-9916595 on Build GRCh38
rs927582082 Chr.12: 9916745-9916745 on Build GRCh38
rs526680 Chr.12: 9916703-9916703 on Build GRCh38
rs753238164 Chr.12: 9916724-9916724 on Build GRCh38
rs531019592 Chr.12: 9916876-9916876 on Build GRCh38
rs181118308 Chr.12: 9916996-9916996 on Build GRCh38
rs779321490 Chr.12: 9916720-9916720 on Build GRCh38
rs185978029 Chr.12: 9916562-9916562 on Build GRCh38
rs368715067 Chr.12: 9916829-9916829 on Build GRCh38
rs560101406 Chr.12: 9916759-9916759 on Build GRCh38

View Full Product Details