Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9971617-9972118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC12A
Entrez Gene ID
160364
Gene Name
C-type lectin domain family 12 member A
Gene Aliases
CD371, CLL-1, CLL1, DCAL-2, MICL
UniGene
Hs.190519
Gene Chromosome Location
Chr.12: 9951316-9995209 on Build GRCh38
Transcripts
NM_001207010.1 NM_001300730.1 NM_138337.5 NM_201623.3 XM_005253324.2 XM_006719033.3 XM_006719035.3 XM_006719036.3 XM_011520570.1 XM_011520571.2 XM_011520573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1298811 c.63C>T p.I21I Chr.12: 9971659-9971659 on Build GRCh38
4672714 c.85_86insA p.A31fs*53 Chr.12: 9971681-9971682 on Build GRCh38
1298810 c.93C>T p.I31I Chr.12: 9971659-9971659 on Build GRCh38
1248404 c.80T>G p.F27C Chr.12: 9971676-9971676 on Build GRCh38
1248403 c.110T>G p.F37C Chr.12: 9971676-9971676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141711354 Chr.12: 9971871-9971871 on Build GRCh38
rs766312863 Chr.12: 9971651-9971652 on Build GRCh38
rs2984956 Chr.12: 9971737-9971737 on Build GRCh38
rs201144260 Chr.12: 9972060-9972060 on Build GRCh38
rs368532397 Chr.12: 9971723-9971723 on Build GRCh38
rs755301368 Chr.12: 9971673-9971679 on Build GRCh38
rs1016710408 Chr.12: 9971855-9971855 on Build GRCh38
rs962549545 Chr.12: 9972071-9972071 on Build GRCh38
rs149569771 Chr.12: 9972082-9972082 on Build GRCh38
rs763749828 Chr.12: 9971668-9971668 on Build GRCh38
rs759377720 Chr.12: 9972057-9972064 on Build GRCh38
rs892107874 Chr.12: 9971845-9971845 on Build GRCh38
rs921195644 Chr.12: 9971888-9971888 on Build GRCh38
rs986469818 Chr.12: 9971921-9971921 on Build GRCh38
rs779038063 Chr.12: 9971804-9971805 on Build GRCh38
rs144231894 Chr.12: 9972086-9972086 on Build GRCh38
rs907450633 Chr.12: 9972008-9972008 on Build GRCh38
rs1047713754 Chr.12: 9972046-9972046 on Build GRCh38
rs762396359 Chr.12: 9971666-9971666 on Build GRCh38
rs145705192 Chr.12: 9971929-9971929 on Build GRCh38
rs201875211 Chr.12: 9972058-9972058 on Build GRCh38
rs67786371 Chr.12: 9972053-9972053 on Build GRCh38
rs755892723 Chr.12: 9971739-9971739 on Build GRCh38
rs693494 Chr.12: 9972076-9972076 on Build GRCh38
rs936370239 Chr.12: 9971826-9971826 on Build GRCh38
rs942542141 Chr.12: 9971768-9971768 on Build GRCh38
rs755404795 Chr.12: 9972056-9972057 on Build GRCh38
rs749556690 Chr.12: 9972043-9972048 on Build GRCh38
rs764635204 Chr.12: 9971662-9971662 on Build GRCh38
rs779966756 Chr.12: 9971686-9971686 on Build GRCh38
rs10550334 Chr.12: 9972057-9972062 on Build GRCh38
rs60417066 Chr.12: 9972070-9972071 on Build GRCh38
rs768529836 Chr.12: 9972048-9972049 on Build GRCh38
rs773764131 Chr.12: 9971699-9971699 on Build GRCh38
rs189179326 Chr.12: 9971890-9971890 on Build GRCh38
rs185524074 Chr.12: 9971712-9971712 on Build GRCh38
rs138302751 Chr.12: 9972006-9972006 on Build GRCh38
rs533589131 Chr.12: 9971665-9971665 on Build GRCh38
rs373897817 Chr.12: 9971707-9971707 on Build GRCh38
rs761510038 Chr.12: 9971722-9971722 on Build GRCh38
rs550964283 Chr.12: 9971816-9971816 on Build GRCh38
rs531336470 Chr.12: 9971654-9971654 on Build GRCh38
rs141326355 Chr.12: 9971657-9971657 on Build GRCh38
rs779212116 Chr.12: 9971694-9971694 on Build GRCh38
rs143296013 Chr.12: 9971669-9971669 on Build GRCh38
rs80107756 Chr.12: 9972069-9972071 on Build GRCh38
rs774508497 Chr.12: 9972053-9972059 on Build GRCh38
rs762464486 Chr.12: 9971708-9971708 on Build GRCh38
rs71441781 Chr.12: 9972056-9972056 on Build GRCh38
rs1026316415 Chr.12: 9971803-9971803 on Build GRCh38
rs567155810 Chr.12: 9971698-9971698 on Build GRCh38
rs371255349 Chr.12: 9971696-9971696 on Build GRCh38
rs975336076 Chr.12: 9971874-9971874 on Build GRCh38
rs974132527 Chr.12: 9971966-9971966 on Build GRCh38
rs1025654662 Chr.12: 9971990-9971990 on Build GRCh38
rs912393609 Chr.12: 9971767-9971767 on Build GRCh38
rs936442317 Chr.12: 9972070-9972070 on Build GRCh38
rs869065604 Chr.12: 9972055-9972057 on Build GRCh38
rs987156865 Chr.12: 9972001-9972001 on Build GRCh38
rs781030619 Chr.12: 9971679-9971679 on Build GRCh38
rs796297333 Chr.12: 9972053-9972057 on Build GRCh38
rs201816371 Chr.12: 9971660-9971660 on Build GRCh38
rs200619603 Chr.12: 9972043-9972044 on Build GRCh38
rs756976989 Chr.12: 9971673-9971673 on Build GRCh38
rs948913102 Chr.12: 9972000-9972000 on Build GRCh38
rs761171835 Chr.12: 9972053-9972057 on Build GRCh38
rs751991188 Chr.12: 9971650-9971650 on Build GRCh38
rs756124445 Chr.12: 9971682-9971682 on Build GRCh38
rs867656488 Chr.12: 9971640-9971640 on Build GRCh38
rs530928031 Chr.12: 9971782-9971782 on Build GRCh38
rs200050138 Chr.12: 9972059-9972059 on Build GRCh38
rs1010906846 Chr.12: 9971866-9971866 on Build GRCh38
rs903594815 Chr.12: 9971821-9971821 on Build GRCh38
rs751206019 Chr.12: 9971714-9971714 on Build GRCh38
rs1016321916 Chr.12: 9972065-9972065 on Build GRCh38
rs531225337 Chr.12: 9972075-9972075 on Build GRCh38
rs951381367 Chr.12: 9971994-9971994 on Build GRCh38
rs758905699 Chr.12: 9972063-9972063 on Build GRCh38
rs1001706105 Chr.12: 9971895-9971895 on Build GRCh38
rs757707883 Chr.12: 9971893-9971893 on Build GRCh38
rs538314105 Chr.12: 9971838-9971838 on Build GRCh38
rs201936514 Chr.12: 9972062-9972062 on Build GRCh38
rs756506623 Chr.12: 9971756-9971756 on Build GRCh38
rs866380852 Chr.12: 9971687-9971687 on Build GRCh38
rs138074498 Chr.12: 9972057-9972061 on Build GRCh38
rs762212618 Chr.12: 9971650-9971650 on Build GRCh38
rs1003164975 Chr.12: 9972017-9972017 on Build GRCh38
rs771433619 Chr.12: 9971704-9971704 on Build GRCh38
rs767277542 Chr.12: 9971736-9971736 on Build GRCh38
rs770759055 Chr.12: 9972061-9972061 on Build GRCh38
rs77858080 Chr.12: 9972061-9972062 on Build GRCh38
rs1797526 Chr.12: 9971934-9971934 on Build GRCh38
rs750345792 Chr.12: 9971681-9971681 on Build GRCh38
rs942552284 Chr.12: 9972024-9972024 on Build GRCh38
rs2961542 Chr.12: 9971774-9971774 on Build GRCh38
rs769550186 Chr.12: 9972058-9972063 on Build GRCh38
rs148336258 Chr.12: 9971688-9971688 on Build GRCh38
rs368369689 Chr.12: 9971838-9971838 on Build GRCh38
rs1054227421 Chr.12: 9971955-9971955 on Build GRCh38

View Full Product Details