Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 29223575-29224083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAR2
Entrez Gene ID
55711
Gene Name
fatty acyl-CoA reductase 2
Gene Aliases
HEL-S-81, MLSTD1, SDR10E2
UniGene
Hs.684482
Gene Chromosome Location
Chr.12: 29149003-29335616 on Build GRCh38
Transcripts
NM_001271783.1 NM_001271784.1 NM_018099.4 XM_011520747.2 XM_011520748.2 XM_017019624.1 XM_017019625.1

SNP Detail

SNP ID Chromosome Location
rs765327513 Chr.12: 29223767-29223767 on Build GRCh38
rs910826319 Chr.12: 29223793-29223793 on Build GRCh38
rs761945407 Chr.12: 29223858-29223858 on Build GRCh38
rs917229696 Chr.12: 29223979-29223979 on Build GRCh38
rs577521989 Chr.12: 29223679-29223679 on Build GRCh38
rs936172625 Chr.12: 29223713-29223713 on Build GRCh38
rs895887075 Chr.12: 29224006-29224006 on Build GRCh38
rs777367545 Chr.12: 29223889-29223889 on Build GRCh38
rs993398786 Chr.12: 29223832-29223832 on Build GRCh38
rs991436829 Chr.12: 29223601-29223601 on Build GRCh38
rs191589209 Chr.12: 29223849-29223849 on Build GRCh38
rs192564800 Chr.12: 29223608-29223608 on Build GRCh38
rs958764761 Chr.12: 29223940-29223940 on Build GRCh38
rs1040042456 Chr.12: 29223812-29223812 on Build GRCh38
rs1057252777 Chr.12: 29224001-29224001 on Build GRCh38
rs527566632 Chr.12: 29223732-29223732 on Build GRCh38
rs984618738 Chr.12: 29223690-29223690 on Build GRCh38
rs922936076 Chr.12: 29223746-29223746 on Build GRCh38
rs998949185 Chr.12: 29223911-29223911 on Build GRCh38
rs186785951 Chr.12: 29223847-29223847 on Build GRCh38
rs950308232 Chr.12: 29223628-29223628 on Build GRCh38
rs752680250 Chr.12: 29223790-29223790 on Build GRCh38
rs145015760 Chr.12: 29223874-29223874 on Build GRCh38
rs977664793 Chr.12: 29224017-29224017 on Build GRCh38
rs776656926 Chr.12: 29223614-29223614 on Build GRCh38
rs901522527 Chr.12: 29223829-29223829 on Build GRCh38
rs933034026 Chr.12: 29223782-29223782 on Build GRCh38
rs1013326572 Chr.12: 29223645-29223645 on Build GRCh38
rs570443516 Chr.12: 29223701-29223701 on Build GRCh38
rs960036333 Chr.12: 29223638-29223638 on Build GRCh38
rs184191826 Chr.12: 29223691-29223691 on Build GRCh38
rs991553137 Chr.12: 29223953-29223953 on Build GRCh38
rs971747028 Chr.12: 29223717-29223717 on Build GRCh38
rs1054671491 Chr.12: 29223757-29223757 on Build GRCh38
rs971814558 Chr.12: 29223981-29223981 on Build GRCh38
rs1033502865 Chr.12: 29223928-29223928 on Build GRCh38
rs544926554 Chr.12: 29223689-29223689 on Build GRCh38

View Full Product Details