Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56642292-56642833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5B
Entrez Gene ID
506
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases
ATPMB, ATPSB, HEL-S-271
UniGene
Hs.406510
Gene Chromosome Location
Chr.12: 56638175-56646068 on Build GRCh38
Transcripts
NM_001686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3376318 c.898G>C p.D300H Chr.12: 56642726-56642726 on Build GRCh38
4633702 c.854C>A p.T285N Chr.12: 56642770-56642770 on Build GRCh38
7322292 c.952-1G>A p.? Chr.12: 56642581-56642581 on Build GRCh38
4513387 c.936C>T p.T312T Chr.12: 56642688-56642688 on Build GRCh38
7305352 c.839C>T p.A280V Chr.12: 56642785-56642785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767468553 Chr.12: 56642470-56642470 on Build GRCh38
rs1044985091 Chr.12: 56642793-56642793 on Build GRCh38
rs749480463 Chr.12: 56642627-56642627 on Build GRCh38
rs750895114 Chr.12: 56642727-56642727 on Build GRCh38
rs780433436 Chr.12: 56642770-56642770 on Build GRCh38
rs200004957 Chr.12: 56642443-56642443 on Build GRCh38
rs369008512 Chr.12: 56642662-56642662 on Build GRCh38
rs749568756 Chr.12: 56642784-56642784 on Build GRCh38
rs995281650 Chr.12: 56642340-56642340 on Build GRCh38
rs778366547 Chr.12: 56642586-56642586 on Build GRCh38
rs774460776 Chr.12: 56642463-56642463 on Build GRCh38
rs999118788 Chr.12: 56642394-56642394 on Build GRCh38
rs201302572 Chr.12: 56642625-56642625 on Build GRCh38
rs562906295 Chr.12: 56642441-56642441 on Build GRCh38
rs770139894 Chr.12: 56642604-56642604 on Build GRCh38
rs1028097410 Chr.12: 56642426-56642426 on Build GRCh38
rs752204083 Chr.12: 56642716-56642716 on Build GRCh38
rs371608577 Chr.12: 56642605-56642605 on Build GRCh38
rs760397757 Chr.12: 56642652-56642652 on Build GRCh38
rs895828024 Chr.12: 56642676-56642676 on Build GRCh38
rs189451411 Chr.12: 56642630-56642630 on Build GRCh38
rs542763979 Chr.12: 56642699-56642699 on Build GRCh38
rs11542644 Chr.12: 56642680-56642680 on Build GRCh38
rs140513120 Chr.12: 56642718-56642718 on Build GRCh38
rs200319393 Chr.12: 56642744-56642744 on Build GRCh38
rs368222309 Chr.12: 56642422-56642422 on Build GRCh38
rs916031834 Chr.12: 56642756-56642756 on Build GRCh38
rs957073878 Chr.12: 56642414-56642414 on Build GRCh38
rs754502933 Chr.12: 56642545-56642545 on Build GRCh38
rs532478420 Chr.12: 56642321-56642321 on Build GRCh38
rs778003361 Chr.12: 56642723-56642723 on Build GRCh38
rs781669179 Chr.12: 56642595-56642595 on Build GRCh38
rs764295123 Chr.12: 56642757-56642758 on Build GRCh38
rs201322205 Chr.12: 56642742-56642742 on Build GRCh38
rs201980085 Chr.12: 56642587-56642587 on Build GRCh38
rs373472201 Chr.12: 56642634-56642634 on Build GRCh38
rs760801755 Chr.12: 56642663-56642664 on Build GRCh38
rs544634277 Chr.12: 56642356-56642356 on Build GRCh38
rs769905198 Chr.12: 56642428-56642428 on Build GRCh38
rs775583034 Chr.12: 56642425-56642425 on Build GRCh38
rs903410364 Chr.12: 56642369-56642369 on Build GRCh38
rs746162896 Chr.12: 56642425-56642425 on Build GRCh38
rs779338647 Chr.12: 56642631-56642631 on Build GRCh38
rs955187649 Chr.12: 56642569-56642569 on Build GRCh38
rs755340059 Chr.12: 56642562-56642562 on Build GRCh38
rs765948611 Chr.12: 56642486-56642486 on Build GRCh38
rs1014415079 Chr.12: 56642353-56642353 on Build GRCh38
rs750171418 Chr.12: 56642471-56642471 on Build GRCh38
rs764789281 Chr.12: 56642700-56642700 on Build GRCh38
rs375233620 Chr.12: 56642626-56642626 on Build GRCh38
rs762958204 Chr.12: 56642433-56642433 on Build GRCh38
rs773130792 Chr.12: 56642635-56642635 on Build GRCh38
rs1042001 Chr.12: 56642804-56642804 on Build GRCh38
rs761809872 Chr.12: 56642464-56642464 on Build GRCh38
rs775715405 Chr.12: 56642432-56642432 on Build GRCh38
rs755858646 Chr.12: 56642474-56642474 on Build GRCh38
rs758934230 Chr.12: 56642717-56642717 on Build GRCh38
rs753476589 Chr.12: 56642665-56642665 on Build GRCh38
rs369343050 Chr.12: 56642542-56642542 on Build GRCh38
rs867442552 Chr.12: 56642753-56642753 on Build GRCh38
rs772276020 Chr.12: 56642424-56642425 on Build GRCh38
rs747477525 Chr.12: 56642411-56642411 on Build GRCh38
rs1042002 Chr.12: 56642512-56642512 on Build GRCh38
rs752112335 Chr.12: 56642588-56642588 on Build GRCh38
rs778262428 Chr.12: 56642409-56642409 on Build GRCh38
rs867106580 Chr.12: 56642447-56642447 on Build GRCh38
rs746223095 Chr.12: 56642599-56642599 on Build GRCh38
rs776841680 Chr.12: 56642424-56642424 on Build GRCh38

View Full Product Details