Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89351924-89352419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP6
Entrez Gene ID
1848
Gene Name
dual specificity phosphatase 6
Gene Aliases
HH19, MKP3, PYST1
UniGene
Hs.298654
Gene Chromosome Location
Chr.12: 89347825-89352859 on Build GRCh38
Transcripts
NM_001946.3 NM_022652.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
431954 c.73G>A p.E25K Chr.12: 89351967-89351967 on Build GRCh38
1995021 c.49A>T p.S17C Chr.12: 89351991-89351991 on Build GRCh38
4883397 c.3G>A p.M1I Chr.12: 89352037-89352037 on Build GRCh38
1364742 c.93C>A p.N31K Chr.12: 89351947-89351947 on Build GRCh38
3376396 c.66G>A p.W22* Chr.12: 89351974-89351974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189094409 Chr.12: 89352379-89352379 on Build GRCh38
rs967592324 Chr.12: 89352082-89352082 on Build GRCh38
rs1014088065 Chr.12: 89351978-89351978 on Build GRCh38
rs955003812 Chr.12: 89352017-89352017 on Build GRCh38
rs764109153 Chr.12: 89352076-89352076 on Build GRCh38
rs543873089 Chr.12: 89352331-89352331 on Build GRCh38
rs966892763 Chr.12: 89352271-89352271 on Build GRCh38
rs538971597 Chr.12: 89352192-89352192 on Build GRCh38
rs1027559997 Chr.12: 89352238-89352238 on Build GRCh38
rs148576220 Chr.12: 89351989-89351989 on Build GRCh38
rs201858156 Chr.12: 89352033-89352033 on Build GRCh38
rs753875746 Chr.12: 89352077-89352077 on Build GRCh38
rs949806515 Chr.12: 89352344-89352344 on Build GRCh38
rs1015687828 Chr.12: 89352245-89352245 on Build GRCh38
rs991752060 Chr.12: 89352305-89352305 on Build GRCh38
rs574573165 Chr.12: 89352266-89352266 on Build GRCh38
rs978018463 Chr.12: 89352090-89352090 on Build GRCh38
rs183833024 Chr.12: 89352332-89352332 on Build GRCh38
rs926207447 Chr.12: 89352313-89352313 on Build GRCh38
rs536864459 Chr.12: 89352151-89352151 on Build GRCh38
rs936273181 Chr.12: 89352119-89352119 on Build GRCh38
rs1045872811 Chr.12: 89352351-89352351 on Build GRCh38
rs952672259 Chr.12: 89352043-89352043 on Build GRCh38
rs772457203 Chr.12: 89352273-89352273 on Build GRCh38
rs141049010 Chr.12: 89351946-89351946 on Build GRCh38
rs961448955 Chr.12: 89352270-89352270 on Build GRCh38
rs79053845 Chr.12: 89352355-89352355 on Build GRCh38
rs766301392 Chr.12: 89352016-89352016 on Build GRCh38
rs756143915 Chr.12: 89352008-89352008 on Build GRCh38
rs758485593 Chr.12: 89351996-89351996 on Build GRCh38
rs765005164 Chr.12: 89352392-89352392 on Build GRCh38
rs567001636 Chr.12: 89352350-89352351 on Build GRCh38
rs760935219 Chr.12: 89352368-89352368 on Build GRCh38
rs751604114 Chr.12: 89352072-89352072 on Build GRCh38
rs752643011 Chr.12: 89352005-89352005 on Build GRCh38
rs988403057 Chr.12: 89352334-89352334 on Build GRCh38
rs201138185 Chr.12: 89352019-89352019 on Build GRCh38
rs745473293 Chr.12: 89352088-89352088 on Build GRCh38
rs985661437 Chr.12: 89352132-89352132 on Build GRCh38
rs1020601976 Chr.12: 89352269-89352269 on Build GRCh38
rs547284305 Chr.12: 89352028-89352028 on Build GRCh38
rs865817237 Chr.12: 89352217-89352217 on Build GRCh38
rs940105077 Chr.12: 89352197-89352197 on Build GRCh38
rs780038667 Chr.12: 89351997-89351997 on Build GRCh38
rs765239006 Chr.12: 89352286-89352287 on Build GRCh38
rs562832283 Chr.12: 89351965-89351965 on Build GRCh38
rs778602279 Chr.12: 89352083-89352083 on Build GRCh38
rs888482227 Chr.12: 89352242-89352242 on Build GRCh38
rs1027986241 Chr.12: 89352015-89352015 on Build GRCh38
rs935662574 Chr.12: 89352180-89352180 on Build GRCh38
rs761953520 Chr.12: 89351968-89351968 on Build GRCh38
rs776032408 Chr.12: 89351953-89351953 on Build GRCh38
rs559157035 Chr.12: 89352206-89352206 on Build GRCh38
rs950062405 Chr.12: 89352179-89352179 on Build GRCh38
rs370257627 Chr.12: 89352181-89352181 on Build GRCh38
rs774626250 Chr.12: 89352130-89352130 on Build GRCh38
rs777192674 Chr.12: 89352048-89352048 on Build GRCh38
rs761181689 Chr.12: 89352035-89352035 on Build GRCh38
rs192081420 Chr.12: 89351952-89351952 on Build GRCh38
rs985712229 Chr.12: 89352283-89352283 on Build GRCh38
rs116765964 Chr.12: 89352126-89352126 on Build GRCh38
rs1029805409 Chr.12: 89352276-89352276 on Build GRCh38
rs570195466 Chr.12: 89352187-89352187 on Build GRCh38
rs151076733 Chr.12: 89352226-89352226 on Build GRCh38
rs757085812 Chr.12: 89352079-89352079 on Build GRCh38
rs1038644380 Chr.12: 89352202-89352202 on Build GRCh38
rs749828471 Chr.12: 89352116-89352116 on Build GRCh38
rs769997603 Chr.12: 89352047-89352047 on Build GRCh38
rs1049045138 Chr.12: 89352370-89352370 on Build GRCh38
rs765444393 Chr.12: 89351971-89351971 on Build GRCh38
rs778684685 Chr.12: 89352020-89352020 on Build GRCh38
rs554674631 Chr.12: 89352243-89352243 on Build GRCh38
rs749090170 Chr.12: 89352013-89352013 on Build GRCh38
rs577697011 Chr.12: 89352196-89352196 on Build GRCh38
rs575824258 Chr.12: 89352250-89352250 on Build GRCh38
rs976906296 Chr.12: 89352104-89352104 on Build GRCh38
rs775794238 Chr.12: 89352342-89352342 on Build GRCh38
rs922851269 Chr.12: 89352111-89352111 on Build GRCh38
rs201126929 Chr.12: 89352060-89352060 on Build GRCh38
rs142927078 Chr.12: 89352209-89352209 on Build GRCh38
rs527240303 Chr.12: 89352073-89352073 on Build GRCh38
rs553295680 Chr.12: 89352029-89352029 on Build GRCh38
rs766434885 Chr.12: 89352064-89352064 on Build GRCh38
rs375201922 Chr.12: 89352010-89352010 on Build GRCh38
rs763117998 Chr.12: 89352062-89352062 on Build GRCh38
rs764390682 Chr.12: 89351961-89351961 on Build GRCh38
rs981877883 Chr.12: 89352343-89352343 on Build GRCh38
rs957491339 Chr.12: 89352287-89352287 on Build GRCh38
rs777653842 Chr.12: 89352012-89352012 on Build GRCh38
rs796691930 Chr.12: 89352351-89352351 on Build GRCh38
rs767848655 Chr.12: 89352039-89352040 on Build GRCh38
rs547561739 Chr.12: 89352099-89352099 on Build GRCh38
rs199734366 Chr.12: 89352054-89352054 on Build GRCh38
rs905793653 Chr.12: 89352218-89352218 on Build GRCh38
rs760626772 Chr.12: 89352281-89352281 on Build GRCh38

View Full Product Details