Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11076468-11076968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC10
Entrez Gene ID
5394
Gene Name
exosome component 10
Gene Aliases
PM-Scl, PM/Scl-100, PMSCL, PMSCL2, RRP6, Rrp6p, p2, p3, p4
UniGene
Hs.632368
Gene Chromosome Location
Chr.1: 11066613-11099910 on Build GRCh38
Transcripts
NM_001001998.2 NM_002685.3 XM_005263475.2 XM_017001494.1 XM_017001495.1 XM_017001496.1 XM_017001497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735250 c.1906A>G p.S636G Chr.1: 11076922-11076922 on Build GRCh38
7133347 c.1974C>T p.I658I Chr.1: 11076854-11076854 on Build GRCh38
6657291 c.1905G>A p.A635A Chr.1: 11076923-11076923 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs186263031 Chr.1: 11076584-11076584 on Build GRCh38
rs983853905 Chr.1: 11076606-11076606 on Build GRCh38
rs370147496 Chr.1: 11076876-11076876 on Build GRCh38
rs374057128 Chr.1: 11076853-11076853 on Build GRCh38
rs998124832 Chr.1: 11076897-11076897 on Build GRCh38
rs1007220508 Chr.1: 11076530-11076530 on Build GRCh38
rs769202354 Chr.1: 11076855-11076855 on Build GRCh38
rs377142383 Chr.1: 11076809-11076809 on Build GRCh38
rs765298874 Chr.1: 11076920-11076920 on Build GRCh38
rs183257312 Chr.1: 11076490-11076490 on Build GRCh38
rs148055003 Chr.1: 11076550-11076550 on Build GRCh38
rs72870088 Chr.1: 11076758-11076758 on Build GRCh38
rs777489759 Chr.1: 11076848-11076848 on Build GRCh38
rs149349714 Chr.1: 11076919-11076919 on Build GRCh38
rs909114223 Chr.1: 11076522-11076522 on Build GRCh38
rs199670680 Chr.1: 11076883-11076883 on Build GRCh38
rs191575932 Chr.1: 11076589-11076589 on Build GRCh38
rs935509267 Chr.1: 11076736-11076736 on Build GRCh38
rs753818672 Chr.1: 11076791-11076791 on Build GRCh38
rs560089561 Chr.1: 11076684-11076684 on Build GRCh38
rs944713870 Chr.1: 11076801-11076801 on Build GRCh38
rs372295246 Chr.1: 11076537-11076537 on Build GRCh38
rs763574577 Chr.1: 11076924-11076924 on Build GRCh38
rs748229694 Chr.1: 11076870-11076870 on Build GRCh38
rs904076439 Chr.1: 11076760-11076760 on Build GRCh38
rs372778448 Chr.1: 11076940-11076940 on Build GRCh38
rs752296401 Chr.1: 11076738-11076738 on Build GRCh38
rs751220639 Chr.1: 11076926-11076926 on Build GRCh38
rs1040965511 Chr.1: 11076821-11076821 on Build GRCh38
rs763221041 Chr.1: 11076509-11076509 on Build GRCh38
rs755063801 Chr.1: 11076631-11076631 on Build GRCh38
rs928187156 Chr.1: 11076691-11076691 on Build GRCh38
rs1044594114 Chr.1: 11076551-11076551 on Build GRCh38
rs143463195 Chr.1: 11076549-11076549 on Build GRCh38
rs764816118 Chr.1: 11076804-11076804 on Build GRCh38
rs780351106 Chr.1: 11076852-11076852 on Build GRCh38
rs375142573 Chr.1: 11076805-11076805 on Build GRCh38
rs865965759 Chr.1: 11076666-11076667 on Build GRCh38
rs753180765 Chr.1: 11076746-11076746 on Build GRCh38
rs755002258 Chr.1: 11076798-11076798 on Build GRCh38
rs759495336 Chr.1: 11076893-11076893 on Build GRCh38
rs1025441481 Chr.1: 11076878-11076878 on Build GRCh38
rs75232193 Chr.1: 11076489-11076489 on Build GRCh38
rs368146460 Chr.1: 11076856-11076856 on Build GRCh38
rs901044968 Chr.1: 11076541-11076541 on Build GRCh38
rs772126275 Chr.1: 11076874-11076874 on Build GRCh38
rs988325095 Chr.1: 11076745-11076745 on Build GRCh38
rs756626208 Chr.1: 11076747-11076747 on Build GRCh38
rs776613377 Chr.1: 11076892-11076892 on Build GRCh38
rs938553364 Chr.1: 11076659-11076659 on Build GRCh38
rs369040494 Chr.1: 11076849-11076849 on Build GRCh38
rs950824946 Chr.1: 11076932-11076932 on Build GRCh38
rs756488116 Chr.1: 11076851-11076851 on Build GRCh38
rs35791772 Chr.1: 11076668-11076669 on Build GRCh38
rs1007819958 Chr.1: 11076772-11076772 on Build GRCh38
rs752190706 Chr.1: 11076921-11076921 on Build GRCh38
rs758087431 Chr.1: 11076838-11076838 on Build GRCh38
rs145033383 Chr.1: 11076888-11076888 on Build GRCh38
rs17036297 Chr.1: 11076781-11076781 on Build GRCh38
rs752280430 Chr.1: 11076813-11076813 on Build GRCh38
rs17036295 Chr.1: 11076511-11076511 on Build GRCh38
rs969078490 Chr.1: 11076645-11076645 on Build GRCh38
rs756900921 Chr.1: 11076939-11076939 on Build GRCh38
rs1025410467 Chr.1: 11076596-11076596 on Build GRCh38
rs751771453 Chr.1: 11076600-11076600 on Build GRCh38
rs560785624 Chr.1: 11076706-11076706 on Build GRCh38
rs762542262 Chr.1: 11076923-11076923 on Build GRCh38
rs889152135 Chr.1: 11076784-11076784 on Build GRCh38
rs148821124 Chr.1: 11076502-11076502 on Build GRCh38
rs866827597 Chr.1: 11076629-11076629 on Build GRCh38

View Full Product Details