Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101921835-101922347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRAM1
Entrez Gene ID
55332
Gene Name
DNA damage regulated autophagy modulator 1
Gene Aliases
DRAM
UniGene
Hs.525634
Gene Chromosome Location
Chr.12: 101877327-101923623 on Build GRCh38
Transcripts
NM_018370.2 XM_005269004.2 XM_005269005.2 XM_017019578.1

SNP Detail

SNP ID Chromosome Location
rs4764679 Chr.12: 101922147-101922147 on Build GRCh38
rs967960546 Chr.12: 101922101-101922101 on Build GRCh38
rs7135515 Chr.12: 101921968-101921968 on Build GRCh38
rs150973156 Chr.12: 101921880-101921880 on Build GRCh38
rs4764680 Chr.12: 101922187-101922187 on Build GRCh38
rs971595808 Chr.12: 101922012-101922012 on Build GRCh38
rs17434787 Chr.12: 101922013-101922013 on Build GRCh38
rs61936586 Chr.12: 101922240-101922240 on Build GRCh38
rs1028302864 Chr.12: 101921959-101921959 on Build GRCh38
rs4764682 Chr.12: 101922243-101922243 on Build GRCh38
rs914572026 Chr.12: 101922097-101922097 on Build GRCh38
rs977934389 Chr.12: 101922008-101922008 on Build GRCh38
rs55954375 Chr.12: 101922298-101922298 on Build GRCh38
rs145841429 Chr.12: 101922126-101922126 on Build GRCh38
rs140785522 Chr.12: 101921967-101921967 on Build GRCh38
rs185752065 Chr.12: 101922132-101922132 on Build GRCh38
rs995883344 Chr.12: 101921935-101921935 on Build GRCh38
rs959301433 Chr.12: 101921947-101921947 on Build GRCh38
rs772541013 Chr.12: 101922016-101922016 on Build GRCh38
rs944574286 Chr.12: 101922128-101922128 on Build GRCh38
rs990144968 Chr.12: 101922082-101922082 on Build GRCh38
rs376741646 Chr.12: 101922287-101922287 on Build GRCh38
rs900167099 Chr.12: 101921920-101921920 on Build GRCh38
rs902122074 Chr.12: 101922055-101922055 on Build GRCh38
rs545074328 Chr.12: 101921941-101921941 on Build GRCh38
rs1034463983 Chr.12: 101922056-101922056 on Build GRCh38
rs970653400 Chr.12: 101922236-101922236 on Build GRCh38
rs867114065 Chr.12: 101921862-101921862 on Build GRCh38
rs147778696 Chr.12: 101922306-101922306 on Build GRCh38
rs978414352 Chr.12: 101922106-101922106 on Build GRCh38
rs943379691 Chr.12: 101922049-101922049 on Build GRCh38
rs990478187 Chr.12: 101921957-101921957 on Build GRCh38
rs775829905 Chr.12: 101922235-101922235 on Build GRCh38
rs577383591 Chr.12: 101921881-101921881 on Build GRCh38
rs921710457 Chr.12: 101922136-101922136 on Build GRCh38
rs17032069 Chr.12: 101921982-101921982 on Build GRCh38
rs914637046 Chr.12: 101922042-101922042 on Build GRCh38
rs4764681 Chr.12: 101922204-101922204 on Build GRCh38
rs776693695 Chr.12: 101922174-101922174 on Build GRCh38
rs1039023271 Chr.12: 101922054-101922054 on Build GRCh38
rs534474797 Chr.12: 101922250-101922250 on Build GRCh38
rs546593590 Chr.12: 101922080-101922080 on Build GRCh38
rs1024593383 Chr.12: 101922234-101922234 on Build GRCh38
rs1015066147 Chr.12: 101922010-101922010 on Build GRCh38
rs544184356 Chr.12: 101921980-101921980 on Build GRCh38

View Full Product Details