Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101922767-101923313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRAM1
Entrez Gene ID
55332
Gene Name
DNA damage regulated autophagy modulator 1
Gene Aliases
DRAM
UniGene
Hs.525634
Gene Chromosome Location
Chr.12: 101877327-101923623 on Build GRCh38
Transcripts
NM_018370.2 XM_005269004.2 XM_005269005.2 XM_017019578.1

SNP Detail

SNP ID Chromosome Location
rs146142574 Chr.12: 101923093-101923094 on Build GRCh38
rs116370125 Chr.12: 101923254-101923254 on Build GRCh38
rs1024644012 Chr.12: 101923053-101923053 on Build GRCh38
rs1011988004 Chr.12: 101923051-101923051 on Build GRCh38
rs549238813 Chr.12: 101923152-101923155 on Build GRCh38
rs113706528 Chr.12: 101923185-101923185 on Build GRCh38
rs576725258 Chr.12: 101923065-101923065 on Build GRCh38
rs143348179 Chr.12: 101923211-101923211 on Build GRCh38
rs190415232 Chr.12: 101922794-101922794 on Build GRCh38
rs563617505 Chr.12: 101923151-101923151 on Build GRCh38
rs542021592 Chr.12: 101923114-101923114 on Build GRCh38
rs76866994 Chr.12: 101922813-101922813 on Build GRCh38
rs546567433 Chr.12: 101923160-101923161 on Build GRCh38
rs554435833 Chr.12: 101922870-101922870 on Build GRCh38
rs544308889 Chr.12: 101923134-101923134 on Build GRCh38
rs923312790 Chr.12: 101923173-101923173 on Build GRCh38
rs56363228 Chr.12: 101923277-101923277 on Build GRCh38
rs775774849 Chr.12: 101923094-101923097 on Build GRCh38
rs569887070 Chr.12: 101922895-101922895 on Build GRCh38
rs534662203 Chr.12: 101923150-101923150 on Build GRCh38
rs533968599 Chr.12: 101922860-101922860 on Build GRCh38
rs181583213 Chr.12: 101923164-101923164 on Build GRCh38
rs781706279 Chr.12: 101923262-101923262 on Build GRCh38
rs752438831 Chr.12: 101922975-101922975 on Build GRCh38
rs1030888959 Chr.12: 101923097-101923097 on Build GRCh38
rs780944328 Chr.12: 101923097-101923098 on Build GRCh38
rs540138023 Chr.12: 101922963-101922963 on Build GRCh38
rs149627780 Chr.12: 101922904-101922904 on Build GRCh38
rs186541210 Chr.12: 101923259-101923259 on Build GRCh38
rs1018104621 Chr.12: 101923142-101923142 on Build GRCh38
rs58464864 Chr.12: 101923113-101923114 on Build GRCh38
rs568429087 Chr.12: 101922823-101922823 on Build GRCh38
rs34054548 Chr.12: 101923088-101923089 on Build GRCh38
rs1037854520 Chr.12: 101922995-101922995 on Build GRCh38
rs554731477 Chr.12: 101923141-101923141 on Build GRCh38
rs747267556 Chr.12: 101923110-101923110 on Build GRCh38
rs778210021 Chr.12: 101923063-101923063 on Build GRCh38
rs537307298 Chr.12: 101922926-101922926 on Build GRCh38
rs117146578 Chr.12: 101922985-101922985 on Build GRCh38
rs894926637 Chr.12: 101923034-101923034 on Build GRCh38

View Full Product Details