Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113185608-113186099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RITA1
Entrez Gene ID
84934
Gene Name
RBPJ interacting and tubulin associated 1
Gene Aliases
C12orf52, RITA
UniGene
Hs.524762
Gene Chromosome Location
Chr.12: 113185526-113192368 on Build GRCh38
Transcripts
NM_001286215.1 NM_032848.2 XM_005253972.2

SNP Detail

SNP ID Chromosome Location
rs989996732 Chr.12: 113185871-113185871 on Build GRCh38
rs376754506 Chr.12: 113185686-113185686 on Build GRCh38
rs935890932 Chr.12: 113185729-113185729 on Build GRCh38
rs910815020 Chr.12: 113185725-113185725 on Build GRCh38
rs921075848 Chr.12: 113185887-113185887 on Build GRCh38
rs1050737080 Chr.12: 113185638-113185638 on Build GRCh38
rs921211808 Chr.12: 113185742-113185742 on Build GRCh38
rs890758405 Chr.12: 113185761-113185761 on Build GRCh38
rs994993703 Chr.12: 113185793-113185793 on Build GRCh38
rs1003943142 Chr.12: 113185646-113185646 on Build GRCh38
rs977824214 Chr.12: 113185817-113185817 on Build GRCh38
rs1036821920 Chr.12: 113185785-113185785 on Build GRCh38
rs1013032872 Chr.12: 113186068-113186068 on Build GRCh38
rs952109240 Chr.12: 113185872-113185872 on Build GRCh38
rs971457797 Chr.12: 113185810-113185810 on Build GRCh38
rs757028028 Chr.12: 113185972-113185972 on Build GRCh38
rs890893594 Chr.12: 113185639-113185639 on Build GRCh38
rs931039803 Chr.12: 113185965-113185965 on Build GRCh38
rs904419244 Chr.12: 113185634-113185634 on Build GRCh38
rs550867280 Chr.12: 113185839-113185839 on Build GRCh38
rs958932608 Chr.12: 113185694-113185694 on Build GRCh38
rs536618420 Chr.12: 113186052-113186052 on Build GRCh38
rs1024234909 Chr.12: 113185712-113185712 on Build GRCh38
rs11544008 Chr.12: 113185754-113185754 on Build GRCh38
rs143072480 Chr.12: 113185857-113185857 on Build GRCh38
rs866075719 Chr.12: 113185809-113185809 on Build GRCh38
rs752575020 Chr.12: 113186003-113186003 on Build GRCh38
rs775637061 Chr.12: 113185730-113185730 on Build GRCh38
rs924684066 Chr.12: 113185726-113185726 on Build GRCh38
rs976228229 Chr.12: 113185946-113185946 on Build GRCh38
rs977502198 Chr.12: 113185721-113185721 on Build GRCh38
rs1050768302 Chr.12: 113185758-113185758 on Build GRCh38
rs57137190 Chr.12: 113185868-113185868 on Build GRCh38
rs1037091119 Chr.12: 113185955-113185955 on Build GRCh38
rs989965770 Chr.12: 113185737-113185737 on Build GRCh38
rs531598176 Chr.12: 113185669-113185669 on Build GRCh38
rs4397912 Chr.12: 113185851-113185851 on Build GRCh38
rs932558505 Chr.12: 113185892-113185892 on Build GRCh38
rs962085306 Chr.12: 113185832-113185832 on Build GRCh38
rs1024319896 Chr.12: 113185662-113185663 on Build GRCh38
rs376198185 Chr.12: 113185797-113185797 on Build GRCh38
rs914108966 Chr.12: 113185950-113185950 on Build GRCh38
rs962116381 Chr.12: 113185679-113185679 on Build GRCh38
rs991461321 Chr.12: 113185699-113185699 on Build GRCh38
rs11544009 Chr.12: 113185870-113185870 on Build GRCh38
rs920619783 Chr.12: 113185716-113185716 on Build GRCh38
rs945800824 Chr.12: 113185786-113185786 on Build GRCh38
rs76390746 Chr.12: 113185956-113185956 on Build GRCh38
rs971888374 Chr.12: 113185718-113185718 on Build GRCh38
rs569965491 Chr.12: 113186016-113186016 on Build GRCh38
rs972026740 Chr.12: 113185706-113185706 on Build GRCh38
rs1015515406 Chr.12: 113185656-113185656 on Build GRCh38
rs778960195 Chr.12: 113185939-113185940 on Build GRCh38
rs1013080405 Chr.12: 113185803-113185803 on Build GRCh38
rs1049220026 Chr.12: 113186006-113186006 on Build GRCh38
rs898027683 Chr.12: 113185657-113185657 on Build GRCh38
rs894371694 Chr.12: 113185972-113185973 on Build GRCh38
rs776567047 Chr.12: 113185801-113185801 on Build GRCh38
rs1055418154 Chr.12: 113185830-113185830 on Build GRCh38
rs374538335 Chr.12: 113185953-113185953 on Build GRCh38
rs146684478 Chr.12: 113185682-113185682 on Build GRCh38
rs1031781014 Chr.12: 113185820-113185820 on Build GRCh38
rs532443329 Chr.12: 113185776-113185776 on Build GRCh38
rs900908354 Chr.12: 113185771-113185771 on Build GRCh38
rs57210700 Chr.12: 113185894-113185894 on Build GRCh38
rs1046023130 Chr.12: 113186078-113186078 on Build GRCh38
rs762397479 Chr.12: 113185936-113185936 on Build GRCh38
rs768760649 Chr.12: 113185789-113185789 on Build GRCh38
rs945769500 Chr.12: 113185952-113185952 on Build GRCh38
rs930908517 Chr.12: 113185717-113185717 on Build GRCh38
rs545304892 Chr.12: 113185818-113185818 on Build GRCh38
rs932330891 Chr.12: 113185634-113185634 on Build GRCh38
rs139235943 Chr.12: 113185942-113185942 on Build GRCh38
rs899749065 Chr.12: 113185630-113185630 on Build GRCh38
rs993581826 Chr.12: 113185842-113185842 on Build GRCh38
rs548186802 Chr.12: 113186000-113186000 on Build GRCh38

View Full Product Details