Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 116949485-116949982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW8
Entrez Gene ID
26259
Gene Name
F-box and WD repeat domain containing 8
Gene Aliases
FBW6, FBW8, FBX29, FBXO29, FBXW6
UniGene
Hs.624537
Gene Chromosome Location
Chr.12: 116910949-117032498 on Build GRCh38
Transcripts
NM_012174.1 NM_153348.2 XM_005253867.2 XM_005253868.3 XM_017019175.1 XM_017019176.1 XM_017019177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7367974 c.634G>C p.V212L Chr.12: 116949663-116949663 on Build GRCh38
4188046 c.667G>T p.V223F Chr.12: 116949696-116949696 on Build GRCh38
2173417 c.593G>A p.R198H Chr.12: 116949622-116949622 on Build GRCh38
7413903 c.638T>C p.L213S Chr.12: 116949667-116949667 on Build GRCh38
6891800 c.616G>A p.E206K Chr.12: 116949645-116949645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772181283 Chr.12: 116949589-116949589 on Build GRCh38
rs751163360 Chr.12: 116949641-116949641 on Build GRCh38
rs575752045 Chr.12: 116949622-116949622 on Build GRCh38
rs755710991 Chr.12: 116949686-116949687 on Build GRCh38
rs962644275 Chr.12: 116949701-116949701 on Build GRCh38
rs911004192 Chr.12: 116949830-116949830 on Build GRCh38
rs950623349 Chr.12: 116949527-116949527 on Build GRCh38
rs552160533 Chr.12: 116949853-116949853 on Build GRCh38
rs145405264 Chr.12: 116949657-116949657 on Build GRCh38
rs983306958 Chr.12: 116949531-116949531 on Build GRCh38
rs1047102448 Chr.12: 116949903-116949903 on Build GRCh38
rs758133267 Chr.12: 116949630-116949630 on Build GRCh38
rs1045113370 Chr.12: 116949930-116949930 on Build GRCh38
rs948068978 Chr.12: 116949915-116949915 on Build GRCh38
rs760910615 Chr.12: 116949699-116949699 on Build GRCh38
rs145823135 Chr.12: 116949687-116949687 on Build GRCh38
rs935210779 Chr.12: 116949882-116949882 on Build GRCh38
rs373550067 Chr.12: 116949570-116949570 on Build GRCh38
rs961058355 Chr.12: 116949542-116949542 on Build GRCh38
rs778152410 Chr.12: 116949579-116949579 on Build GRCh38
rs753094701 Chr.12: 116949632-116949632 on Build GRCh38
rs7136629 Chr.12: 116949840-116949840 on Build GRCh38
rs1016922910 Chr.12: 116949950-116949950 on Build GRCh38
rs758023365 Chr.12: 116949568-116949568 on Build GRCh38
rs17583110 Chr.12: 116949765-116949765 on Build GRCh38
rs1054013262 Chr.12: 116949631-116949631 on Build GRCh38
rs771554969 Chr.12: 116949677-116949677 on Build GRCh38
rs894072723 Chr.12: 116949639-116949639 on Build GRCh38
rs753325159 Chr.12: 116949715-116949715 on Build GRCh38
rs750114718 Chr.12: 116949567-116949567 on Build GRCh38
rs764615831 Chr.12: 116949742-116949742 on Build GRCh38
rs987196962 Chr.12: 116949827-116949827 on Build GRCh38
rs985035414 Chr.12: 116949508-116949508 on Build GRCh38
rs1045628843 Chr.12: 116949865-116949865 on Build GRCh38
rs548890737 Chr.12: 116949906-116949906 on Build GRCh38
rs775599245 Chr.12: 116949691-116949691 on Build GRCh38
rs1043991885 Chr.12: 116949861-116949861 on Build GRCh38
rs116549491 Chr.12: 116949881-116949881 on Build GRCh38
rs749918174 Chr.12: 116949623-116949623 on Build GRCh38
rs775717341 Chr.12: 116949592-116949592 on Build GRCh38
rs748455020 Chr.12: 116949571-116949571 on Build GRCh38
rs759751109 Chr.12: 116949613-116949613 on Build GRCh38
rs61748074 Chr.12: 116949688-116949688 on Build GRCh38
rs1045277600 Chr.12: 116949654-116949654 on Build GRCh38
rs887885053 Chr.12: 116949954-116949954 on Build GRCh38
rs559339878 Chr.12: 116949583-116949583 on Build GRCh38
rs754254779 Chr.12: 116949744-116949744 on Build GRCh38
rs1056312527 Chr.12: 116949904-116949904 on Build GRCh38
rs902453236 Chr.12: 116949586-116949586 on Build GRCh38
rs915478087 Chr.12: 116949856-116949856 on Build GRCh38
rs934950936 Chr.12: 116949849-116949849 on Build GRCh38
rs376186920 Chr.12: 116949741-116949741 on Build GRCh38
rs746338707 Chr.12: 116949758-116949758 on Build GRCh38
rs746815866 Chr.12: 116949919-116949919 on Build GRCh38
rs559069589 Chr.12: 116949560-116949560 on Build GRCh38
rs79542857 Chr.12: 116949520-116949520 on Build GRCh38
rs540611091 Chr.12: 116949872-116949872 on Build GRCh38
rs887081027 Chr.12: 116949703-116949703 on Build GRCh38
rs370466421 Chr.12: 116949569-116949569 on Build GRCh38
rs756646324 Chr.12: 116949633-116949633 on Build GRCh38
rs749863935 Chr.12: 116949673-116949673 on Build GRCh38
rs1028852273 Chr.12: 116949784-116949784 on Build GRCh38
rs751213188 Chr.12: 116949738-116949738 on Build GRCh38
rs776406255 Chr.12: 116949555-116949555 on Build GRCh38
rs759548178 Chr.12: 116949686-116949686 on Build GRCh38
rs933696961 Chr.12: 116949557-116949557 on Build GRCh38
rs771178945 Chr.12: 116949581-116949581 on Build GRCh38
rs371809248 Chr.12: 116949706-116949706 on Build GRCh38
rs36021180 Chr.12: 116949660-116949660 on Build GRCh38
rs368238777 Chr.12: 116949702-116949702 on Build GRCh38
rs372725345 Chr.12: 116949578-116949578 on Build GRCh38
rs183809803 Chr.12: 116949842-116949842 on Build GRCh38
rs748755801 Chr.12: 116949741-116949742 on Build GRCh38
rs765151874 Chr.12: 116949718-116949718 on Build GRCh38
rs972831447 Chr.12: 116949549-116949549 on Build GRCh38
rs779576974 Chr.12: 116949750-116949750 on Build GRCh38
rs985181120 Chr.12: 116949824-116949824 on Build GRCh38
rs192129939 Chr.12: 116949719-116949719 on Build GRCh38
rs753804526 Chr.12: 116949621-116949621 on Build GRCh38
rs989151706 Chr.12: 116949852-116949852 on Build GRCh38
rs757629664 Chr.12: 116949747-116949747 on Build GRCh38
rs754639619 Chr.12: 116949739-116949739 on Build GRCh38
rs777413351 Chr.12: 116949740-116949747 on Build GRCh38
rs765861906 Chr.12: 116949731-116949731 on Build GRCh38
rs774699272 Chr.12: 116949563-116949563 on Build GRCh38
rs74977947 Chr.12: 116949614-116949614 on Build GRCh38

View Full Product Details