Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20404526-20404988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYL1
Entrez Gene ID
51084
Gene Name
crystallin lambda 1
Gene Aliases
GDH, HEL30, lambda-CRY
UniGene
Hs.370703
Gene Chromosome Location
Chr.13: 20403667-20525873 on Build GRCh38
Transcripts
NM_015974.2 XM_005266415.3 XM_017020624.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
945877 c.778A>C p.K260Q Chr.13: 20404703-20404703 on Build GRCh38
4779173 c.748A>G p.S250G Chr.13: 20404733-20404733 on Build GRCh38
4046381 c.843C>T p.N281N Chr.13: 20404638-20404638 on Build GRCh38
4572724 c.840T>G p.V280V Chr.13: 20404641-20404641 on Build GRCh38
695836 c.846+1G>T p.? Chr.13: 20404634-20404634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049420891 Chr.13: 20404798-20404798 on Build GRCh38
rs929522372 Chr.13: 20404740-20404740 on Build GRCh38
rs151322996 Chr.13: 20404556-20404556 on Build GRCh38
rs939789061 Chr.13: 20404762-20404762 on Build GRCh38
rs889727613 Chr.13: 20404902-20404902 on Build GRCh38
rs1020977525 Chr.13: 20404583-20404583 on Build GRCh38
rs751840935 Chr.13: 20404601-20404601 on Build GRCh38
rs1019879612 Chr.13: 20404929-20404929 on Build GRCh38
rs578063236 Chr.13: 20404894-20404894 on Build GRCh38
rs370272058 Chr.13: 20404690-20404690 on Build GRCh38
rs762042341 Chr.13: 20404691-20404691 on Build GRCh38
rs367679286 Chr.13: 20404666-20404666 on Build GRCh38
rs953897591 Chr.13: 20404700-20404700 on Build GRCh38
rs771025628 Chr.13: 20404661-20404661 on Build GRCh38
rs745867128 Chr.13: 20404753-20404753 on Build GRCh38
rs768014340 Chr.13: 20404699-20404699 on Build GRCh38
rs763632884 Chr.13: 20404685-20404685 on Build GRCh38
rs750870267 Chr.13: 20404703-20404703 on Build GRCh38
rs769824555 Chr.13: 20404770-20404770 on Build GRCh38
rs528412596 Chr.13: 20404938-20404938 on Build GRCh38
rs770796343 Chr.13: 20404738-20404738 on Build GRCh38
rs546505969 Chr.13: 20404559-20404559 on Build GRCh38
rs949505500 Chr.13: 20404828-20404828 on Build GRCh38
rs759134464 Chr.13: 20404668-20404668 on Build GRCh38
rs780268367 Chr.13: 20404632-20404632 on Build GRCh38
rs902658696 Chr.13: 20404874-20404874 on Build GRCh38
rs773807408 Chr.13: 20404793-20404793 on Build GRCh38
rs776646147 Chr.13: 20404662-20404662 on Build GRCh38
rs369395294 Chr.13: 20404727-20404727 on Build GRCh38
rs201075706 Chr.13: 20404646-20404646 on Build GRCh38
rs756119140 Chr.13: 20404629-20404629 on Build GRCh38
rs779181144 Chr.13: 20404777-20404778 on Build GRCh38
rs545468443 Chr.13: 20404898-20404898 on Build GRCh38
rs780171118 Chr.13: 20404713-20404713 on Build GRCh38
rs141893577 Chr.13: 20404557-20404557 on Build GRCh38
rs564413728 Chr.13: 20404553-20404553 on Build GRCh38
rs386768630 Chr.13: 20404888-20404889 on Build GRCh38
rs781626657 Chr.13: 20404627-20404627 on Build GRCh38
rs762500431 Chr.13: 20404592-20404592 on Build GRCh38
rs762707870 Chr.13: 20404683-20404683 on Build GRCh38
rs568132908 Chr.13: 20404598-20404598 on Build GRCh38
rs879902329 Chr.13: 20404888-20404889 on Build GRCh38
rs573819678 Chr.13: 20404555-20404557 on Build GRCh38
rs752343725 Chr.13: 20404589-20404589 on Build GRCh38
rs1025387703 Chr.13: 20404957-20404957 on Build GRCh38
rs368728558 Chr.13: 20404659-20404659 on Build GRCh38
rs902511671 Chr.13: 20404860-20404860 on Build GRCh38
rs142394913 Chr.13: 20404803-20404803 on Build GRCh38
rs938173969 Chr.13: 20404796-20404796 on Build GRCh38
rs147724770 Chr.13: 20404725-20404725 on Build GRCh38
rs1006703268 Chr.13: 20404921-20404921 on Build GRCh38
rs200417608 Chr.13: 20404780-20404780 on Build GRCh38
rs970407601 Chr.13: 20404961-20404961 on Build GRCh38
rs750531464 Chr.13: 20404628-20404628 on Build GRCh38
rs377245202 Chr.13: 20404652-20404652 on Build GRCh38
rs577167120 Chr.13: 20404797-20404797 on Build GRCh38
rs775135564 Chr.13: 20404675-20404675 on Build GRCh38
rs184596127 Chr.13: 20404712-20404712 on Build GRCh38
rs67871298 Chr.13: 20404820-20404821 on Build GRCh38
rs1019344558 Chr.13: 20404714-20404714 on Build GRCh38
rs35768694 Chr.13: 20404889-20404889 on Build GRCh38
rs751973920 Chr.13: 20404730-20404730 on Build GRCh38
rs145530540 Chr.13: 20404724-20404724 on Build GRCh38
rs769490135 Chr.13: 20404669-20404669 on Build GRCh38
rs902394914 Chr.13: 20404590-20404590 on Build GRCh38
rs7331073 Chr.13: 20404888-20404888 on Build GRCh38
rs6490564 Chr.13: 20404965-20404965 on Build GRCh38
rs746828889 Chr.13: 20404728-20404728 on Build GRCh38
rs3818618 Chr.13: 20404889-20404889 on Build GRCh38
rs564353928 Chr.13: 20404935-20404935 on Build GRCh38
rs749577909 Chr.13: 20404636-20404636 on Build GRCh38
rs745810845 Chr.13: 20404679-20404679 on Build GRCh38
rs116116968 Chr.13: 20404792-20404792 on Build GRCh38
rs999549359 Chr.13: 20404686-20404686 on Build GRCh38
rs757833199 Chr.13: 20404609-20404609 on Build GRCh38
rs556557136 Chr.13: 20404891-20404891 on Build GRCh38
rs781125397 Chr.13: 20404750-20404750 on Build GRCh38
rs746272882 Chr.13: 20404778-20404778 on Build GRCh38
rs188195058 Chr.13: 20404947-20404947 on Build GRCh38
rs555716908 Chr.13: 20404837-20404837 on Build GRCh38
rs748700308 Chr.13: 20404784-20404784 on Build GRCh38
rs775472496 Chr.13: 20404771-20404771 on Build GRCh38

View Full Product Details