Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51932978-51933486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP7B
Entrez Gene ID
540
Gene Name
ATPase copper transporting beta
Gene Aliases
PWD, WC1, WD, WND
UniGene
Hs.492280
Gene Chromosome Location
Chr.13: 51932669-52012130 on Build GRCh38
Transcripts
NM_000053.3 NM_001005918.2 NM_001243182.1 XM_005266423.2 XM_005266424.4 XM_005266427.2 XM_005266428.1 XM_005266430.4 XM_005266431.3 XM_005266432.2 XM_006719837.3 XM_006719838.1 XM_006719839.1 XM_011535117.2 XM_011535118.1 XM_011535119.1 XM_011535122.2 XM_017020627.1 XM_017020628.1

SNP Detail

SNP ID Chromosome Location
rs151138735 Chr.13: 51933087-51933087 on Build GRCh38
rs936464874 Chr.13: 51933076-51933076 on Build GRCh38
rs1007747915 Chr.13: 51933170-51933170 on Build GRCh38
rs747998965 Chr.13: 51933240-51933240 on Build GRCh38
rs886050298 Chr.13: 51933198-51933198 on Build GRCh38
rs139850360 Chr.13: 51933011-51933011 on Build GRCh38
rs188364702 Chr.13: 51933179-51933179 on Build GRCh38
rs77770386 Chr.13: 51933039-51933039 on Build GRCh38
rs1000565729 Chr.13: 51933222-51933222 on Build GRCh38
rs139053981 Chr.13: 51933265-51933265 on Build GRCh38
rs943483127 Chr.13: 51933425-51933425 on Build GRCh38
rs926332692 Chr.13: 51933302-51933302 on Build GRCh38
rs773178127 Chr.13: 51933387-51933387 on Build GRCh38
rs17076111 Chr.13: 51933048-51933048 on Build GRCh38
rs890608411 Chr.13: 51933164-51933164 on Build GRCh38
rs945045204 Chr.13: 51933047-51933047 on Build GRCh38
rs545813070 Chr.13: 51933029-51933029 on Build GRCh38
rs975847737 Chr.13: 51933263-51933263 on Build GRCh38
rs537012324 Chr.13: 51933347-51933347 on Build GRCh38
rs966629866 Chr.13: 51933277-51933277 on Build GRCh38
rs897373492 Chr.13: 51933173-51933173 on Build GRCh38
rs904612527 Chr.13: 51933198-51933198 on Build GRCh38
rs746211484 Chr.13: 51933010-51933010 on Build GRCh38
rs113688195 Chr.13: 51933097-51933097 on Build GRCh38
rs568184687 Chr.13: 51933320-51933320 on Build GRCh38
rs567043686 Chr.13: 51933257-51933257 on Build GRCh38
rs916283148 Chr.13: 51933401-51933401 on Build GRCh38
rs1006992965 Chr.13: 51933247-51933247 on Build GRCh38
rs548262136 Chr.13: 51933307-51933307 on Build GRCh38
rs1037380425 Chr.13: 51933068-51933068 on Build GRCh38
rs973602424 Chr.13: 51933328-51933328 on Build GRCh38
rs980644724 Chr.13: 51933279-51933279 on Build GRCh38
rs41292780 Chr.13: 51933371-51933371 on Build GRCh38
rs949535301 Chr.13: 51933407-51933407 on Build GRCh38
rs555136047 Chr.13: 51933138-51933138 on Build GRCh38
rs143285115 Chr.13: 51933050-51933050 on Build GRCh38
rs770432302 Chr.13: 51933460-51933460 on Build GRCh38
rs964877517 Chr.13: 51933027-51933027 on Build GRCh38
rs1013734395 Chr.13: 51933145-51933146 on Build GRCh38
rs9535794 Chr.13: 51933012-51933012 on Build GRCh38
rs886050297 Chr.13: 51933164-51933164 on Build GRCh38
rs191892694 Chr.13: 51933234-51933234 on Build GRCh38
rs928976951 Chr.13: 51933363-51933363 on Build GRCh38
rs1039499463 Chr.13: 51933178-51933178 on Build GRCh38
rs1009426310 Chr.13: 51933256-51933256 on Build GRCh38
rs192653965 Chr.13: 51933009-51933009 on Build GRCh38
rs368996327 Chr.13: 51933437-51933437 on Build GRCh38
rs956075093 Chr.13: 51933253-51933253 on Build GRCh38
rs886050299 Chr.13: 51933263-51933263 on Build GRCh38
rs564357604 Chr.13: 51933112-51933112 on Build GRCh38
rs977963892 Chr.13: 51933283-51933283 on Build GRCh38
rs573002084 Chr.13: 51933045-51933045 on Build GRCh38

View Full Product Details