Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35046271-35046788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM177A1
Entrez Gene ID
283635
Gene Name
family with sequence similarity 177 member A1
Gene Aliases
C14orf24
UniGene
Hs.446357
Gene Chromosome Location
Chr.14: 35044907-35083383 on Build GRCh38
Transcripts
NM_001079519.1 NM_001289022.1 NM_173607.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402591 c.25G>A p.V9M Chr.14: 35046557-35046557 on Build GRCh38
5456435 c.43G>A p.A15T Chr.14: 35046506-35046506 on Build GRCh38
7430403 c.45C>G p.V15V Chr.14: 35046577-35046577 on Build GRCh38
1734318 c.28C>G p.L10V Chr.14: 35046491-35046491 on Build GRCh38
6402590 c.94G>A p.V32M Chr.14: 35046557-35046557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146253684 Chr.14: 35046540-35046540 on Build GRCh38
rs978135830 Chr.14: 35046750-35046750 on Build GRCh38
rs374120883 Chr.14: 35046490-35046490 on Build GRCh38
rs377271090 Chr.14: 35046442-35046442 on Build GRCh38
rs768447281 Chr.14: 35046422-35046422 on Build GRCh38
rs201725571 Chr.14: 35046748-35046748 on Build GRCh38
rs139171323 Chr.14: 35046532-35046532 on Build GRCh38
rs760824126 Chr.14: 35046499-35046499 on Build GRCh38
rs1012024808 Chr.14: 35046361-35046361 on Build GRCh38
rs776356310 Chr.14: 35046423-35046423 on Build GRCh38
rs200121612 Chr.14: 35046533-35046533 on Build GRCh38
rs764244120 Chr.14: 35046587-35046587 on Build GRCh38
rs1016668544 Chr.14: 35046365-35046365 on Build GRCh38
rs765381513 Chr.14: 35046458-35046458 on Build GRCh38
rs761636749 Chr.14: 35046488-35046488 on Build GRCh38
rs974811522 Chr.14: 35046485-35046485 on Build GRCh38
rs546015694 Chr.14: 35046733-35046733 on Build GRCh38
rs953069563 Chr.14: 35046299-35046299 on Build GRCh38
rs957905132 Chr.14: 35046732-35046732 on Build GRCh38
rs961755213 Chr.14: 35046657-35046657 on Build GRCh38
rs566154599 Chr.14: 35046325-35046325 on Build GRCh38
rs765650226 Chr.14: 35046599-35046599 on Build GRCh38
rs370076717 Chr.14: 35046335-35046335 on Build GRCh38
rs528303053 Chr.14: 35046752-35046752 on Build GRCh38
rs368712835 Chr.14: 35046555-35046555 on Build GRCh38
rs1015850016 Chr.14: 35046630-35046630 on Build GRCh38
rs766835801 Chr.14: 35046611-35046611 on Build GRCh38
rs577501612 Chr.14: 35046526-35046526 on Build GRCh38
rs765467452 Chr.14: 35046531-35046531 on Build GRCh38
rs905834051 Chr.14: 35046360-35046360 on Build GRCh38
rs955053270 Chr.14: 35046669-35046669 on Build GRCh38
rs373639704 Chr.14: 35046538-35046538 on Build GRCh38
rs201181123 Chr.14: 35046601-35046601 on Build GRCh38
rs754312052 Chr.14: 35046467-35046467 on Build GRCh38
rs200451528 Chr.14: 35046510-35046510 on Build GRCh38
rs917834721 Chr.14: 35046341-35046341 on Build GRCh38
rs937353622 Chr.14: 35046363-35046363 on Build GRCh38
rs537791034 Chr.14: 35046359-35046359 on Build GRCh38
rs776261868 Chr.14: 35046484-35046484 on Build GRCh38
rs1054492773 Chr.14: 35046534-35046534 on Build GRCh38
rs1054523879 Chr.14: 35046370-35046370 on Build GRCh38
rs768359400 Chr.14: 35046482-35046482 on Build GRCh38
rs756336210 Chr.14: 35046462-35046462 on Build GRCh38
rs1025569523 Chr.14: 35046426-35046426 on Build GRCh38
rs201860851 Chr.14: 35046544-35046544 on Build GRCh38
rs746335490 Chr.14: 35046476-35046476 on Build GRCh38
rs970599792 Chr.14: 35046520-35046520 on Build GRCh38
rs949287336 Chr.14: 35046355-35046355 on Build GRCh38
rs772100207 Chr.14: 35046764-35046765 on Build GRCh38
rs762757572 Chr.14: 35046492-35046492 on Build GRCh38
rs902727681 Chr.14: 35046294-35046294 on Build GRCh38
rs770832124 Chr.14: 35046582-35046582 on Build GRCh38
rs1045006350 Chr.14: 35046358-35046358 on Build GRCh38
rs771646413 Chr.14: 35046479-35046479 on Build GRCh38
rs773224837 Chr.14: 35046491-35046491 on Build GRCh38
rs752040724 Chr.14: 35046612-35046612 on Build GRCh38
rs747752812 Chr.14: 35046559-35046559 on Build GRCh38
rs750298028 Chr.14: 35046453-35046453 on Build GRCh38
rs754507124 Chr.14: 35046614-35046614 on Build GRCh38
rs914406201 Chr.14: 35046721-35046721 on Build GRCh38
rs766614773 Chr.14: 35046716-35046718 on Build GRCh38
rs769662828 Chr.14: 35046562-35046562 on Build GRCh38
rs758790505 Chr.14: 35046605-35046605 on Build GRCh38
rs370003655 Chr.14: 35046435-35046435 on Build GRCh38
rs757742252 Chr.14: 35046530-35046530 on Build GRCh38
rs1025765091 Chr.14: 35046660-35046660 on Build GRCh38
rs762063469 Chr.14: 35046594-35046594 on Build GRCh38
rs963854261 Chr.14: 35046712-35046712 on Build GRCh38
rs777011967 Chr.14: 35046593-35046593 on Build GRCh38
rs142982614 Chr.14: 35046699-35046699 on Build GRCh38
rs760207725 Chr.14: 35046415-35046415 on Build GRCh38
rs2295103 Chr.14: 35046714-35046714 on Build GRCh38
rs200956304 Chr.14: 35046496-35046496 on Build GRCh38
rs757687036 Chr.14: 35046473-35046473 on Build GRCh38
rs1024754518 Chr.14: 35046670-35046670 on Build GRCh38
rs759712498 Chr.14: 35046456-35046456 on Build GRCh38
rs780983492 Chr.14: 35046686-35046686 on Build GRCh38
rs962381403 Chr.14: 35046386-35046386 on Build GRCh38
rs373148442 Chr.14: 35046436-35046436 on Build GRCh38
rs747801958 Chr.14: 35046674-35046674 on Build GRCh38
rs548591220 Chr.14: 35046385-35046385 on Build GRCh38
rs758537555 Chr.14: 35046635-35046636 on Build GRCh38
rs45530731 Chr.14: 35046751-35046751 on Build GRCh38
rs543069717 Chr.14: 35046689-35046689 on Build GRCh38
rs749104104 Chr.14: 35046701-35046701 on Build GRCh38
rs760873641 Chr.14: 35046586-35046586 on Build GRCh38
rs898208465 Chr.14: 35046610-35046610 on Build GRCh38
rs972348017 Chr.14: 35046332-35046332 on Build GRCh38
rs767636712 Chr.14: 35046493-35046493 on Build GRCh38
rs993914392 Chr.14: 35046652-35046652 on Build GRCh38
rs746682313 Chr.14: 35046480-35046480 on Build GRCh38
rs750740906 Chr.14: 35046556-35046558 on Build GRCh38
rs780642611 Chr.14: 35046642-35046642 on Build GRCh38
rs926517186 Chr.14: 35046722-35046722 on Build GRCh38
rs911074205 Chr.14: 35046470-35046470 on Build GRCh38
rs548654520 Chr.14: 35046763-35046763 on Build GRCh38
rs370763722 Chr.14: 35046465-35046465 on Build GRCh38
rs4007475 Chr.14: 35046401-35046402 on Build GRCh38
rs560825552 Chr.14: 35046585-35046585 on Build GRCh38
rs752993000 Chr.14: 35046459-35046459 on Build GRCh38
rs1003799232 Chr.14: 35046305-35046305 on Build GRCh38
rs535170003 Chr.14: 35046379-35046379 on Build GRCh38
rs745748327 Chr.14: 35046727-35046727 on Build GRCh38
rs772814193 Chr.14: 35046572-35046572 on Build GRCh38
rs777535071 Chr.14: 35046319-35046319 on Build GRCh38
rs775254793 Chr.14: 35046414-35046414 on Build GRCh38

View Full Product Details