Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39290938-39291436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE5
Entrez Gene ID
4253
Gene Name
CTAGE family member 5, ER export factor
Gene Aliases
MEA6, MGEA, MGEA11, MGEA6
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265264-39351193 on Build GRCh38
Transcripts
NM_001247988.1 NM_001247989.1 NM_001247990.1 NM_005930.3 NM_203354.2 NM_203355.2 NM_203356.2 XM_005267648.4 XM_006720148.2 XM_011536775.2 XM_011536778.2 XM_011536784.2 XM_011536785.2 XM_017021314.1 XM_017021315.1 XM_017021316.1 XM_017021317.1 XM_017021318.1 XM_017021319.1 XM_017021320.1 XM_017021321.1 XM_017021322.1 XM_017021323.1 XM_017021324.1 XM_017021325.1 XM_017021326.1 XM_017021327.1 XM_017021328.1 XM_017021329.1 XM_017021330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6531901 c.384+3A>C p.? Chr.14: 39291099-39291099 on Build GRCh38
6341939 c.337A>G p.K113E Chr.14: 39291049-39291049 on Build GRCh38
6341941 c.337A>G p.K113E Chr.14: 39291049-39291049 on Build GRCh38
6531903 c.384+3A>C p.? Chr.14: 39291099-39291099 on Build GRCh38
4978840 c.315C>A p.G105G Chr.14: 39291027-39291027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766722324 Chr.14: 39291047-39291047 on Build GRCh38
rs765388324 Chr.14: 39291100-39291101 on Build GRCh38
rs775353159 Chr.14: 39291018-39291018 on Build GRCh38
rs764159496 Chr.14: 39291164-39291164 on Build GRCh38
rs189981106 Chr.14: 39291374-39291374 on Build GRCh38
rs539941849 Chr.14: 39291369-39291369 on Build GRCh38
rs985201866 Chr.14: 39291156-39291156 on Build GRCh38
rs557411519 Chr.14: 39291249-39291249 on Build GRCh38
rs778762082 Chr.14: 39290991-39290991 on Build GRCh38
rs777330091 Chr.14: 39290980-39290980 on Build GRCh38
rs745681501 Chr.14: 39291108-39291108 on Build GRCh38
rs751382675 Chr.14: 39291115-39291116 on Build GRCh38
rs565915367 Chr.14: 39290977-39290977 on Build GRCh38
rs753499187 Chr.14: 39291093-39291093 on Build GRCh38
rs536369298 Chr.14: 39290989-39290989 on Build GRCh38
rs1055233331 Chr.14: 39291256-39291256 on Build GRCh38
rs765658437 Chr.14: 39291393-39291393 on Build GRCh38
rs906727867 Chr.14: 39291224-39291227 on Build GRCh38
rs780010596 Chr.14: 39291119-39291119 on Build GRCh38
rs746947506 Chr.14: 39291125-39291125 on Build GRCh38
rs372208960 Chr.14: 39291075-39291075 on Build GRCh38
rs1054138102 Chr.14: 39291225-39291225 on Build GRCh38
rs140533519 Chr.14: 39291254-39291254 on Build GRCh38
rs528756977 Chr.14: 39291112-39291112 on Build GRCh38
rs558900790 Chr.14: 39291258-39291260 on Build GRCh38
rs754953254 Chr.14: 39291133-39291133 on Build GRCh38
rs576394156 Chr.14: 39291181-39291181 on Build GRCh38
rs1022591544 Chr.14: 39291354-39291354 on Build GRCh38
rs1004233437 Chr.14: 39291342-39291342 on Build GRCh38
rs941052030 Chr.14: 39291173-39291173 on Build GRCh38
rs1010913984 Chr.14: 39291299-39291299 on Build GRCh38
rs1037410814 Chr.14: 39290984-39290984 on Build GRCh38
rs552843649 Chr.14: 39291373-39291373 on Build GRCh38
rs763645570 Chr.14: 39291092-39291092 on Build GRCh38
rs1033927185 Chr.14: 39291363-39291363 on Build GRCh38
rs759922970 Chr.14: 39291073-39291073 on Build GRCh38
rs899418415 Chr.14: 39291184-39291184 on Build GRCh38
rs533063800 Chr.14: 39291028-39291028 on Build GRCh38
rs566267600 Chr.14: 39291103-39291103 on Build GRCh38
rs760010878 Chr.14: 39291158-39291158 on Build GRCh38
rs745593334 Chr.14: 39290995-39290995 on Build GRCh38
rs989779552 Chr.14: 39290968-39290968 on Build GRCh38
rs564064510 Chr.14: 39290962-39290962 on Build GRCh38
rs763225626 Chr.14: 39291039-39291039 on Build GRCh38
rs2274399 Chr.14: 39291189-39291189 on Build GRCh38
rs774667421 Chr.14: 39291147-39291147 on Build GRCh38
rs774861603 Chr.14: 39291054-39291054 on Build GRCh38
rs769825701 Chr.14: 39291138-39291138 on Build GRCh38
rs772007092 Chr.14: 39291003-39291003 on Build GRCh38
rs891514014 Chr.14: 39291241-39291241 on Build GRCh38
rs749262424 Chr.14: 39291142-39291142 on Build GRCh38
rs752122388 Chr.14: 39291243-39291243 on Build GRCh38
rs933926433 Chr.14: 39291132-39291132 on Build GRCh38
rs376694144 Chr.14: 39291084-39291084 on Build GRCh38
rs971071585 Chr.14: 39291288-39291288 on Build GRCh38
rs773357791 Chr.14: 39291029-39291029 on Build GRCh38
rs1047918486 Chr.14: 39291213-39291213 on Build GRCh38
rs753224655 Chr.14: 39291077-39291077 on Build GRCh38
rs771198897 Chr.14: 39291146-39291146 on Build GRCh38
rs910858050 Chr.14: 39291165-39291165 on Build GRCh38
rs772542657 Chr.14: 39291161-39291161 on Build GRCh38
rs377507981 Chr.14: 39291139-39291139 on Build GRCh38
rs1009920417 Chr.14: 39291279-39291279 on Build GRCh38
rs552343772 Chr.14: 39291280-39291280 on Build GRCh38
rs543373795 Chr.14: 39291178-39291178 on Build GRCh38
rs552468023 Chr.14: 39291099-39291099 on Build GRCh38
rs966595293 Chr.14: 39291368-39291368 on Build GRCh38
rs554609385 Chr.14: 39291058-39291058 on Build GRCh38
rs530502978 Chr.14: 39291276-39291276 on Build GRCh38
rs760899932 Chr.14: 39291020-39291020 on Build GRCh38
rs748946760 Chr.14: 39290986-39290986 on Build GRCh38

View Full Product Details