Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 53046013-53046520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDHD1
Entrez Gene ID
80821
Gene Name
DDHD domain containing 1
Gene Aliases
PA-PLA1, PAPLA1, SPG28
UniGene
Hs.125525
Gene Chromosome Location
Chr.14: 53036740-53153328 on Build GRCh38
Transcripts
NM_001160147.1 NM_001160148.1 NM_030637.2 XM_005268102.2 XM_005268103.2 XM_005268105.2 XM_011537188.2 XM_011537189.2 XM_017021668.1 XM_017021669.1

SNP Detail

SNP ID Chromosome Location
rs774065647 Chr.14: 53046348-53046348 on Build GRCh38
rs535107705 Chr.14: 53046235-53046235 on Build GRCh38
rs995843382 Chr.14: 53046286-53046286 on Build GRCh38
rs921765470 Chr.14: 53046483-53046483 on Build GRCh38
rs183766440 Chr.14: 53046410-53046410 on Build GRCh38
rs768594166 Chr.14: 53046285-53046285 on Build GRCh38
rs926431367 Chr.14: 53046406-53046406 on Build GRCh38
rs926481980 Chr.14: 53046048-53046049 on Build GRCh38
rs936567332 Chr.14: 53046474-53046474 on Build GRCh38
rs958865539 Chr.14: 53046355-53046355 on Build GRCh38
rs946109281 Chr.14: 53046194-53046194 on Build GRCh38
rs957015586 Chr.14: 53046341-53046341 on Build GRCh38
rs917373772 Chr.14: 53046376-53046376 on Build GRCh38
rs746386998 Chr.14: 53046044-53046044 on Build GRCh38
rs372183849 Chr.14: 53046039-53046039 on Build GRCh38
rs759505075 Chr.14: 53046388-53046388 on Build GRCh38
rs917226224 Chr.14: 53046109-53046109 on Build GRCh38
rs1015571342 Chr.14: 53046343-53046343 on Build GRCh38
rs370669233 Chr.14: 53046057-53046057 on Build GRCh38
rs1042690356 Chr.14: 53046211-53046211 on Build GRCh38
rs1053632474 Chr.14: 53046481-53046481 on Build GRCh38
rs980508369 Chr.14: 53046391-53046391 on Build GRCh38
rs1051498180 Chr.14: 53046305-53046305 on Build GRCh38
rs867889918 Chr.14: 53046353-53046353 on Build GRCh38
rs958752351 Chr.14: 53046147-53046147 on Build GRCh38
rs1029919665 Chr.14: 53046049-53046049 on Build GRCh38
rs780028186 Chr.14: 53046081-53046081 on Build GRCh38
rs772305129 Chr.14: 53046426-53046432 on Build GRCh38
rs992922552 Chr.14: 53046158-53046158 on Build GRCh38
rs1005541219 Chr.14: 53046075-53046075 on Build GRCh38
rs552408794 Chr.14: 53046056-53046056 on Build GRCh38
rs191717543 Chr.14: 53046120-53046120 on Build GRCh38
rs963640627 Chr.14: 53046351-53046351 on Build GRCh38
rs923910318 Chr.14: 53046213-53046213 on Build GRCh38
rs776979885 Chr.14: 53046303-53046303 on Build GRCh38
rs143125034 Chr.14: 53046119-53046119 on Build GRCh38
rs967770861 Chr.14: 53046385-53046385 on Build GRCh38
rs555074615 Chr.14: 53046240-53046240 on Build GRCh38
rs545697453 Chr.14: 53046145-53046145 on Build GRCh38
rs199844954 Chr.14: 53046113-53046113 on Build GRCh38
rs569116402 Chr.14: 53046087-53046087 on Build GRCh38
rs570830667 Chr.14: 53046357-53046357 on Build GRCh38
rs770727390 Chr.14: 53046335-53046335 on Build GRCh38
rs762010889 Chr.14: 53046321-53046321 on Build GRCh38
rs936602266 Chr.14: 53046233-53046233 on Build GRCh38
rs899233237 Chr.14: 53046216-53046216 on Build GRCh38
rs1034485391 Chr.14: 53046354-53046354 on Build GRCh38
rs1036405481 Chr.14: 53046160-53046160 on Build GRCh38
rs369319368 Chr.14: 53046325-53046325 on Build GRCh38
rs556538855 Chr.14: 53046417-53046417 on Build GRCh38
rs374591946 Chr.14: 53046284-53046284 on Build GRCh38
rs917216098 Chr.14: 53046179-53046179 on Build GRCh38
rs973621852 Chr.14: 53046131-53046131 on Build GRCh38
rs148254741 Chr.14: 53046137-53046137 on Build GRCh38

View Full Product Details