Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55381468-55382015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG14
Entrez Gene ID
22863
Gene Name
autophagy related 14
Gene Aliases
ATG14L, BARKOR, KIAA0831
UniGene
Hs.414809
Gene Chromosome Location
Chr.14: 55366391-55411858 on Build GRCh38
Transcripts
NM_014924.4 XM_011536563.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
698325 c.868C>T p.Q290* Chr.14: 55381971-55381971 on Build GRCh38
4511386 c.866C>T p.T289I Chr.14: 55381973-55381973 on Build GRCh38
398926 c.858G>C p.K286N Chr.14: 55381981-55381981 on Build GRCh38
4953153 c.863C>T p.T288I Chr.14: 55381976-55381976 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753869673 Chr.14: 55381941-55381941 on Build GRCh38
rs999256528 Chr.14: 55381698-55381698 on Build GRCh38
rs917735195 Chr.14: 55381503-55381503 on Build GRCh38
rs775946589 Chr.14: 55381973-55381973 on Build GRCh38
rs770504770 Chr.14: 55381856-55381856 on Build GRCh38
rs115525601 Chr.14: 55381933-55381933 on Build GRCh38
rs903392106 Chr.14: 55381975-55381975 on Build GRCh38
rs954821673 Chr.14: 55381778-55381778 on Build GRCh38
rs899192188 Chr.14: 55381563-55381563 on Build GRCh38
rs553079368 Chr.14: 55381744-55381744 on Build GRCh38
rs752796116 Chr.14: 55381859-55381859 on Build GRCh38
rs547691327 Chr.14: 55381625-55381625 on Build GRCh38
rs60796919 Chr.14: 55381527-55381531 on Build GRCh38
rs113415883 Chr.14: 55381652-55381652 on Build GRCh38
rs1031956369 Chr.14: 55381700-55381700 on Build GRCh38
rs34996227 Chr.14: 55381610-55381611 on Build GRCh38
rs771843903 Chr.14: 55381973-55381975 on Build GRCh38
rs765127252 Chr.14: 55381656-55381656 on Build GRCh38
rs1010366817 Chr.14: 55381654-55381654 on Build GRCh38
rs564896129 Chr.14: 55381577-55381577 on Build GRCh38
rs761031047 Chr.14: 55381984-55381984 on Build GRCh38
rs149789789 Chr.14: 55381743-55381743 on Build GRCh38
rs769560096 Chr.14: 55381931-55381931 on Build GRCh38
rs771299743 Chr.14: 55381773-55381774 on Build GRCh38
rs371762468 Chr.14: 55381659-55381659 on Build GRCh38
rs186606732 Chr.14: 55381663-55381663 on Build GRCh38
rs943715177 Chr.14: 55381901-55381901 on Build GRCh38
rs755003166 Chr.14: 55381690-55381690 on Build GRCh38
rs142760782 Chr.14: 55381764-55381765 on Build GRCh38
rs375978125 Chr.14: 55381930-55381930 on Build GRCh38
rs373265085 Chr.14: 55381842-55381842 on Build GRCh38
rs759951442 Chr.14: 55381952-55381952 on Build GRCh38
rs181705231 Chr.14: 55381502-55381502 on Build GRCh38
rs201771367 Chr.14: 55381558-55381561 on Build GRCh38
rs946058 Chr.14: 55381574-55381574 on Build GRCh38
rs768658830 Chr.14: 55381926-55381926 on Build GRCh38
rs773138557 Chr.14: 55381920-55381923 on Build GRCh38
rs973496499 Chr.14: 55381887-55381887 on Build GRCh38
rs1040764747 Chr.14: 55381929-55381929 on Build GRCh38
rs532383026 Chr.14: 55381578-55381578 on Build GRCh38
rs763735051 Chr.14: 55381549-55381549 on Build GRCh38
rs189444741 Chr.14: 55381928-55381928 on Build GRCh38
rs772528327 Chr.14: 55381953-55381953 on Build GRCh38
rs536713516 Chr.14: 55381833-55381833 on Build GRCh38
rs750458523 Chr.14: 55381513-55381516 on Build GRCh38
rs992900518 Chr.14: 55381725-55381725 on Build GRCh38
rs1037686524 Chr.14: 55381546-55381546 on Build GRCh38
rs746877341 Chr.14: 55381923-55381923 on Build GRCh38
rs74324636 Chr.14: 55381535-55381535 on Build GRCh38
rs80089534 Chr.14: 55381748-55381748 on Build GRCh38
rs943782229 Chr.14: 55381551-55381551 on Build GRCh38
rs1002223797 Chr.14: 55381681-55381681 on Build GRCh38
rs777535440 Chr.14: 55381703-55381704 on Build GRCh38
rs369781356 Chr.14: 55381523-55381527 on Build GRCh38
rs1041852303 Chr.14: 55381949-55381949 on Build GRCh38
rs530859878 Chr.14: 55381543-55381543 on Build GRCh38
rs919790623 Chr.14: 55381765-55381765 on Build GRCh38
rs775241579 Chr.14: 55381517-55381517 on Build GRCh38
rs558655835 Chr.14: 55381906-55381906 on Build GRCh38
rs1028912923 Chr.14: 55381775-55381775 on Build GRCh38
rs569655579 Chr.14: 55381799-55381799 on Build GRCh38

View Full Product Details