Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74288622-74289164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD4
Entrez Gene ID
5826
Gene Name
ATP binding cassette subfamily D member 4
Gene Aliases
ABC41, EST352188, MAHCJ, P70R, P79R, PMP69, PXMP1L
UniGene
Hs.94395
Gene Chromosome Location
Chr.14: 74285277-74303064 on Build GRCh38
Transcripts
NM_005050.3 NR_003256.2 XM_005267939.3 XM_005267940.3 XM_005267941.4 XM_005267942.4 XM_005267946.3 XM_005267949.3 XM_006720223.2 XM_011537041.2 XM_017021531.1 XM_017021532.1 XM_017021533.1 XM_017021534.1 XM_017021535.1 XM_017021536.1 XM_017021537.1 XM_017021538.1 XM_017021539.1 XM_017021540.1 XM_017021541.1 XM_017021542.1 XM_017021543.1 XM_017021544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7353668 c.1465G>A p.D489N Chr.14: 74288757-74288757 on Build GRCh38
2137625 c.1488C>A p.F496L Chr.14: 74288734-74288734 on Build GRCh38
7353671 c.1498G>A p.A500T Chr.14: 74288724-74288724 on Build GRCh38
6590627 c.1463C>T p.A488V Chr.14: 74288759-74288759 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs139901585 Chr.14: 74288757-74288757 on Build GRCh38
rs764928836 Chr.14: 74288787-74288787 on Build GRCh38
rs763049476 Chr.14: 74288651-74288651 on Build GRCh38
rs777790540 Chr.14: 74288691-74288691 on Build GRCh38
rs796147224 Chr.14: 74288872-74288872 on Build GRCh38
rs752208815 Chr.14: 74288686-74288686 on Build GRCh38
rs944872785 Chr.14: 74288664-74288664 on Build GRCh38
rs905048987 Chr.14: 74289036-74289036 on Build GRCh38
rs746065923 Chr.14: 74288642-74288642 on Build GRCh38
rs940709344 Chr.14: 74288991-74288991 on Build GRCh38
rs751538711 Chr.14: 74288694-74288694 on Build GRCh38
rs761126304 Chr.14: 74288782-74288782 on Build GRCh38
rs144816144 Chr.14: 74288953-74288953 on Build GRCh38
rs776602222 Chr.14: 74288999-74288999 on Build GRCh38
rs561519173 Chr.14: 74288727-74288727 on Build GRCh38
rs1056622684 Chr.14: 74288964-74288964 on Build GRCh38
rs557093129 Chr.14: 74289061-74289061 on Build GRCh38
rs946401990 Chr.14: 74288893-74288893 on Build GRCh38
rs386778837 Chr.14: 74288973-74288974 on Build GRCh38
rs757999682 Chr.14: 74288687-74288687 on Build GRCh38
rs887820438 Chr.14: 74288980-74288980 on Build GRCh38
rs775514847 Chr.14: 74288648-74288648 on Build GRCh38
rs369566429 Chr.14: 74288645-74288645 on Build GRCh38
rs757278296 Chr.14: 74288719-74288719 on Build GRCh38
rs1045364584 Chr.14: 74289005-74289005 on Build GRCh38
rs571380056 Chr.14: 74289011-74289011 on Build GRCh38
rs956381271 Chr.14: 74288890-74288890 on Build GRCh38
rs557254765 Chr.14: 74288653-74288653 on Build GRCh38
rs533175143 Chr.14: 74288879-74288879 on Build GRCh38
rs76925461 Chr.14: 74289111-74289111 on Build GRCh38
rs1019258473 Chr.14: 74289136-74289136 on Build GRCh38
rs763683625 Chr.14: 74288801-74288801 on Build GRCh38
rs879754242 Chr.14: 74289003-74289003 on Build GRCh38
rs952431326 Chr.14: 74288986-74288986 on Build GRCh38
rs866254045 Chr.14: 74288732-74288732 on Build GRCh38
rs985737967 Chr.14: 74289094-74289095 on Build GRCh38
rs772681992 Chr.14: 74288758-74288758 on Build GRCh38
rs373333360 Chr.14: 74288798-74288798 on Build GRCh38
rs972519546 Chr.14: 74288789-74288789 on Build GRCh38
rs865778997 Chr.14: 74288901-74288901 on Build GRCh38
rs549971933 Chr.14: 74288912-74288912 on Build GRCh38
rs369359552 Chr.14: 74288670-74288670 on Build GRCh38
rs1023456626 Chr.14: 74288852-74288852 on Build GRCh38
rs541820889 Chr.14: 74288671-74288671 on Build GRCh38
rs956454264 Chr.14: 74289095-74289095 on Build GRCh38
rs60498100 Chr.14: 74289130-74289138 on Build GRCh38
rs148595545 Chr.14: 74289002-74289002 on Build GRCh38
rs1013873103 Chr.14: 74288833-74288833 on Build GRCh38
rs1032023372 Chr.14: 74288880-74288880 on Build GRCh38
rs932035002 Chr.14: 74288688-74288688 on Build GRCh38
rs575145063 Chr.14: 74289112-74289120 on Build GRCh38
rs913509127 Chr.14: 74288934-74288934 on Build GRCh38
rs557542079 Chr.14: 74289060-74289060 on Build GRCh38
rs1013487369 Chr.14: 74289004-74289004 on Build GRCh38
rs772131218 Chr.14: 74288772-74288772 on Build GRCh38
rs60940187 Chr.14: 74288973-74288973 on Build GRCh38
rs922140142 Chr.14: 74288970-74288970 on Build GRCh38
rs146600870 Chr.14: 74288868-74288868 on Build GRCh38
rs768726077 Chr.14: 74288754-74288754 on Build GRCh38
rs376287005 Chr.14: 74288677-74288677 on Build GRCh38
rs751166310 Chr.14: 74288805-74288805 on Build GRCh38
rs773414987 Chr.14: 74288770-74288770 on Build GRCh38
rs150594251 Chr.14: 74288724-74288724 on Build GRCh38
rs572155878 Chr.14: 74288755-74288755 on Build GRCh38
rs749308023 Chr.14: 74288744-74288744 on Build GRCh38
rs867765583 Chr.14: 74288906-74288906 on Build GRCh38
rs564304996 Chr.14: 74288870-74288870 on Build GRCh38
rs12590962 Chr.14: 74288822-74288822 on Build GRCh38
rs573949742 Chr.14: 74288669-74288669 on Build GRCh38
rs1000621155 Chr.14: 74289037-74289037 on Build GRCh38
rs56200118 Chr.14: 74288938-74288938 on Build GRCh38
rs769102171 Chr.14: 74288652-74288652 on Build GRCh38
rs66758529 Chr.14: 74289134-74289138 on Build GRCh38
rs55737040 Chr.14: 74288929-74288929 on Build GRCh38
rs373129475 Chr.14: 74288674-74288674 on Build GRCh38
rs922407673 Chr.14: 74289023-74289023 on Build GRCh38
rs752405350 Chr.14: 74288794-74288794 on Build GRCh38
rs537251303 Chr.14: 74289015-74289015 on Build GRCh38
rs780274763 Chr.14: 74288737-74288737 on Build GRCh38
rs370984701 Chr.14: 74288746-74288746 on Build GRCh38
rs1009675463 Chr.14: 74289132-74289132 on Build GRCh38
rs55807426 Chr.14: 74288974-74288974 on Build GRCh38
rs762095272 Chr.14: 74288657-74288657 on Build GRCh38
rs757111675 Chr.14: 74288816-74288816 on Build GRCh38

View Full Product Details