Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102929349-102929853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN
Entrez Gene ID
81693
Gene Name
amnion associated transmembrane protein
Gene Aliases
PRO1028, amnionless
UniGene
Hs.534494
Gene Chromosome Location
Chr.14: 102922430-102930842 on Build GRCh38
Transcripts
NM_030943.3 XM_011537202.2 XM_011537203.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6387259 c.770T>C p.V257A Chr.14: 102929664-102929664 on Build GRCh38
6035612 c.773T>C p.L258S Chr.14: 102929667-102929667 on Build GRCh38
5727400 c.843+1G>A p.? Chr.14: 102929738-102929738 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368430211 Chr.14: 102929677-102929677 on Build GRCh38
rs374692212 Chr.14: 102929732-102929732 on Build GRCh38
rs916791081 Chr.14: 102929730-102929730 on Build GRCh38
rs756429503 Chr.14: 102929698-102929698 on Build GRCh38
rs369631445 Chr.14: 102929766-102929766 on Build GRCh38
rs774460041 Chr.14: 102929775-102929775 on Build GRCh38
rs767700967 Chr.14: 102929692-102929692 on Build GRCh38
rs933555566 Chr.14: 102929389-102929389 on Build GRCh38
rs919847262 Chr.14: 102929379-102929379 on Build GRCh38
rs1032554285 Chr.14: 102929506-102929506 on Build GRCh38
rs975146429 Chr.14: 102929370-102929370 on Build GRCh38
rs965147792 Chr.14: 102929612-102929612 on Build GRCh38
rs936786504 Chr.14: 102929760-102929760 on Build GRCh38
rs888359987 Chr.14: 102929772-102929772 on Build GRCh38
rs902671345 Chr.14: 102929460-102929460 on Build GRCh38
rs562891794 Chr.14: 102929548-102929548 on Build GRCh38
rs200712130 Chr.14: 102929699-102929699 on Build GRCh38
rs190222721 Chr.14: 102929667-102929667 on Build GRCh38
rs763110632 Chr.14: 102929674-102929674 on Build GRCh38
rs760534051 Chr.14: 102929636-102929636 on Build GRCh38
rs991168089 Chr.14: 102929724-102929724 on Build GRCh38
rs369361298 Chr.14: 102929748-102929748 on Build GRCh38
rs115489959 Chr.14: 102929781-102929781 on Build GRCh38
rs386834174 Chr.14: 102929439-102929439 on Build GRCh38
rs765147150 Chr.14: 102929705-102929705 on Build GRCh38
rs1054734003 Chr.14: 102929403-102929403 on Build GRCh38
rs756662668 Chr.14: 102929488-102929488 on Build GRCh38
rs747751960 Chr.14: 102929552-102929552 on Build GRCh38
rs12883949 Chr.14: 102929654-102929654 on Build GRCh38
rs902265398 Chr.14: 102929757-102929757 on Build GRCh38
rs386834178 Chr.14: 102929655-102929655 on Build GRCh38
rs767790603 Chr.14: 102929782-102929782 on Build GRCh38
rs972154786 Chr.14: 102929634-102929634 on Build GRCh38
rs140902778 Chr.14: 102929749-102929749 on Build GRCh38
rs143303478 Chr.14: 102929751-102929751 on Build GRCh38
rs571160852 Chr.14: 102929708-102929708 on Build GRCh38
rs372304599 Chr.14: 102929694-102929694 on Build GRCh38
rs924209069 Chr.14: 102929413-102929413 on Build GRCh38
rs371426960 Chr.14: 102929729-102929729 on Build GRCh38
rs386834176 Chr.14: 102929477-102929477 on Build GRCh38
rs1005458364 Chr.14: 102929777-102929777 on Build GRCh38
rs916279736 Chr.14: 102929478-102929478 on Build GRCh38
rs1046934064 Chr.14: 102929508-102929508 on Build GRCh38
rs370572037 Chr.14: 102929613-102929613 on Build GRCh38
rs947871053 Chr.14: 102929479-102929479 on Build GRCh38
rs896201571 Chr.14: 102929404-102929404 on Build GRCh38
rs778609113 Chr.14: 102929722-102929722 on Build GRCh38
rs1015463312 Chr.14: 102929652-102929652 on Build GRCh38
rs757507992 Chr.14: 102929691-102929691 on Build GRCh38
rs983550174 Chr.14: 102929632-102929632 on Build GRCh38
rs572852233 Chr.14: 102929378-102929378 on Build GRCh38
rs767981046 Chr.14: 102929544-102929544 on Build GRCh38
rs899688048 Chr.14: 102929823-102929823 on Build GRCh38
rs200393580 Chr.14: 102929669-102929669 on Build GRCh38
rs996147843 Chr.14: 102929551-102929551 on Build GRCh38
rs746199829 Chr.14: 102929467-102929467 on Build GRCh38
rs1031037361 Chr.14: 102929787-102929787 on Build GRCh38
rs954905847 Chr.14: 102929624-102929624 on Build GRCh38
rs780412383 Chr.14: 102929499-102929499 on Build GRCh38
rs146499374 Chr.14: 102929723-102929723 on Build GRCh38
rs386834177 Chr.14: 102929518-102929518 on Build GRCh38
rs1042558809 Chr.14: 102929455-102929455 on Build GRCh38
rs939893197 Chr.14: 102929376-102929376 on Build GRCh38
rs577843988 Chr.14: 102929481-102929481 on Build GRCh38
rs1053967680 Chr.14: 102929461-102929461 on Build GRCh38
rs746808374 Chr.14: 102929736-102929736 on Build GRCh38
rs76832951 Chr.14: 102929675-102929675 on Build GRCh38
rs531701170 Chr.14: 102929651-102929651 on Build GRCh38
rs1018102384 Chr.14: 102929590-102929590 on Build GRCh38
rs546245909 Chr.14: 102929511-102929511 on Build GRCh38
rs1030348310 Chr.14: 102929573-102929573 on Build GRCh38
rs771820682 Chr.14: 102929585-102929585 on Build GRCh38
rs890460212 Chr.14: 102929830-102929830 on Build GRCh38
rs569327877 Chr.14: 102929735-102929735 on Build GRCh38
rs949145775 Chr.14: 102929414-102929414 on Build GRCh38
rs781494385 Chr.14: 102929380-102929380 on Build GRCh38
rs386834175 Chr.14: 102929459-102929506 on Build GRCh38

View Full Product Details