Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59116950-59117472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB2
Entrez Gene ID
9133
Gene Name
cyclin B2
Gene Aliases
HsT17299
UniGene
Hs.194698
Gene Chromosome Location
Chr.15: 59105085-59125045 on Build GRCh38
Transcripts
NM_004701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
470893 c.929C>T p.A310V Chr.15: 59117322-59117322 on Build GRCh38
6400604 c.941G>A p.C314Y Chr.15: 59117334-59117334 on Build GRCh38
4578377 c.847C>T p.Q283* Chr.15: 59117240-59117240 on Build GRCh38
3936797 c.918T>C p.S306S Chr.15: 59117311-59117311 on Build GRCh38
3969180 c.850C>T p.H284Y Chr.15: 59117243-59117243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047933502 Chr.15: 59117378-59117378 on Build GRCh38
rs776783136 Chr.15: 59117250-59117250 on Build GRCh38
rs962792738 Chr.15: 59117091-59117091 on Build GRCh38
rs1003858087 Chr.15: 59117397-59117397 on Build GRCh38
rs886811673 Chr.15: 59117071-59117071 on Build GRCh38
rs372870051 Chr.15: 59117342-59117342 on Build GRCh38
rs769739976 Chr.15: 59117383-59117383 on Build GRCh38
rs773543738 Chr.15: 59117278-59117278 on Build GRCh38
rs990575775 Chr.15: 59117231-59117231 on Build GRCh38
rs142849994 Chr.15: 59117356-59117356 on Build GRCh38
rs774061171 Chr.15: 59117282-59117282 on Build GRCh38
rs1016643409 Chr.15: 59117086-59117086 on Build GRCh38
rs774900402 Chr.15: 59117371-59117371 on Build GRCh38
rs556218585 Chr.15: 59116994-59116994 on Build GRCh38
rs774714734 Chr.15: 59117393-59117393 on Build GRCh38
rs376477102 Chr.15: 59117419-59117419 on Build GRCh38
rs769923427 Chr.15: 59117208-59117208 on Build GRCh38
rs934640466 Chr.15: 59117216-59117216 on Build GRCh38
rs767021893 Chr.15: 59117284-59117284 on Build GRCh38
rs545643959 Chr.15: 59117054-59117054 on Build GRCh38
rs762302609 Chr.15: 59117416-59117416 on Build GRCh38
rs1004295416 Chr.15: 59117083-59117083 on Build GRCh38
rs757092056 Chr.15: 59117209-59117209 on Build GRCh38
rs199501636 Chr.15: 59117207-59117207 on Build GRCh38
rs767219342 Chr.15: 59117399-59117399 on Build GRCh38
rs376602782 Chr.15: 59117191-59117191 on Build GRCh38
rs748536373 Chr.15: 59117247-59117247 on Build GRCh38
rs746892887 Chr.15: 59117058-59117058 on Build GRCh38
rs28376133 Chr.15: 59117296-59117296 on Build GRCh38
rs777529737 Chr.15: 59117204-59117204 on Build GRCh38
rs778836336 Chr.15: 59117338-59117338 on Build GRCh38
rs144150396 Chr.15: 59116999-59116999 on Build GRCh38
rs141427555 Chr.15: 59117251-59117251 on Build GRCh38
rs948891999 Chr.15: 59116983-59116983 on Build GRCh38
rs745599085 Chr.15: 59117351-59117351 on Build GRCh38
rs201814723 Chr.15: 59117295-59117295 on Build GRCh38
rs28383543 Chr.15: 59116979-59116979 on Build GRCh38
rs201333257 Chr.15: 59117407-59117407 on Build GRCh38
rs370315049 Chr.15: 59117422-59117422 on Build GRCh38
rs750954670 Chr.15: 59117415-59117417 on Build GRCh38
rs553648682 Chr.15: 59117019-59117019 on Build GRCh38
rs1028188843 Chr.15: 59117160-59117160 on Build GRCh38
rs547886264 Chr.15: 59117281-59117281 on Build GRCh38
rs975241884 Chr.15: 59117154-59117154 on Build GRCh38
rs573500710 Chr.15: 59117418-59117418 on Build GRCh38
rs576020311 Chr.15: 59117143-59117143 on Build GRCh38
rs774979748 Chr.15: 59117404-59117404 on Build GRCh38
rs368620705 Chr.15: 59117292-59117292 on Build GRCh38
rs747229243 Chr.15: 59117245-59117245 on Build GRCh38
rs568061623 Chr.15: 59117439-59117439 on Build GRCh38
rs986754173 Chr.15: 59117167-59117167 on Build GRCh38
rs751191379 Chr.15: 59117183-59117183 on Build GRCh38
rs1044971485 Chr.15: 59116993-59116993 on Build GRCh38
rs752279669 Chr.15: 59117286-59117286 on Build GRCh38
rs780463202 Chr.15: 59117219-59117219 on Build GRCh38
rs11548101 Chr.15: 59117326-59117326 on Build GRCh38
rs1003105295 Chr.15: 59117023-59117023 on Build GRCh38
rs766046275 Chr.15: 59117180-59117180 on Build GRCh38
rs779993602 Chr.15: 59117272-59117272 on Build GRCh38
rs773532612 Chr.15: 59117358-59117358 on Build GRCh38
rs148681047 Chr.15: 59117048-59117048 on Build GRCh38
rs757184223 Chr.15: 59117314-59117314 on Build GRCh38
rs537843302 Chr.15: 59117380-59117380 on Build GRCh38
rs564047294 Chr.15: 59117055-59117055 on Build GRCh38
rs768741504 Chr.15: 59117364-59117364 on Build GRCh38
rs150819267 Chr.15: 59117394-59117394 on Build GRCh38
rs183917477 Chr.15: 59117005-59117005 on Build GRCh38
rs368459438 Chr.15: 59117242-59117242 on Build GRCh38
rs552092989 Chr.15: 59117384-59117384 on Build GRCh38
rs754523706 Chr.15: 59117189-59117189 on Build GRCh38
rs771346685 Chr.15: 59117390-59117390 on Build GRCh38
rs368038064 Chr.15: 59117190-59117190 on Build GRCh38
rs763267453 Chr.15: 59117178-59117178 on Build GRCh38
rs768913561 Chr.15: 59117246-59117246 on Build GRCh38
rs763357025 Chr.15: 59117280-59117280 on Build GRCh38
rs955255998 Chr.15: 59117163-59117163 on Build GRCh38

View Full Product Details