Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 88512720-88513229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DET1
Entrez Gene ID
55070
Gene Name
de-etiolated homolog 1 (Arabidopsis)
Gene Aliases
-
UniGene
Hs.655064
Gene Chromosome Location
Chr.15: 88501924-88546703 on Build GRCh38
Transcripts
NM_001144074.2 NM_001321594.1 NM_001321596.1 NM_017996.4 NR_026645.2 NR_135739.1 NR_135740.1 NR_135741.1 NR_135742.1 NR_135743.1 NR_135744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3401987 c.1669C>T p.R557* Chr.15: 88512968-88512968 on Build GRCh38
6143213 c.1633A>G p.R545G Chr.15: 88513004-88513004 on Build GRCh38
5850009 c.1540G>A p.A514T Chr.15: 88513097-88513097 on Build GRCh38
5850008 c.1603_1604CC>TT p.P535F Chr.15: 88513033-88513034 on Build GRCh38
6078124 c.1556G>T p.R519L Chr.15: 88513081-88513081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010970178 Chr.15: 88512749-88512749 on Build GRCh38
rs199619310 Chr.15: 88512966-88512966 on Build GRCh38
rs766698359 Chr.15: 88513028-88513028 on Build GRCh38
rs891230869 Chr.15: 88513091-88513091 on Build GRCh38
rs202049856 Chr.15: 88512970-88512970 on Build GRCh38
rs373057397 Chr.15: 88512955-88512955 on Build GRCh38
rs368418254 Chr.15: 88513082-88513082 on Build GRCh38
rs771173086 Chr.15: 88512848-88512848 on Build GRCh38
rs184689884 Chr.15: 88513173-88513173 on Build GRCh38
rs775183903 Chr.15: 88512967-88512967 on Build GRCh38
rs376592225 Chr.15: 88513058-88513058 on Build GRCh38
rs771667500 Chr.15: 88513108-88513108 on Build GRCh38
rs577478682 Chr.15: 88512752-88512752 on Build GRCh38
rs1022229067 Chr.15: 88512812-88512812 on Build GRCh38
rs184099925 Chr.15: 88512828-88512828 on Build GRCh38
rs967952493 Chr.15: 88512815-88512815 on Build GRCh38
rs758451300 Chr.15: 88513079-88513079 on Build GRCh38
rs944435988 Chr.15: 88512918-88512918 on Build GRCh38
rs777263695 Chr.15: 88513130-88513130 on Build GRCh38
rs190664811 Chr.15: 88512851-88512851 on Build GRCh38
rs754051413 Chr.15: 88512927-88512927 on Build GRCh38
rs766781407 Chr.15: 88513127-88513127 on Build GRCh38
rs778904277 Chr.15: 88512936-88512936 on Build GRCh38
rs368867850 Chr.15: 88513122-88513122 on Build GRCh38
rs780304332 Chr.15: 88512925-88512925 on Build GRCh38
rs962311970 Chr.15: 88512774-88512774 on Build GRCh38
rs370366433 Chr.15: 88512916-88512916 on Build GRCh38
rs774211122 Chr.15: 88512969-88512969 on Build GRCh38
rs768608942 Chr.15: 88513095-88513095 on Build GRCh38
rs761465043 Chr.15: 88512909-88512909 on Build GRCh38
rs753629207 Chr.15: 88512770-88512770 on Build GRCh38
rs751607822 Chr.15: 88513169-88513169 on Build GRCh38
rs2289415 Chr.15: 88512772-88512772 on Build GRCh38
rs1040447180 Chr.15: 88512960-88512960 on Build GRCh38
rs779843792 Chr.15: 88513086-88513086 on Build GRCh38
rs13835 Chr.15: 88512809-88512809 on Build GRCh38
rs761619650 Chr.15: 88512958-88512958 on Build GRCh38
rs764097765 Chr.15: 88513168-88513168 on Build GRCh38
rs754166318 Chr.15: 88513041-88513041 on Build GRCh38
rs752868372 Chr.15: 88513150-88513150 on Build GRCh38
rs746402502 Chr.15: 88513123-88513123 on Build GRCh38
rs182003477 Chr.15: 88512968-88512968 on Build GRCh38
rs200501194 Chr.15: 88512954-88512954 on Build GRCh38
rs1030974973 Chr.15: 88512864-88512864 on Build GRCh38
rs969470894 Chr.15: 88512767-88512767 on Build GRCh38
rs370620890 Chr.15: 88513132-88513132 on Build GRCh38
rs533617357 Chr.15: 88513180-88513180 on Build GRCh38
rs368557263 Chr.15: 88513133-88513133 on Build GRCh38
rs774121152 Chr.15: 88513097-88513097 on Build GRCh38
rs374410511 Chr.15: 88513073-88513073 on Build GRCh38
rs1037133559 Chr.15: 88513034-88513034 on Build GRCh38
rs767182017 Chr.15: 88513011-88513011 on Build GRCh38
rs750889030 Chr.15: 88513016-88513016 on Build GRCh38
rs756475603 Chr.15: 88512917-88512917 on Build GRCh38
rs985936634 Chr.15: 88512850-88512850 on Build GRCh38
rs772601304 Chr.15: 88513110-88513110 on Build GRCh38
rs148455500 Chr.15: 88512827-88512827 on Build GRCh38
rs761116751 Chr.15: 88513020-88513020 on Build GRCh38
rs141866692 Chr.15: 88512750-88512750 on Build GRCh38
rs757902237 Chr.15: 88513057-88513057 on Build GRCh38
rs752187164 Chr.15: 88513026-88513026 on Build GRCh38
rs751121615 Chr.15: 88512901-88512901 on Build GRCh38
rs745843188 Chr.15: 88512956-88512956 on Build GRCh38
rs765639184 Chr.15: 88513144-88513144 on Build GRCh38
rs978518533 Chr.15: 88512818-88512818 on Build GRCh38
rs905280038 Chr.15: 88513146-88513146 on Build GRCh38
rs972000751 Chr.15: 88512777-88512777 on Build GRCh38
rs962060710 Chr.15: 88513181-88513181 on Build GRCh38
rs777852532 Chr.15: 88513081-88513081 on Build GRCh38
rs757705007 Chr.15: 88513198-88513198 on Build GRCh38
rs1023725879 Chr.15: 88513129-88513129 on Build GRCh38
rs758654675 Chr.15: 88513151-88513151 on Build GRCh38
rs75714837 Chr.15: 88513061-88513061 on Build GRCh38
rs146230552 Chr.15: 88512769-88512769 on Build GRCh38
rs546165634 Chr.15: 88512857-88512857 on Build GRCh38
rs548006693 Chr.15: 88513092-88513092 on Build GRCh38
rs749294534 Chr.15: 88513183-88513183 on Build GRCh38
rs548274685 Chr.15: 88512761-88512761 on Build GRCh38
rs993435573 Chr.15: 88513197-88513197 on Build GRCh38

View Full Product Details