Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90619495-90619894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC3
Entrez Gene ID
64784
Gene Name
CREB regulated transcription coactivator 3
Gene Aliases
TORC-3, TORC3
UniGene
Hs.567572
Gene Chromosome Location
Chr.15: 90529886-90645345 on Build GRCh38
Transcripts
NM_001042574.2 NM_022769.4 XM_005254968.2 XM_011521906.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2140111 c.718C>T p.R240C Chr.15: 90619759-90619759 on Build GRCh38
702124 c.724C>A p.R242R Chr.15: 90619765-90619765 on Build GRCh38
6442929 c.707C>T p.S236F Chr.15: 90619748-90619748 on Build GRCh38
1708549 c.728_729CC>AT p.S243Y Chr.15: 90619769-90619770 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768161924 Chr.15: 90619823-90619823 on Build GRCh38
rs202181818 Chr.15: 90619696-90619696 on Build GRCh38
rs746277982 Chr.15: 90619628-90619628 on Build GRCh38
rs756453361 Chr.15: 90619748-90619748 on Build GRCh38
rs778977437 Chr.15: 90619759-90619759 on Build GRCh38
rs878948487 Chr.15: 90619565-90619565 on Build GRCh38
rs766761894 Chr.15: 90619831-90619831 on Build GRCh38
rs1029976234 Chr.15: 90619745-90619745 on Build GRCh38
rs770884806 Chr.15: 90619764-90619764 on Build GRCh38
rs1009051962 Chr.15: 90619552-90619552 on Build GRCh38
rs988676351 Chr.15: 90619567-90619567 on Build GRCh38
rs539860842 Chr.15: 90619644-90619644 on Build GRCh38
rs914358040 Chr.15: 90619579-90619579 on Build GRCh38
rs564641028 Chr.15: 90619566-90619566 on Build GRCh38
rs577544474 Chr.15: 90619867-90619867 on Build GRCh38
rs929518444 Chr.15: 90619672-90619672 on Build GRCh38
rs780188941 Chr.15: 90619750-90619750 on Build GRCh38
rs772925307 Chr.15: 90619766-90619766 on Build GRCh38
rs1010532197 Chr.15: 90619852-90619852 on Build GRCh38
rs754454231 Chr.15: 90619695-90619695 on Build GRCh38
rs752174201 Chr.15: 90619711-90619711 on Build GRCh38
rs1030227899 Chr.15: 90619533-90619533 on Build GRCh38
rs887902371 Chr.15: 90619695-90619695 on Build GRCh38
rs997194436 Chr.15: 90619532-90619532 on Build GRCh38
rs558209233 Chr.15: 90619690-90619690 on Build GRCh38
rs773156723 Chr.15: 90619603-90619603 on Build GRCh38
rs144806135 Chr.15: 90619632-90619632 on Build GRCh38
rs900647267 Chr.15: 90619580-90619580 on Build GRCh38
rs745696186 Chr.15: 90619787-90619787 on Build GRCh38
rs763785720 Chr.15: 90619812-90619812 on Build GRCh38
rs956129858 Chr.15: 90619539-90619539 on Build GRCh38
rs376110475 Chr.15: 90619760-90619760 on Build GRCh38
rs763689426 Chr.15: 90619709-90619709 on Build GRCh38
rs1056372602 Chr.15: 90619645-90619645 on Build GRCh38
rs200839301 Chr.15: 90619741-90619741 on Build GRCh38
rs540837583 Chr.15: 90619821-90619821 on Build GRCh38
rs117878260 Chr.15: 90619837-90619837 on Build GRCh38
rs548079476 Chr.15: 90619629-90619629 on Build GRCh38
rs774004767 Chr.15: 90619813-90619813 on Build GRCh38
rs114667786 Chr.15: 90619708-90619708 on Build GRCh38
rs200123845 Chr.15: 90619691-90619691 on Build GRCh38
rs755197230 Chr.15: 90619839-90619839 on Build GRCh38
rs375418782 Chr.15: 90619659-90619659 on Build GRCh38
rs750921748 Chr.15: 90619827-90619827 on Build GRCh38
rs762657004 Chr.15: 90619810-90619810 on Build GRCh38
rs756476080 Chr.15: 90619829-90619829 on Build GRCh38
rs1033254452 Chr.15: 90619607-90619607 on Build GRCh38
rs755107523 Chr.15: 90619757-90619757 on Build GRCh38
rs202128462 Chr.15: 90619798-90619798 on Build GRCh38
rs752061307 Chr.15: 90619806-90619806 on Build GRCh38
rs148780728 Chr.15: 90619623-90619623 on Build GRCh38
rs918186746 Chr.15: 90619671-90619671 on Build GRCh38
rs768072100 Chr.15: 90619726-90619726 on Build GRCh38
rs147910113 Chr.15: 90619713-90619713 on Build GRCh38

View Full Product Details