Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99997303-99997804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS17
Entrez Gene ID
170691
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 17
Gene Aliases
-
UniGene
Hs.513200
Gene Chromosome Location
Chr.15: 99971438-100342924 on Build GRCh38
Transcripts
NM_139057.3 XM_005254872.3 XM_011521312.2 XM_017021973.1 XM_017021974.1 XM_017021975.1 XM_017021976.1 XM_017021978.1 XM_017021979.1 XM_017021980.1 XM_017021982.1 XM_017021983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1740201 c.2650C>T p.R884W Chr.15: 99997531-99997531 on Build GRCh38
1244325 c.2683G>A p.G895S Chr.15: 99997498-99997498 on Build GRCh38
6594006 c.2606C>T p.P869L Chr.15: 99997575-99997575 on Build GRCh38
6222776 c.2726G>A p.R909Q Chr.15: 99997455-99997455 on Build GRCh38
5774679 c.2702G>A p.R901Q Chr.15: 99997479-99997479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563938783 Chr.15: 99997544-99997544 on Build GRCh38
rs375519462 Chr.15: 99997608-99997608 on Build GRCh38
rs773881815 Chr.15: 99997633-99997633 on Build GRCh38
rs1014151316 Chr.15: 99997462-99997462 on Build GRCh38
rs888313242 Chr.15: 99997409-99997409 on Build GRCh38
rs763500860 Chr.15: 99997511-99997511 on Build GRCh38
rs886050974 Chr.15: 99997450-99997450 on Build GRCh38
rs982823860 Chr.15: 99997666-99997666 on Build GRCh38
rs770811586 Chr.15: 99997338-99997338 on Build GRCh38
rs747817058 Chr.15: 99997428-99997428 on Build GRCh38
rs545940908 Chr.15: 99997505-99997505 on Build GRCh38
rs780661268 Chr.15: 99997410-99997410 on Build GRCh38
rs780077915 Chr.15: 99997556-99997556 on Build GRCh38
rs977439848 Chr.15: 99997660-99997660 on Build GRCh38
rs748706912 Chr.15: 99997729-99997729 on Build GRCh38
rs761847274 Chr.15: 99997334-99997334 on Build GRCh38
rs756721876 Chr.15: 99997401-99997401 on Build GRCh38
rs776589136 Chr.15: 99997446-99997446 on Build GRCh38
rs1000211796 Chr.15: 99997359-99997359 on Build GRCh38
rs371958509 Chr.15: 99997342-99997342 on Build GRCh38
rs747049463 Chr.15: 99997620-99997620 on Build GRCh38
rs746767578 Chr.15: 99997443-99997443 on Build GRCh38
rs765738277 Chr.15: 99997499-99997499 on Build GRCh38
rs780979498 Chr.15: 99997541-99997541 on Build GRCh38
rs1041035340 Chr.15: 99997331-99997331 on Build GRCh38
rs140577767 Chr.15: 99997583-99997583 on Build GRCh38
rs779560660 Chr.15: 99997662-99997662 on Build GRCh38
rs754451095 Chr.15: 99997458-99997458 on Build GRCh38
rs917184116 Chr.15: 99997644-99997644 on Build GRCh38
rs775650700 Chr.15: 99997624-99997624 on Build GRCh38
rs749156998 Chr.15: 99997557-99997557 on Build GRCh38
rs754802167 Chr.15: 99997616-99997616 on Build GRCh38
rs756837804 Chr.15: 99997531-99997531 on Build GRCh38
rs777524361 Chr.15: 99997475-99997475 on Build GRCh38
rs766914279 Chr.15: 99997490-99997490 on Build GRCh38
rs765499241 Chr.15: 99997596-99997596 on Build GRCh38
rs949857831 Chr.15: 99997749-99997749 on Build GRCh38
rs112856607 Chr.15: 99997645-99997645 on Build GRCh38
rs369460266 Chr.15: 99997622-99997622 on Build GRCh38
rs773418364 Chr.15: 99997559-99997559 on Build GRCh38
rs762081124 Chr.15: 99997368-99997368 on Build GRCh38
rs757978096 Chr.15: 99997507-99997507 on Build GRCh38
rs371076042 Chr.15: 99997335-99997335 on Build GRCh38
rs766989655 Chr.15: 99997637-99997637 on Build GRCh38
rs202079882 Chr.15: 99997605-99997605 on Build GRCh38
rs753876970 Chr.15: 99997615-99997615 on Build GRCh38
rs761476488 Chr.15: 99997601-99997601 on Build GRCh38
rs373083345 Chr.15: 99997482-99997482 on Build GRCh38
rs551904169 Chr.15: 99997734-99997734 on Build GRCh38
rs138772164 Chr.15: 99997451-99997451 on Build GRCh38
rs771685042 Chr.15: 99997437-99997437 on Build GRCh38
rs373132940 Chr.15: 99997516-99997516 on Build GRCh38
rs182846439 Chr.15: 99997489-99997489 on Build GRCh38
rs1008859063 Chr.15: 99997693-99997693 on Build GRCh38
rs866950737 Chr.15: 99997390-99997390 on Build GRCh38
rs779698939 Chr.15: 99997610-99997610 on Build GRCh38
rs780220308 Chr.15: 99997480-99997480 on Build GRCh38
rs767728567 Chr.15: 99997452-99997452 on Build GRCh38
rs776124364 Chr.15: 99997597-99997597 on Build GRCh38
rs751012843 Chr.15: 99997599-99997599 on Build GRCh38
rs569576215 Chr.15: 99997669-99997669 on Build GRCh38
rs767635581 Chr.15: 99997454-99997454 on Build GRCh38
rs760236403 Chr.15: 99997639-99997639 on Build GRCh38
rs777456566 Chr.15: 99997442-99997442 on Build GRCh38
rs113426361 Chr.15: 99997661-99997661 on Build GRCh38
rs922631314 Chr.15: 99997530-99997530 on Build GRCh38
rs567084956 Chr.15: 99997746-99997746 on Build GRCh38
rs766826340 Chr.15: 99997456-99997456 on Build GRCh38
rs758020667 Chr.15: 99997470-99997470 on Build GRCh38
rs756394096 Chr.15: 99997479-99997479 on Build GRCh38
rs368855635 Chr.15: 99997467-99997467 on Build GRCh38
rs1055650935 Chr.15: 99997589-99997589 on Build GRCh38
rs750044969 Chr.15: 99997607-99997607 on Build GRCh38
rs534270404 Chr.15: 99997750-99997750 on Build GRCh38
rs750733743 Chr.15: 99997400-99997400 on Build GRCh38
rs201644342 Chr.15: 99997455-99997455 on Build GRCh38
rs200054320 Chr.15: 99997355-99997355 on Build GRCh38
rs533467085 Chr.15: 99997698-99997698 on Build GRCh38
rs957131238 Chr.15: 99997641-99997641 on Build GRCh38
rs1055870061 Chr.15: 99997741-99997741 on Build GRCh38
rs540074131 Chr.15: 99997337-99997337 on Build GRCh38
rs768473488 Chr.15: 99997607-99997610 on Build GRCh38
rs550989841 Chr.15: 99997656-99997656 on Build GRCh38
rs143323522 Chr.15: 99997547-99997547 on Build GRCh38
rs746067605 Chr.15: 99997623-99997623 on Build GRCh38
rs376009030 Chr.15: 99997463-99997463 on Build GRCh38
rs569332582 Chr.15: 99997336-99997336 on Build GRCh38
rs751794220 Chr.15: 99997358-99997358 on Build GRCh38
rs778009417 Chr.15: 99997619-99997619 on Build GRCh38
rs377387249 Chr.15: 99997560-99997560 on Build GRCh38
rs755357010 Chr.15: 99997654-99997654 on Build GRCh38
rs748217171 Chr.15: 99997487-99997487 on Build GRCh38
rs151305163 Chr.15: 99997574-99997574 on Build GRCh38
rs572716880 Chr.15: 99997554-99997554 on Build GRCh38
rs770688711 Chr.15: 99997445-99997445 on Build GRCh38
rs763067738 Chr.15: 99997344-99997344 on Build GRCh38
rs369161179 Chr.15: 99997449-99997449 on Build GRCh38
rs778485857 Chr.15: 99997424-99997424 on Build GRCh38
rs769102110 Chr.15: 99997632-99997632 on Build GRCh38
rs952237627 Chr.15: 99997614-99997614 on Build GRCh38
rs760021311 Chr.15: 99997575-99997575 on Build GRCh38
rs149373184 Chr.15: 99997481-99997481 on Build GRCh38
rs903118054 Chr.15: 99997766-99997766 on Build GRCh38
rs767786439 Chr.15: 99997382-99997382 on Build GRCh38
rs202099735 Chr.15: 99997488-99997488 on Build GRCh38
rs774316306 Chr.15: 99997486-99997486 on Build GRCh38
rs373353462 Chr.15: 99997598-99997598 on Build GRCh38
rs200834418 Chr.15: 99997413-99997413 on Build GRCh38
rs760146001 Chr.15: 99997498-99997498 on Build GRCh38
rs139105066 Chr.15: 99997448-99997448 on Build GRCh38
rs778303777 Chr.15: 99997464-99997464 on Build GRCh38
rs891850525 Chr.15: 99997681-99997681 on Build GRCh38
rs376596927 Chr.15: 99997415-99997415 on Build GRCh38

View Full Product Details