Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 101234999-101235512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHSY1
Entrez Gene ID
22856
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
CHSY, CSS1, ChSy-1, TPBS
UniGene
Hs.110488
Gene Chromosome Location
Chr.15: 101175723-101251932 on Build GRCh38
Transcripts
NM_014918.4 XM_011521364.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629040 c.471C>T p.H157H Chr.15: 101235427-101235427 on Build GRCh38
1629463 c.402G>A p.V134V Chr.15: 101235496-101235496 on Build GRCh38
7289999 c.678C>A p.I226I Chr.15: 101235220-101235220 on Build GRCh38
6522416 c.590G>T p.G197V Chr.15: 101235308-101235308 on Build GRCh38
7279233 c.432G>A p.P144P Chr.15: 101235466-101235466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761321271 Chr.15: 101235235-101235235 on Build GRCh38
rs189132695 Chr.15: 101235025-101235025 on Build GRCh38
rs577569646 Chr.15: 101235466-101235466 on Build GRCh38
rs937156623 Chr.15: 101235474-101235474 on Build GRCh38
rs999890893 Chr.15: 101235352-101235352 on Build GRCh38
rs748806147 Chr.15: 101235225-101235225 on Build GRCh38
rs368009047 Chr.15: 101235188-101235188 on Build GRCh38
rs755444789 Chr.15: 101235418-101235418 on Build GRCh38
rs762321510 Chr.15: 101235231-101235232 on Build GRCh38
rs747150513 Chr.15: 101235060-101235060 on Build GRCh38
rs778545156 Chr.15: 101235419-101235419 on Build GRCh38
rs371453938 Chr.15: 101235354-101235354 on Build GRCh38
rs376691136 Chr.15: 101235479-101235479 on Build GRCh38
rs747903571 Chr.15: 101235345-101235345 on Build GRCh38
rs139411585 Chr.15: 101235241-101235241 on Build GRCh38
rs758494719 Chr.15: 101235201-101235201 on Build GRCh38
rs199985166 Chr.15: 101235075-101235075 on Build GRCh38
rs761799376 Chr.15: 101235467-101235467 on Build GRCh38
rs779085439 Chr.15: 101235040-101235040 on Build GRCh38
rs145660234 Chr.15: 101235142-101235142 on Build GRCh38
rs111801329 Chr.15: 101235428-101235428 on Build GRCh38
rs747414153 Chr.15: 101235074-101235074 on Build GRCh38
rs371508474 Chr.15: 101235195-101235195 on Build GRCh38
rs984872328 Chr.15: 101235437-101235437 on Build GRCh38
rs759237249 Chr.15: 101235070-101235074 on Build GRCh38
rs34886070 Chr.15: 101235032-101235033 on Build GRCh38
rs143062356 Chr.15: 101235379-101235379 on Build GRCh38
rs752265345 Chr.15: 101235111-101235111 on Build GRCh38
rs754504772 Chr.15: 101235334-101235334 on Build GRCh38
rs372021507 Chr.15: 101235044-101235044 on Build GRCh38
rs146047900 Chr.15: 101235409-101235409 on Build GRCh38
rs778262914 Chr.15: 101235340-101235340 on Build GRCh38
rs911919414 Chr.15: 101235091-101235091 on Build GRCh38
rs771613004 Chr.15: 101235346-101235346 on Build GRCh38
rs777322137 Chr.15: 101235426-101235426 on Build GRCh38
rs953788547 Chr.15: 101235037-101235037 on Build GRCh38
rs369982845 Chr.15: 101235488-101235488 on Build GRCh38
rs757906025 Chr.15: 101235213-101235213 on Build GRCh38
rs569937094 Chr.15: 101235360-101235360 on Build GRCh38
rs772881207 Chr.15: 101235244-101235244 on Build GRCh38
rs3743192 Chr.15: 101235364-101235364 on Build GRCh38
rs148344653 Chr.15: 101235322-101235322 on Build GRCh38
rs746006586 Chr.15: 101235374-101235374 on Build GRCh38
rs386352292 Chr.15: 101235167-101235167 on Build GRCh38
rs763160520 Chr.15: 101235279-101235279 on Build GRCh38
rs567957962 Chr.15: 101235253-101235253 on Build GRCh38
rs764225372 Chr.15: 101235283-101235283 on Build GRCh38
rs757857730 Chr.15: 101235424-101235424 on Build GRCh38
rs141538980 Chr.15: 101235324-101235324 on Build GRCh38
rs149315848 Chr.15: 101235109-101235109 on Build GRCh38
rs74545602 Chr.15: 101235232-101235232 on Build GRCh38
rs372381623 Chr.15: 101235105-101235105 on Build GRCh38
rs778341316 Chr.15: 101235068-101235068 on Build GRCh38
rs140045066 Chr.15: 101235421-101235421 on Build GRCh38
rs867562787 Chr.15: 101235120-101235120 on Build GRCh38
rs757665488 Chr.15: 101235212-101235212 on Build GRCh38
rs764708620 Chr.15: 101235232-101235232 on Build GRCh38
rs977820708 Chr.15: 101235029-101235029 on Build GRCh38
rs768279919 Chr.15: 101235118-101235118 on Build GRCh38
rs779593100 Chr.15: 101235222-101235222 on Build GRCh38
rs754390209 Chr.15: 101235413-101235413 on Build GRCh38
rs1020012250 Chr.15: 101235318-101235318 on Build GRCh38
rs770277928 Chr.15: 101235221-101235221 on Build GRCh38
rs115447262 Chr.15: 101235475-101235475 on Build GRCh38
rs201433733 Chr.15: 101235039-101235039 on Build GRCh38
rs745870858 Chr.15: 101235447-101235447 on Build GRCh38
rs756501308 Chr.15: 101235397-101235397 on Build GRCh38
rs147152103 Chr.15: 101235325-101235325 on Build GRCh38
rs776539443 Chr.15: 101235444-101235444 on Build GRCh38
rs758690160 Chr.15: 101235064-101235064 on Build GRCh38
rs780196023 Chr.15: 101235433-101235433 on Build GRCh38
rs769602325 Chr.15: 101235458-101235458 on Build GRCh38
rs753762807 Chr.15: 101235265-101235265 on Build GRCh38
rs773444662 Chr.15: 101235478-101235478 on Build GRCh38
rs146924321 Chr.15: 101235200-101235200 on Build GRCh38
rs369052544 Chr.15: 101235297-101235297 on Build GRCh38
rs149739984 Chr.15: 101235100-101235100 on Build GRCh38
rs751695766 Chr.15: 101235208-101235208 on Build GRCh38
rs546349346 Chr.15: 101235250-101235250 on Build GRCh38
rs762150136 Chr.15: 101235368-101235368 on Build GRCh38
rs1001898226 Chr.15: 101235233-101235233 on Build GRCh38
rs750394065 Chr.15: 101235176-101235176 on Build GRCh38
rs746614193 Chr.15: 101235427-101235427 on Build GRCh38
rs780992038 Chr.15: 101235373-101235373 on Build GRCh38
rs774947799 Chr.15: 101235107-101235107 on Build GRCh38
rs768087787 Chr.15: 101235226-101235226 on Build GRCh38
rs574572282 Chr.15: 101235085-101235085 on Build GRCh38
rs770812991 Chr.15: 101235359-101235359 on Build GRCh38
rs374959506 Chr.15: 101235206-101235206 on Build GRCh38
rs573785211 Chr.15: 101235287-101235287 on Build GRCh38
rs942146414 Chr.15: 101235094-101235094 on Build GRCh38
rs74040399 Chr.15: 101235168-101235168 on Build GRCh38
rs145031427 Chr.15: 101235286-101235286 on Build GRCh38
rs138311444 Chr.15: 101235363-101235363 on Build GRCh38
rs942299713 Chr.15: 101235358-101235358 on Build GRCh38
rs957978709 Chr.15: 101235198-101235198 on Build GRCh38
rs1001866321 Chr.15: 101235439-101235439 on Build GRCh38

View Full Product Details