Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 693806-694296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL16
Entrez Gene ID
146330
Gene Name
F-box and leucine rich repeat protein 16
Gene Aliases
C16orf22, Fbl16, c380A1.1
UniGene
Hs.513244
Gene Chromosome Location
Chr.16: 692500-705825 on Build GRCh38
Transcripts
NM_153350.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2690713 c.1429G>C p.V477L Chr.16: 694286-694286 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs929328656 Chr.16: 694184-694184 on Build GRCh38
rs935309222 Chr.16: 694205-694205 on Build GRCh38
rs993725255 Chr.16: 693884-693884 on Build GRCh38
rs1048029406 Chr.16: 694040-694043 on Build GRCh38
rs954586104 Chr.16: 693958-693958 on Build GRCh38
rs1055221430 Chr.16: 693916-693916 on Build GRCh38
rs879178290 Chr.16: 694037-694038 on Build GRCh38
rs534561522 Chr.16: 693988-693988 on Build GRCh38
rs958457058 Chr.16: 694111-694111 on Build GRCh38
rs1055312130 Chr.16: 694209-694209 on Build GRCh38
rs760469740 Chr.16: 694269-694269 on Build GRCh38
rs3833954 Chr.16: 693982-693983 on Build GRCh38
rs928208441 Chr.16: 694188-694188 on Build GRCh38
rs1003695692 Chr.16: 693911-693911 on Build GRCh38
rs1047936962 Chr.16: 693891-693892 on Build GRCh38
rs964837042 Chr.16: 694118-694118 on Build GRCh38
rs1039539172 Chr.16: 694246-694246 on Build GRCh38
rs1035483518 Chr.16: 693912-693912 on Build GRCh38
rs546775157 Chr.16: 693993-693993 on Build GRCh38
rs188252904 Chr.16: 694069-694069 on Build GRCh38
rs768103665 Chr.16: 694258-694258 on Build GRCh38
rs148739956 Chr.16: 694260-694260 on Build GRCh38
rs552533161 Chr.16: 693917-693917 on Build GRCh38
rs1031531355 Chr.16: 693956-693956 on Build GRCh38
rs181135441 Chr.16: 694136-694136 on Build GRCh38
rs113541943 Chr.16: 693980-693981 on Build GRCh38
rs373803558 Chr.16: 693938-693938 on Build GRCh38
rs548564355 Chr.16: 693842-693842 on Build GRCh38
rs1006998517 Chr.16: 694243-694243 on Build GRCh38
rs1017834035 Chr.16: 694116-694116 on Build GRCh38
rs543438240 Chr.16: 694039-694040 on Build GRCh38
rs987325775 Chr.16: 694114-694114 on Build GRCh38
rs983442978 Chr.16: 694187-694187 on Build GRCh38
rs949647855 Chr.16: 694127-694127 on Build GRCh38
rs894227384 Chr.16: 694065-694066 on Build GRCh38
rs909377082 Chr.16: 693900-693900 on Build GRCh38
rs990918483 Chr.16: 694120-694120 on Build GRCh38
rs942242083 Chr.16: 693913-693913 on Build GRCh38
rs909594931 Chr.16: 694088-694088 on Build GRCh38
rs748918224 Chr.16: 694257-694257 on Build GRCh38
rs976720029 Chr.16: 694060-694060 on Build GRCh38
rs904222004 Chr.16: 694071-694072 on Build GRCh38
rs185623566 Chr.16: 694220-694220 on Build GRCh38
rs906726046 Chr.16: 693910-693910 on Build GRCh38
rs965265888 Chr.16: 694059-694059 on Build GRCh38
rs12934629 Chr.16: 694115-694115 on Build GRCh38
rs929696845 Chr.16: 693846-693846 on Build GRCh38
rs562180431 Chr.16: 694072-694073 on Build GRCh38
rs1053798178 Chr.16: 694207-694207 on Build GRCh38
rs1046803503 Chr.16: 693945-693945 on Build GRCh38
rs968571580 Chr.16: 693840-693840 on Build GRCh38
rs973445163 Chr.16: 694032-694032 on Build GRCh38
rs367614744 Chr.16: 694273-694273 on Build GRCh38
rs899418489 Chr.16: 693864-693865 on Build GRCh38
rs1025719450 Chr.16: 694068-694068 on Build GRCh38
rs563688878 Chr.16: 693845-693845 on Build GRCh38
rs1000003040 Chr.16: 693954-693954 on Build GRCh38
rs1046671595 Chr.16: 694154-694154 on Build GRCh38
rs910647797 Chr.16: 693995-693995 on Build GRCh38
rs771986831 Chr.16: 694264-694264 on Build GRCh38
rs766158199 Chr.16: 694272-694272 on Build GRCh38
rs1038074706 Chr.16: 693853-693853 on Build GRCh38
rs1018135345 Chr.16: 694053-694053 on Build GRCh38
rs568021031 Chr.16: 694057-694057 on Build GRCh38
rs943410895 Chr.16: 694026-694026 on Build GRCh38
rs908676214 Chr.16: 694054-694055 on Build GRCh38
rs998475266 Chr.16: 694072-694072 on Build GRCh38
rs902534320 Chr.16: 693950-693950 on Build GRCh38
rs1013053855 Chr.16: 694065-694065 on Build GRCh38
rs1006697342 Chr.16: 694044-694044 on Build GRCh38
rs559639611 Chr.16: 693840-693840 on Build GRCh38
rs535549618 Chr.16: 694058-694058 on Build GRCh38
rs574919273 Chr.16: 694080-694080 on Build GRCh38
rs916763102 Chr.16: 694122-694122 on Build GRCh38
rs578011648 Chr.16: 693841-693841 on Build GRCh38
rs983916141 Chr.16: 693854-693854 on Build GRCh38
rs954595932 Chr.16: 694103-694103 on Build GRCh38
rs530798048 Chr.16: 693915-693915 on Build GRCh38
rs537335868 Chr.16: 694095-694095 on Build GRCh38
rs965298592 Chr.16: 693829-693829 on Build GRCh38
rs895367037 Chr.16: 693926-693926 on Build GRCh38
rs773077964 Chr.16: 694267-694267 on Build GRCh38
rs893899722 Chr.16: 694004-694004 on Build GRCh38
rs923936785 Chr.16: 694160-694160 on Build GRCh38

View Full Product Details