Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 695628-696137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL16
Entrez Gene ID
146330
Gene Name
F-box and leucine rich repeat protein 16
Gene Aliases
C16orf22, Fbl16, c380A1.1
UniGene
Hs.513244
Gene Chromosome Location
Chr.16: 692500-705825 on Build GRCh38
Transcripts
NM_153350.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6357359 c.851C>T p.A284V Chr.16: 695706-695706 on Build GRCh38
4848489 c.789C>T p.I263I Chr.16: 695768-695768 on Build GRCh38
6240139 c.892A>T p.T298S Chr.16: 695665-695665 on Build GRCh38
6379690 c.844G>T p.D282Y Chr.16: 695713-695713 on Build GRCh38
4912518 c.700G>T p.A234S Chr.16: 695857-695857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980588764 Chr.16: 696110-696110 on Build GRCh38
rs1006006574 Chr.16: 696049-696049 on Build GRCh38
rs777513857 Chr.16: 695966-695966 on Build GRCh38
rs779715251 Chr.16: 695651-695651 on Build GRCh38
rs374285491 Chr.16: 696072-696072 on Build GRCh38
rs866391028 Chr.16: 695746-695746 on Build GRCh38
rs752808901 Chr.16: 695678-695678 on Build GRCh38
rs746826837 Chr.16: 695975-695975 on Build GRCh38
rs1022483575 Chr.16: 695685-695685 on Build GRCh38
rs532836569 Chr.16: 696061-696061 on Build GRCh38
rs1014664601 Chr.16: 696052-696052 on Build GRCh38
rs752061772 Chr.16: 695708-695708 on Build GRCh38
rs762404968 Chr.16: 695935-695935 on Build GRCh38
rs775511332 Chr.16: 695894-695894 on Build GRCh38
rs962752198 Chr.16: 696056-696056 on Build GRCh38
rs773055351 Chr.16: 695849-695849 on Build GRCh38
rs758353826 Chr.16: 695701-695701 on Build GRCh38
rs964265716 Chr.16: 695734-695734 on Build GRCh38
rs892495207 Chr.16: 695958-695958 on Build GRCh38
rs1011348324 Chr.16: 696013-696013 on Build GRCh38
rs763146281 Chr.16: 695899-695899 on Build GRCh38
rs1015568524 Chr.16: 696051-696051 on Build GRCh38
rs776892160 Chr.16: 695657-695657 on Build GRCh38
rs917846451 Chr.16: 696100-696100 on Build GRCh38
rs754535777 Chr.16: 695953-695953 on Build GRCh38
rs962783108 Chr.16: 695940-695940 on Build GRCh38
rs1052795688 Chr.16: 696008-696008 on Build GRCh38
rs779406317 Chr.16: 695860-695860 on Build GRCh38
rs781661544 Chr.16: 695745-695745 on Build GRCh38
rs755335069 Chr.16: 695852-695852 on Build GRCh38
rs757743985 Chr.16: 695744-695744 on Build GRCh38
rs187006034 Chr.16: 696021-696021 on Build GRCh38
rs540143540 Chr.16: 695986-695986 on Build GRCh38
rs113497984 Chr.16: 695932-695932 on Build GRCh38
rs769164060 Chr.16: 695956-695958 on Build GRCh38
rs755773705 Chr.16: 695750-695750 on Build GRCh38
rs1030364303 Chr.16: 696029-696029 on Build GRCh38
rs778846761 Chr.16: 695782-695782 on Build GRCh38
rs539959274 Chr.16: 695944-695944 on Build GRCh38
rs997027865 Chr.16: 696012-696012 on Build GRCh38
rs538668302 Chr.16: 696044-696044 on Build GRCh38
rs748154237 Chr.16: 695961-695961 on Build GRCh38
rs764703969 Chr.16: 695704-695704 on Build GRCh38
rs997098254 Chr.16: 696026-696026 on Build GRCh38
rs370288425 Chr.16: 695943-695943 on Build GRCh38
rs759777166 Chr.16: 695666-695666 on Build GRCh38
rs78996697 Chr.16: 695978-695978 on Build GRCh38
rs970335655 Chr.16: 696078-696078 on Build GRCh38
rs997531244 Chr.16: 695738-695738 on Build GRCh38
rs779510755 Chr.16: 695759-695759 on Build GRCh38
rs373260304 Chr.16: 695861-695861 on Build GRCh38
rs748586575 Chr.16: 695957-695957 on Build GRCh38
rs551483114 Chr.16: 696106-696106 on Build GRCh38
rs768112232 Chr.16: 695774-695774 on Build GRCh38
rs764246057 Chr.16: 695909-695909 on Build GRCh38
rs1004586101 Chr.16: 696046-696046 on Build GRCh38
rs748059924 Chr.16: 695816-695816 on Build GRCh38
rs1043899283 Chr.16: 696017-696017 on Build GRCh38
rs765301732 Chr.16: 695672-695672 on Build GRCh38
rs770924183 Chr.16: 696111-696111 on Build GRCh38
rs140400011 Chr.16: 695765-695765 on Build GRCh38
rs982882726 Chr.16: 695832-695832 on Build GRCh38
rs981301189 Chr.16: 696097-696097 on Build GRCh38
rs112597043 Chr.16: 696011-696011 on Build GRCh38
rs543964492 Chr.16: 696031-696031 on Build GRCh38
rs934281788 Chr.16: 696006-696006 on Build GRCh38
rs768047585 Chr.16: 695942-695942 on Build GRCh38
rs867528378 Chr.16: 695742-695742 on Build GRCh38
rs771898848 Chr.16: 695825-695825 on Build GRCh38
rs557842529 Chr.16: 695952-695952 on Build GRCh38

View Full Product Details