Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22081382-22081877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf52
Entrez Gene ID
730094
Gene Name
chromosome 16 open reading frame 52
Gene Aliases
-
UniGene
Hs.498890
Gene Chromosome Location
Chr.16: 22008135-22084654 on Build GRCh38
Transcripts
NM_001164579.1

SNP Detail

SNP ID Chromosome Location
rs951724523 Chr.16: 22081539-22081539 on Build GRCh38
rs888370240 Chr.16: 22081622-22081622 on Build GRCh38
rs985776173 Chr.16: 22081648-22081648 on Build GRCh38
rs1006670739 Chr.16: 22081623-22081623 on Build GRCh38
rs535042487 Chr.16: 22081620-22081620 on Build GRCh38
rs537306920 Chr.16: 22081720-22081721 on Build GRCh38
rs985134919 Chr.16: 22081432-22081432 on Build GRCh38
rs540145041 Chr.16: 22081621-22081622 on Build GRCh38
rs905981350 Chr.16: 22081642-22081642 on Build GRCh38
rs1012187134 Chr.16: 22081538-22081538 on Build GRCh38
rs191490575 Chr.16: 22081744-22081744 on Build GRCh38
rs372692583 Chr.16: 22081461-22081461 on Build GRCh38
rs1035643787 Chr.16: 22081443-22081443 on Build GRCh38
rs985551535 Chr.16: 22081786-22081786 on Build GRCh38
rs956168406 Chr.16: 22081710-22081710 on Build GRCh38
rs930105223 Chr.16: 22081748-22081748 on Build GRCh38
rs896710365 Chr.16: 22081782-22081783 on Build GRCh38
rs187397998 Chr.16: 22081602-22081602 on Build GRCh38
rs1022952996 Chr.16: 22081638-22081638 on Build GRCh38
rs929883749 Chr.16: 22081612-22081612 on Build GRCh38
rs968100708 Chr.16: 22081567-22081567 on Build GRCh38
rs988992158 Chr.16: 22081741-22081741 on Build GRCh38
rs749542281 Chr.16: 22081799-22081799 on Build GRCh38
rs776787829 Chr.16: 22081405-22081405 on Build GRCh38
rs1023613052 Chr.16: 22081555-22081555 on Build GRCh38
rs550465343 Chr.16: 22081820-22081820 on Build GRCh38
rs911002592 Chr.16: 22081563-22081563 on Build GRCh38
rs994355699 Chr.16: 22081434-22081434 on Build GRCh38
rs184211835 Chr.16: 22081522-22081522 on Build GRCh38
rs193118939 Chr.16: 22081492-22081492 on Build GRCh38
rs972025351 Chr.16: 22081531-22081531 on Build GRCh38
rs72784940 Chr.16: 22081493-22081493 on Build GRCh38
rs879616705 Chr.16: 22081817-22081817 on Build GRCh38
rs756228308 Chr.16: 22081658-22081658 on Build GRCh38
rs866656960 Chr.16: 22081543-22081543 on Build GRCh38
rs931915476 Chr.16: 22081646-22081646 on Build GRCh38
rs943749806 Chr.16: 22081568-22081568 on Build GRCh38
rs1027239790 Chr.16: 22081535-22081535 on Build GRCh38
rs548039786 Chr.16: 22081576-22081576 on Build GRCh38
rs548723772 Chr.16: 22081695-22081695 on Build GRCh38
rs903830252 Chr.16: 22081816-22081816 on Build GRCh38
rs537538239 Chr.16: 22081768-22081768 on Build GRCh38
rs923153539 Chr.16: 22081640-22081640 on Build GRCh38
rs151169343 Chr.16: 22081621-22081626 on Build GRCh38
rs1056661488 Chr.16: 22081756-22081756 on Build GRCh38
rs1031479352 Chr.16: 22081833-22081833 on Build GRCh38
rs975960293 Chr.16: 22081631-22081642 on Build GRCh38
rs968983613 Chr.16: 22081755-22081755 on Build GRCh38
rs527257035 Chr.16: 22081597-22081597 on Build GRCh38
rs910865308 Chr.16: 22081805-22081805 on Build GRCh38

View Full Product Details