Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66911104-66911612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH16
Entrez Gene ID
1014
Gene Name
cadherin 16
Gene Aliases
-
UniGene
Hs.513660
Gene Chromosome Location
Chr.16: 66908122-66918984 on Build GRCh38
Transcripts
NM_001204744.1 NM_001204745.1 NM_001204746.1 NM_004062.3 XM_005255770.2 XM_011522807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2993869 c.1876G>A p.A626T Chr.16: 66911230-66911230 on Build GRCh38
4670824 c.1804A>G p.N602D Chr.16: 66911302-66911302 on Build GRCh38
2993866 c.1924G>A p.D642N Chr.16: 66911182-66911182 on Build GRCh38
7177735 c.1803C>T p.V601V Chr.16: 66911303-66911303 on Build GRCh38
4790851 c.1890C>A p.D630E Chr.16: 66911216-66911216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035883027 Chr.16: 66911409-66911409 on Build GRCh38
rs747238150 Chr.16: 66911156-66911156 on Build GRCh38
rs548891975 Chr.16: 66911475-66911475 on Build GRCh38
rs568654888 Chr.16: 66911482-66911482 on Build GRCh38
rs148168796 Chr.16: 66911243-66911243 on Build GRCh38
rs755461559 Chr.16: 66911149-66911149 on Build GRCh38
rs963772520 Chr.16: 66911431-66911431 on Build GRCh38
rs368319887 Chr.16: 66911180-66911180 on Build GRCh38
rs866334243 Chr.16: 66911264-66911265 on Build GRCh38
rs757524926 Chr.16: 66911230-66911230 on Build GRCh38
rs368971731 Chr.16: 66911390-66911390 on Build GRCh38
rs372239455 Chr.16: 66911347-66911347 on Build GRCh38
rs143791536 Chr.16: 66911236-66911236 on Build GRCh38
rs867172915 Chr.16: 66911559-66911559 on Build GRCh38
rs201375709 Chr.16: 66911135-66911135 on Build GRCh38
rs368749991 Chr.16: 66911320-66911320 on Build GRCh38
rs760433894 Chr.16: 66911187-66911187 on Build GRCh38
rs760169737 Chr.16: 66911305-66911305 on Build GRCh38
rs764959149 Chr.16: 66911533-66911533 on Build GRCh38
rs375254137 Chr.16: 66911318-66911318 on Build GRCh38
rs903442013 Chr.16: 66911337-66911337 on Build GRCh38
rs148829305 Chr.16: 66911207-66911207 on Build GRCh38
rs200026605 Chr.16: 66911252-66911252 on Build GRCh38
rs557338218 Chr.16: 66911585-66911585 on Build GRCh38
rs984133484 Chr.16: 66911538-66911538 on Build GRCh38
rs765794991 Chr.16: 66911265-66911266 on Build GRCh38
rs752806942 Chr.16: 66911219-66911219 on Build GRCh38
rs745557002 Chr.16: 66911207-66911208 on Build GRCh38
rs879195602 Chr.16: 66911483-66911485 on Build GRCh38
rs751678641 Chr.16: 66911287-66911287 on Build GRCh38
rs142029576 Chr.16: 66911249-66911249 on Build GRCh38
rs766799191 Chr.16: 66911195-66911195 on Build GRCh38
rs772331009 Chr.16: 66911151-66911151 on Build GRCh38
rs376907450 Chr.16: 66911574-66911574 on Build GRCh38
rs769541411 Chr.16: 66911157-66911157 on Build GRCh38
rs766165348 Chr.16: 66911260-66911260 on Build GRCh38
rs181688764 Chr.16: 66911319-66911319 on Build GRCh38
rs773908175 Chr.16: 66911172-66911172 on Build GRCh38
rs779868205 Chr.16: 66911231-66911231 on Build GRCh38
rs748123110 Chr.16: 66911248-66911248 on Build GRCh38
rs960181491 Chr.16: 66911415-66911415 on Build GRCh38
rs770603846 Chr.16: 66911359-66911359 on Build GRCh38
rs774101546 Chr.16: 66911246-66911246 on Build GRCh38
rs928759026 Chr.16: 66911127-66911127 on Build GRCh38
rs200214830 Chr.16: 66911277-66911277 on Build GRCh38
rs779352583 Chr.16: 66911208-66911208 on Build GRCh38
rs757471088 Chr.16: 66911302-66911302 on Build GRCh38
rs758412639 Chr.16: 66911221-66911221 on Build GRCh38
rs999487457 Chr.16: 66911407-66911407 on Build GRCh38
rs748213704 Chr.16: 66911150-66911150 on Build GRCh38
rs771744221 Chr.16: 66911237-66911238 on Build GRCh38
rs747186297 Chr.16: 66911229-66911229 on Build GRCh38
rs778798558 Chr.16: 66911321-66911321 on Build GRCh38
rs375285751 Chr.16: 66911158-66911158 on Build GRCh38
rs929257813 Chr.16: 66911556-66911556 on Build GRCh38
rs779219458 Chr.16: 66911139-66911140 on Build GRCh38
rs764137312 Chr.16: 66911281-66911281 on Build GRCh38
rs778025045 Chr.16: 66911152-66911152 on Build GRCh38
rs765646166 Chr.16: 66911266-66911266 on Build GRCh38
rs777049139 Chr.16: 66911263-66911263 on Build GRCh38
rs142629933 Chr.16: 66911496-66911496 on Build GRCh38
rs186023100 Chr.16: 66911547-66911547 on Build GRCh38
rs567500263 Chr.16: 66911546-66911546 on Build GRCh38
rs922237350 Chr.16: 66911163-66911163 on Build GRCh38
rs1046432466 Chr.16: 66911304-66911304 on Build GRCh38
rs943123166 Chr.16: 66911317-66911317 on Build GRCh38
rs564468965 Chr.16: 66911280-66911280 on Build GRCh38
rs772814286 Chr.16: 66911356-66911356 on Build GRCh38
rs868458837 Chr.16: 66911181-66911181 on Build GRCh38
rs746585120 Chr.16: 66911357-66911357 on Build GRCh38
rs767151454 Chr.16: 66911182-66911182 on Build GRCh38
rs370889307 Chr.16: 66911225-66911225 on Build GRCh38
rs991564353 Chr.16: 66911450-66911450 on Build GRCh38
rs762929317 Chr.16: 66911162-66911162 on Build GRCh38
rs760378535 Chr.16: 66911250-66911250 on Build GRCh38
rs768516379 Chr.16: 66911170-66911170 on Build GRCh38
rs765861667 Chr.16: 66911301-66911301 on Build GRCh38
rs566561876 Chr.16: 66911412-66911412 on Build GRCh38
rs200181540 Chr.16: 66911264-66911264 on Build GRCh38
rs1038231916 Chr.16: 66911240-66911240 on Build GRCh38
rs750027499 Chr.16: 66911186-66911186 on Build GRCh38
rs963386920 Chr.16: 66911508-66911508 on Build GRCh38
rs535608343 Chr.16: 66911473-66911473 on Build GRCh38
rs370868898 Chr.16: 66911148-66911148 on Build GRCh38

View Full Product Details