Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66934292-66934783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES2
Entrez Gene ID
8824
Gene Name
carboxylesterase 2
Gene Aliases
CE-2, CES2A1, PCE-2, iCE
UniGene
Hs.282975
Gene Chromosome Location
Chr.16: 66934444-66945096 on Build GRCh38
Transcripts
NM_003869.5 NM_198061.2 NR_036684.1 NM_016062.3 NR_024525.2 NR_046109.1

Gene Symbol
FAM96B
Entrez Gene ID
51647
Gene Name
family with sequence similarity 96 member B
Gene Aliases
CGI-128, MIP18
UniGene
Hs.9825
Gene Chromosome Location
Chr.16: 66932055-66934423 on Build GRCh38
Transcripts
NM_003869.5 NM_198061.2 NR_036684.1 NM_016062.3 NR_024525.2 NR_046109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4683313 c.6A>G p.V2V Chr.16: 66934359-66934359 on Build GRCh38
1609498 c.10G>A p.G4S Chr.16: 66934355-66934355 on Build GRCh38
5660837 c.61C>T p.Q21* Chr.16: 66934304-66934304 on Build GRCh38
7075663 c.39G>C p.E13D Chr.16: 66934326-66934326 on Build GRCh38
4407557 c.20T>G p.V7G Chr.16: 66934345-66934345 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751053998 Chr.16: 66934410-66934410 on Build GRCh38
rs747220172 Chr.16: 66934355-66934355 on Build GRCh38
rs896679076 Chr.16: 66934679-66934679 on Build GRCh38
rs574777374 Chr.16: 66934746-66934746 on Build GRCh38
rs769646965 Chr.16: 66934513-66934513 on Build GRCh38
rs935509127 Chr.16: 66934416-66934416 on Build GRCh38
rs757950623 Chr.16: 66934332-66934332 on Build GRCh38
rs753507526 Chr.16: 66934315-66934315 on Build GRCh38
rs375950439 Chr.16: 66934365-66934365 on Build GRCh38
rs765120377 Chr.16: 66934384-66934384 on Build GRCh38
rs964578971 Chr.16: 66934379-66934379 on Build GRCh38
rs780408191 Chr.16: 66934362-66934362 on Build GRCh38
rs1022565719 Chr.16: 66934445-66934445 on Build GRCh38
rs763568182 Chr.16: 66934336-66934347 on Build GRCh38
rs1053622935 Chr.16: 66934549-66934549 on Build GRCh38
rs1043824230 Chr.16: 66934414-66934414 on Build GRCh38
rs756804609 Chr.16: 66934344-66934344 on Build GRCh38
rs1027410186 Chr.16: 66934476-66934476 on Build GRCh38
rs994962186 Chr.16: 66934474-66934474 on Build GRCh38
rs773505111 Chr.16: 66934382-66934382 on Build GRCh38
rs775565018 Chr.16: 66934395-66934395 on Build GRCh38
rs201627927 Chr.16: 66934380-66934380 on Build GRCh38
rs905551368 Chr.16: 66934463-66934463 on Build GRCh38
rs1030113023 Chr.16: 66934413-66934413 on Build GRCh38
rs920205380 Chr.16: 66934485-66934485 on Build GRCh38
rs979879699 Chr.16: 66934655-66934655 on Build GRCh38
rs558683856 Chr.16: 66934447-66934447 on Build GRCh38
rs983608289 Chr.16: 66934442-66934442 on Build GRCh38
rs376528249 Chr.16: 66934670-66934671 on Build GRCh38
rs891853443 Chr.16: 66934582-66934582 on Build GRCh38
rs948281572 Chr.16: 66934688-66934688 on Build GRCh38
rs935441087 Chr.16: 66934657-66934657 on Build GRCh38
rs746181958 Chr.16: 66934352-66934352 on Build GRCh38
rs749856523 Chr.16: 66934364-66934364 on Build GRCh38
rs759228631 Chr.16: 66934383-66934383 on Build GRCh38
rs777492408 Chr.16: 66934334-66934334 on Build GRCh38
rs763043917 Chr.16: 66934399-66934399 on Build GRCh38
rs924039594 Chr.16: 66934656-66934656 on Build GRCh38
rs766789584 Chr.16: 66934345-66934346 on Build GRCh38
rs374829700 Chr.16: 66934368-66934368 on Build GRCh38
rs965928670 Chr.16: 66934545-66934545 on Build GRCh38
rs902640181 Chr.16: 66934415-66934415 on Build GRCh38
rs866384549 Chr.16: 66934321-66934321 on Build GRCh38
rs773140266 Chr.16: 66934335-66934336 on Build GRCh38
rs1056537293 Chr.16: 66934672-66934672 on Build GRCh38
rs752730258 Chr.16: 66934329-66934329 on Build GRCh38
rs950447927 Chr.16: 66934607-66934607 on Build GRCh38
rs866432498 Chr.16: 66934523-66934523 on Build GRCh38
rs11556909 Chr.16: 66934314-66934314 on Build GRCh38
rs1030409924 Chr.16: 66934598-66934598 on Build GRCh38
rs543831316 Chr.16: 66934762-66934762 on Build GRCh38
rs781408556 Chr.16: 66934436-66934436 on Build GRCh38
rs751244375 Chr.16: 66934337-66934337 on Build GRCh38
rs921894443 Chr.16: 66934642-66934642 on Build GRCh38
rs745599731 Chr.16: 66934441-66934441 on Build GRCh38
rs572516144 Chr.16: 66934487-66934487 on Build GRCh38
rs774353687 Chr.16: 66934366-66934366 on Build GRCh38
rs1013865591 Chr.16: 66934331-66934331 on Build GRCh38
rs780903835 Chr.16: 66934346-66934346 on Build GRCh38
rs1043047451 Chr.16: 66934603-66934603 on Build GRCh38
rs1057066721 Chr.16: 66934418-66934418 on Build GRCh38
rs921946838 Chr.16: 66934398-66934398 on Build GRCh38
rs866700118 Chr.16: 66934349-66934349 on Build GRCh38
rs946739491 Chr.16: 66934644-66934644 on Build GRCh38
rs996046182 Chr.16: 66934666-66934666 on Build GRCh38
rs944640076 Chr.16: 66934390-66934390 on Build GRCh38
rs950590013 Chr.16: 66934350-66934350 on Build GRCh38
rs974670094 Chr.16: 66934596-66934596 on Build GRCh38
rs1018819282 Chr.16: 66934532-66934532 on Build GRCh38
rs948943550 Chr.16: 66934501-66934501 on Build GRCh38
rs541140256 Chr.16: 66934508-66934508 on Build GRCh38
rs868711998 Chr.16: 66934717-66934717 on Build GRCh38
rs756526985 Chr.16: 66934360-66934360 on Build GRCh38
rs1044625071 Chr.16: 66934521-66934521 on Build GRCh38
rs562779679 Chr.16: 66934530-66934530 on Build GRCh38
rs759405706 Chr.16: 66934317-66934317 on Build GRCh38
rs764141881 Chr.16: 66934404-66934404 on Build GRCh38
rs67575372 Chr.16: 66934724-66934724 on Build GRCh38

View Full Product Details