Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75238002-75238493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAR1
Entrez Gene ID
9564
Gene Name
BCAR1, Cas family scaffolding protein
Gene Aliases
CAS, CAS1, CASS1, CRKAS, P130Cas
UniGene
Hs.479747
Gene Chromosome Location
Chr.16: 75229030-75268053 on Build GRCh38
Transcripts
NM_001170714.1 NM_001170715.1 NM_001170716.1 NM_001170717.1 NM_001170718.1 NM_001170719.1 NM_001170720.1 NM_001170721.1 NM_014567.3 XM_005256258.4 XM_005256259.4 XM_005256260.4 XM_011523475.2 XM_017023897.1 XM_017023898.1 XM_017023899.1

SNP Detail

SNP ID Chromosome Location
rs892083293 Chr.16: 75238210-75238210 on Build GRCh38
rs550842390 Chr.16: 75238067-75238067 on Build GRCh38
rs997900465 Chr.16: 75238438-75238438 on Build GRCh38
rs905000802 Chr.16: 75238263-75238263 on Build GRCh38
rs931359121 Chr.16: 75238172-75238172 on Build GRCh38
rs368808041 Chr.16: 75238121-75238121 on Build GRCh38
rs757341699 Chr.16: 75238130-75238130 on Build GRCh38
rs866577950 Chr.16: 75238074-75238074 on Build GRCh38
rs988918913 Chr.16: 75238036-75238036 on Build GRCh38
rs141799641 Chr.16: 75238199-75238199 on Build GRCh38
rs370435788 Chr.16: 75238430-75238430 on Build GRCh38
rs115501114 Chr.16: 75238132-75238132 on Build GRCh38
rs955720444 Chr.16: 75238032-75238032 on Build GRCh38
rs953639166 Chr.16: 75238308-75238308 on Build GRCh38
rs976250051 Chr.16: 75238110-75238110 on Build GRCh38
rs992744031 Chr.16: 75238129-75238129 on Build GRCh38
rs13337017 Chr.16: 75238469-75238469 on Build GRCh38
rs925563762 Chr.16: 75238343-75238343 on Build GRCh38
rs373550447 Chr.16: 75238386-75238386 on Build GRCh38
rs556821060 Chr.16: 75238204-75238204 on Build GRCh38
rs559165199 Chr.16: 75238190-75238190 on Build GRCh38
rs891976579 Chr.16: 75238414-75238414 on Build GRCh38
rs774441449 Chr.16: 75238303-75238303 on Build GRCh38
rs750347646 Chr.16: 75238043-75238043 on Build GRCh38
rs968373072 Chr.16: 75238243-75238243 on Build GRCh38
rs542714714 Chr.16: 75238297-75238297 on Build GRCh38
rs766983175 Chr.16: 75238053-75238053 on Build GRCh38
rs575755772 Chr.16: 75238198-75238198 on Build GRCh38
rs892575231 Chr.16: 75238208-75238208 on Build GRCh38
rs556264582 Chr.16: 75238025-75238025 on Build GRCh38
rs1046275782 Chr.16: 75238235-75238235 on Build GRCh38
rs944218944 Chr.16: 75238401-75238401 on Build GRCh38
rs1049859848 Chr.16: 75238187-75238187 on Build GRCh38
rs1034057498 Chr.16: 75238453-75238453 on Build GRCh38
rs567410954 Chr.16: 75238080-75238080 on Build GRCh38
rs949434853 Chr.16: 75238211-75238211 on Build GRCh38
rs538979757 Chr.16: 75238155-75238155 on Build GRCh38
rs753625070 Chr.16: 75238082-75238082 on Build GRCh38
rs1021215851 Chr.16: 75238236-75238236 on Build GRCh38
rs925316481 Chr.16: 75238147-75238147 on Build GRCh38
rs182567000 Chr.16: 75238307-75238307 on Build GRCh38
rs926484085 Chr.16: 75238391-75238391 on Build GRCh38
rs1041570690 Chr.16: 75238409-75238409 on Build GRCh38
rs962237901 Chr.16: 75238291-75238291 on Build GRCh38
rs1001188218 Chr.16: 75238451-75238451 on Build GRCh38
rs34699861 Chr.16: 75238230-75238231 on Build GRCh38
rs372076058 Chr.16: 75238087-75238087 on Build GRCh38
rs369646650 Chr.16: 75238322-75238322 on Build GRCh38
rs1034088651 Chr.16: 75238270-75238270 on Build GRCh38
rs1010880676 Chr.16: 75238425-75238425 on Build GRCh38
rs1010495148 Chr.16: 75238221-75238221 on Build GRCh38
rs919934019 Chr.16: 75238154-75238154 on Build GRCh38
rs1001220978 Chr.16: 75238269-75238269 on Build GRCh38
rs1036993346 Chr.16: 75238140-75238140 on Build GRCh38
rs931328169 Chr.16: 75238363-75238363 on Build GRCh38
rs1049827566 Chr.16: 75238370-75238370 on Build GRCh38
rs1036677128 Chr.16: 75238415-75238415 on Build GRCh38
rs1043312861 Chr.16: 75238431-75238431 on Build GRCh38
rs976449634 Chr.16: 75238288-75238289 on Build GRCh38
rs1027351950 Chr.16: 75238279-75238279 on Build GRCh38
rs1048208768 Chr.16: 75238408-75238408 on Build GRCh38
rs913874958 Chr.16: 75238369-75238369 on Build GRCh38
rs1002128292 Chr.16: 75238272-75238272 on Build GRCh38
rs1003858564 Chr.16: 75238284-75238284 on Build GRCh38
rs907702158 Chr.16: 75238290-75238290 on Build GRCh38
rs1021328476 Chr.16: 75238037-75238037 on Build GRCh38
rs749512091 Chr.16: 75238205-75238205 on Build GRCh38
rs904184346 Chr.16: 75238450-75238450 on Build GRCh38
rs3826109 Chr.16: 75238109-75238109 on Build GRCh38
rs760759011 Chr.16: 75238071-75238071 on Build GRCh38
rs730367 Chr.16: 75238131-75238131 on Build GRCh38
rs190077582 Chr.16: 75238283-75238283 on Build GRCh38
rs571360306 Chr.16: 75238024-75238024 on Build GRCh38
rs958295180 Chr.16: 75238356-75238356 on Build GRCh38
rs377607853 Chr.16: 75238288-75238288 on Build GRCh38

View Full Product Details