Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81351383-81351884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAN
Entrez Gene ID
8139
Gene Name
gigaxonin
Gene Aliases
GAN1, KLHL16
UniGene
Hs.112569
Gene Chromosome Location
Chr.16: 81314941-81380198 on Build GRCh38
Transcripts
NM_022041.3 XM_017023734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
74728 c.218T>C p.I73T Chr.16: 81351633-81351633 on Build GRCh38
6669208 c.183T>C p.Y61Y Chr.16: 81351598-81351598 on Build GRCh38
3421189 c.225T>C p.L75L Chr.16: 81351640-81351640 on Build GRCh38
974087 c.236C>T p.S79L Chr.16: 81351651-81351651 on Build GRCh38
4686377 c.244G>T p.V82F Chr.16: 81351659-81351659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780435880 Chr.16: 81351631-81351631 on Build GRCh38
rs865797654 Chr.16: 81351602-81351602 on Build GRCh38
rs770094216 Chr.16: 81351711-81351712 on Build GRCh38
rs374039254 Chr.16: 81351559-81351559 on Build GRCh38
rs531603690 Chr.16: 81351424-81351424 on Build GRCh38
rs755471804 Chr.16: 81351725-81351725 on Build GRCh38
rs754994754 Chr.16: 81351564-81351565 on Build GRCh38
rs776602704 Chr.16: 81351542-81351542 on Build GRCh38
rs778262818 Chr.16: 81351452-81351452 on Build GRCh38
rs538444025 Chr.16: 81351468-81351468 on Build GRCh38
rs911350150 Chr.16: 81351670-81351670 on Build GRCh38
rs763764629 Chr.16: 81351619-81351619 on Build GRCh38
rs1012533348 Chr.16: 81351846-81351846 on Build GRCh38
rs1051664642 Chr.16: 81351482-81351482 on Build GRCh38
rs113602404 Chr.16: 81351412-81351412 on Build GRCh38
rs149912994 Chr.16: 81351406-81351406 on Build GRCh38
rs371054532 Chr.16: 81351659-81351659 on Build GRCh38
rs1008242723 Chr.16: 81351780-81351780 on Build GRCh38
rs16955109 Chr.16: 81351436-81351436 on Build GRCh38
rs1047056995 Chr.16: 81351767-81351767 on Build GRCh38
rs891147536 Chr.16: 81351756-81351756 on Build GRCh38
rs527429003 Chr.16: 81351407-81351407 on Build GRCh38
rs746067847 Chr.16: 81351587-81351587 on Build GRCh38
rs759464937 Chr.16: 81351679-81351679 on Build GRCh38
rs746821241 Chr.16: 81351538-81351538 on Build GRCh38
rs960747636 Chr.16: 81351527-81351527 on Build GRCh38
rs192761504 Chr.16: 81351415-81351415 on Build GRCh38
rs905008890 Chr.16: 81351807-81351807 on Build GRCh38
rs1049701549 Chr.16: 81351750-81351750 on Build GRCh38
rs770847887 Chr.16: 81351662-81351662 on Build GRCh38
rs146629249 Chr.16: 81351653-81351653 on Build GRCh38
rs757976343 Chr.16: 81351605-81351605 on Build GRCh38
rs766328006 Chr.16: 81351541-81351541 on Build GRCh38
rs545269994 Chr.16: 81351796-81351796 on Build GRCh38
rs748736329 Chr.16: 81351733-81351733 on Build GRCh38
rs9928022 Chr.16: 81351783-81351783 on Build GRCh38
rs748650343 Chr.16: 81351708-81351711 on Build GRCh38
rs897287371 Chr.16: 81351785-81351785 on Build GRCh38
rs759461679 Chr.16: 81351546-81351546 on Build GRCh38
rs35479015 Chr.16: 81351758-81351759 on Build GRCh38
rs184871836 Chr.16: 81351454-81351454 on Build GRCh38
rs748951541 Chr.16: 81351747-81351753 on Build GRCh38
rs778175591 Chr.16: 81351712-81351715 on Build GRCh38
rs9935105 Chr.16: 81351517-81351517 on Build GRCh38
rs1023866952 Chr.16: 81351532-81351532 on Build GRCh38
rs766751614 Chr.16: 81351717-81351717 on Build GRCh38
rs770645867 Chr.16: 81351543-81351543 on Build GRCh38
rs751643674 Chr.16: 81351565-81351565 on Build GRCh38
rs553977605 Chr.16: 81351486-81351486 on Build GRCh38
rs555781582 Chr.16: 81351531-81351531 on Build GRCh38
rs1013423636 Chr.16: 81351530-81351530 on Build GRCh38
rs747278062 Chr.16: 81351661-81351661 on Build GRCh38
rs1038384791 Chr.16: 81351504-81351504 on Build GRCh38
rs891200179 Chr.16: 81351467-81351467 on Build GRCh38
rs749797346 Chr.16: 81351632-81351632 on Build GRCh38
rs1015696356 Chr.16: 81351484-81351484 on Build GRCh38
rs750735081 Chr.16: 81351628-81351628 on Build GRCh38
rs555307037 Chr.16: 81351519-81351519 on Build GRCh38
rs890044739 Chr.16: 81351483-81351483 on Build GRCh38
rs188440858 Chr.16: 81351501-81351501 on Build GRCh38
rs370616133 Chr.16: 81351551-81351551 on Build GRCh38
rs1054133028 Chr.16: 81351434-81351434 on Build GRCh38
rs956586165 Chr.16: 81351855-81351855 on Build GRCh38
rs996080984 Chr.16: 81351788-81351788 on Build GRCh38
rs929968884 Chr.16: 81351762-81351762 on Build GRCh38
rs923029952 Chr.16: 81351612-81351612 on Build GRCh38
rs979139290 Chr.16: 81351446-81351446 on Build GRCh38
rs578225838 Chr.16: 81351791-81351791 on Build GRCh38
rs571071677 Chr.16: 81351641-81351641 on Build GRCh38
rs776351193 Chr.16: 81351676-81351676 on Build GRCh38
rs758519281 Chr.16: 81351739-81351739 on Build GRCh38
rs563977770 Chr.16: 81351700-81351702 on Build GRCh38
rs1024628242 Chr.16: 81351847-81351847 on Build GRCh38
rs376061524 Chr.16: 81351727-81351727 on Build GRCh38
rs754166690 Chr.16: 81351719-81351719 on Build GRCh38
rs377294873 Chr.16: 81351633-81351633 on Build GRCh38
rs778984857 Chr.16: 81351649-81351649 on Build GRCh38
rs924913235 Chr.16: 81351453-81351453 on Build GRCh38
rs16955113 Chr.16: 81351824-81351824 on Build GRCh38
rs1052118995 Chr.16: 81351463-81351463 on Build GRCh38
rs765086970 Chr.16: 81351688-81351688 on Build GRCh38
rs1039007762 Chr.16: 81351697-81351697 on Build GRCh38
rs751181075 Chr.16: 81351704-81351704 on Build GRCh38
rs762469365 Chr.16: 81351696-81351696 on Build GRCh38
rs77078389 Chr.16: 81351535-81351535 on Build GRCh38
rs756446204 Chr.16: 81351630-81351630 on Build GRCh38
rs144007114 Chr.16: 81351652-81351652 on Build GRCh38
rs753427752 Chr.16: 81351416-81351416 on Build GRCh38
rs113906423 Chr.16: 81351520-81351520 on Build GRCh38
rs763640172 Chr.16: 81351701-81351701 on Build GRCh38
rs180907193 Chr.16: 81351822-81351822 on Build GRCh38
rs756969010 Chr.16: 81351713-81351713 on Build GRCh38
rs747217156 Chr.16: 81351742-81351742 on Build GRCh38
rs778063383 Chr.16: 81351740-81351740 on Build GRCh38
rs775308001 Chr.16: 81351423-81351423 on Build GRCh38
rs760570816 Chr.16: 81351537-81351537 on Build GRCh38
rs775021927 Chr.16: 81351694-81351694 on Build GRCh38
rs373346295 Chr.16: 81351851-81351851 on Build GRCh38

View Full Product Details