Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84566495-84566988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COTL1
Entrez Gene ID
23406
Gene Name
coactosin like F-actin binding protein 1
Gene Aliases
CLP
UniGene
Hs.289092
Gene Chromosome Location
Chr.16: 84565596-84618096 on Build GRCh38
Transcripts
NM_021149.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5584894 c.424G>A p.E142K Chr.16: 84566850-84566850 on Build GRCh38
7271813 c.412G>A p.D138N Chr.16: 84566862-84566862 on Build GRCh38
328958 c.359T>A p.L120Q Chr.16: 84566915-84566915 on Build GRCh38
5851638 c.422C>A p.T141K Chr.16: 84566852-84566852 on Build GRCh38
3513011 c.363G>A p.E121E Chr.16: 84566911-84566911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911911944 Chr.16: 84566705-84566705 on Build GRCh38
rs1054017374 Chr.16: 84566934-84566934 on Build GRCh38
rs773484425 Chr.16: 84566904-84566904 on Build GRCh38
rs1014954300 Chr.16: 84566609-84566609 on Build GRCh38
rs961343399 Chr.16: 84566817-84566817 on Build GRCh38
rs752759974 Chr.16: 84566962-84566962 on Build GRCh38
rs750891702 Chr.16: 84566825-84566825 on Build GRCh38
rs940701969 Chr.16: 84566695-84566695 on Build GRCh38
rs372969859 Chr.16: 84566834-84566834 on Build GRCh38
rs899303717 Chr.16: 84566546-84566546 on Build GRCh38
rs570693248 Chr.16: 84566781-84566781 on Build GRCh38
rs762307122 Chr.16: 84566846-84566846 on Build GRCh38
rs247860 Chr.16: 84566741-84566741 on Build GRCh38
rs746629605 Chr.16: 84566747-84566747 on Build GRCh38
rs113170129 Chr.16: 84566686-84566686 on Build GRCh38
rs757430839 Chr.16: 84566633-84566633 on Build GRCh38
rs769290223 Chr.16: 84566879-84566879 on Build GRCh38
rs566860078 Chr.16: 84566826-84566826 on Build GRCh38
rs1046421698 Chr.16: 84566731-84566731 on Build GRCh38
rs780683584 Chr.16: 84566843-84566843 on Build GRCh38
rs764413495 Chr.16: 84566547-84566547 on Build GRCh38
rs199775217 Chr.16: 84566831-84566831 on Build GRCh38
rs568397127 Chr.16: 84566840-84566840 on Build GRCh38
rs757216407 Chr.16: 84566877-84566877 on Build GRCh38
rs796129149 Chr.16: 84566664-84566665 on Build GRCh38
rs771618110 Chr.16: 84566917-84566917 on Build GRCh38
rs970765128 Chr.16: 84566603-84566603 on Build GRCh38
rs745342465 Chr.16: 84566844-84566844 on Build GRCh38
rs548867252 Chr.16: 84566762-84566762 on Build GRCh38
rs921132727 Chr.16: 84566667-84566667 on Build GRCh38
rs998590353 Chr.16: 84566642-84566642 on Build GRCh38
rs769457516 Chr.16: 84566845-84566845 on Build GRCh38
rs534924393 Chr.16: 84566657-84566657 on Build GRCh38
rs374406128 Chr.16: 84566852-84566852 on Build GRCh38
rs1001493291 Chr.16: 84566784-84566784 on Build GRCh38
rs1036468318 Chr.16: 84566520-84566520 on Build GRCh38
rs577103781 Chr.16: 84566950-84566950 on Build GRCh38
rs573143169 Chr.16: 84566553-84566553 on Build GRCh38
rs963230686 Chr.16: 84566645-84566645 on Build GRCh38
rs374951200 Chr.16: 84566863-84566863 on Build GRCh38
rs546735123 Chr.16: 84566795-84566795 on Build GRCh38
rs373624980 Chr.16: 84566717-84566717 on Build GRCh38
rs776309452 Chr.16: 84566845-84566855 on Build GRCh38
rs201571022 Chr.16: 84566929-84566929 on Build GRCh38
rs200867929 Chr.16: 84566932-84566932 on Build GRCh38
rs371729388 Chr.16: 84566833-84566834 on Build GRCh38
rs930754142 Chr.16: 84566712-84566712 on Build GRCh38
rs987760534 Chr.16: 84566687-84566687 on Build GRCh38
rs758268110 Chr.16: 84566868-84566868 on Build GRCh38
rs1046623 Chr.16: 84566665-84566665 on Build GRCh38
rs530508127 Chr.16: 84566544-84566544 on Build GRCh38
rs777439901 Chr.16: 84566871-84566871 on Build GRCh38
rs755286238 Chr.16: 84566833-84566833 on Build GRCh38
rs563456495 Chr.16: 84566715-84566715 on Build GRCh38
rs765593536 Chr.16: 84566862-84566862 on Build GRCh38
rs1024643293 Chr.16: 84566568-84566568 on Build GRCh38
rs746941098 Chr.16: 84566839-84566840 on Build GRCh38
rs141928182 Chr.16: 84566860-84566860 on Build GRCh38
rs779740777 Chr.16: 84566893-84566893 on Build GRCh38
rs545577865 Chr.16: 84566719-84566719 on Build GRCh38
rs955404738 Chr.16: 84566787-84566787 on Build GRCh38
rs1032660813 Chr.16: 84566756-84566756 on Build GRCh38
rs751300532 Chr.16: 84566873-84566873 on Build GRCh38
rs1001851423 Chr.16: 84566767-84566767 on Build GRCh38
rs1034540671 Chr.16: 84566558-84566558 on Build GRCh38
rs962744542 Chr.16: 84566799-84566799 on Build GRCh38
rs766580363 Chr.16: 84566827-84566827 on Build GRCh38
rs41291692 Chr.16: 84566708-84566708 on Build GRCh38
rs996529938 Chr.16: 84566951-84566951 on Build GRCh38
rs71386852 Chr.16: 84566779-84566779 on Build GRCh38
rs891918313 Chr.16: 84566740-84566740 on Build GRCh38
rs761056676 Chr.16: 84566913-84566913 on Build GRCh38
rs753995579 Chr.16: 84566859-84566859 on Build GRCh38
rs768111166 Chr.16: 84566821-84566821 on Build GRCh38
rs936205202 Chr.16: 84566727-84566727 on Build GRCh38
rs1009683711 Chr.16: 84566555-84566555 on Build GRCh38
rs772312264 Chr.16: 84566849-84566849 on Build GRCh38
rs904605440 Chr.16: 84566721-84566721 on Build GRCh38
rs758094068 Chr.16: 84566836-84566836 on Build GRCh38
rs774727198 Chr.16: 84566801-84566801 on Build GRCh38
rs751647301 Chr.16: 84566590-84566590 on Build GRCh38
rs147677885 Chr.16: 84566781-84566782 on Build GRCh38
rs562218563 Chr.16: 84566771-84566771 on Build GRCh38
rs911875608 Chr.16: 84566691-84566691 on Build GRCh38
rs759753858 Chr.16: 84566949-84566949 on Build GRCh38
rs559341417 Chr.16: 84566722-84566722 on Build GRCh38
rs376446340 Chr.16: 84566794-84566794 on Build GRCh38
rs762034210 Chr.16: 84566803-84566803 on Build GRCh38
rs1053285604 Chr.16: 84566733-84566733 on Build GRCh38
rs780718634 Chr.16: 84566565-84566566 on Build GRCh38
rs768575155 Chr.16: 84566840-84566841 on Build GRCh38
rs773568755 Chr.16: 84566851-84566851 on Build GRCh38
rs1024948065 Chr.16: 84566746-84566746 on Build GRCh38
rs554863276 Chr.16: 84566839-84566839 on Build GRCh38
rs557238908 Chr.16: 84566878-84566878 on Build GRCh38
rs777830918 Chr.16: 84566838-84566838 on Build GRCh38
rs369399633 Chr.16: 84566894-84566894 on Build GRCh38
rs1033405267 Chr.16: 84566638-84566638 on Build GRCh38
rs531027081 Chr.16: 84566778-84566778 on Build GRCh38
rs377452050 Chr.16: 84566901-84566901 on Build GRCh38

View Full Product Details