Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89645360-89645871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP1A
Entrez Gene ID
5119
Gene Name
charged multivesicular body protein 1A
Gene Aliases
CHMP1, PCH8, PCOLN3, PRSM1, VPS46-1, VPS46A
UniGene
Hs.589427
Gene Chromosome Location
Chr.16: 89644431-89657785 on Build GRCh38
Transcripts
NM_001083314.3 NM_002768.4 NR_046418.2

SNP Detail

SNP ID Chromosome Location
rs574043057 Chr.16: 89645434-89645434 on Build GRCh38
rs530401153 Chr.16: 89645596-89645596 on Build GRCh38
rs868553025 Chr.16: 89645421-89645421 on Build GRCh38
rs968945437 Chr.16: 89645784-89645784 on Build GRCh38
rs914041763 Chr.16: 89645439-89645439 on Build GRCh38
rs927007555 Chr.16: 89645638-89645638 on Build GRCh38
rs138025964 Chr.16: 89645602-89645602 on Build GRCh38
rs761831775 Chr.16: 89645573-89645573 on Build GRCh38
rs920514645 Chr.16: 89645488-89645488 on Build GRCh38
rs769577731 Chr.16: 89645593-89645593 on Build GRCh38
rs1001316421 Chr.16: 89645474-89645474 on Build GRCh38
rs186070297 Chr.16: 89645570-89645570 on Build GRCh38
rs1055004529 Chr.16: 89645484-89645484 on Build GRCh38
rs900207495 Chr.16: 89645566-89645566 on Build GRCh38
rs1018926660 Chr.16: 89645701-89645701 on Build GRCh38
rs888587640 Chr.16: 89645688-89645688 on Build GRCh38
rs775268822 Chr.16: 89645511-89645511 on Build GRCh38
rs1056848919 Chr.16: 89645651-89645651 on Build GRCh38
rs3751692 Chr.16: 89645730-89645730 on Build GRCh38
rs933125016 Chr.16: 89645392-89645392 on Build GRCh38
rs913094875 Chr.16: 89645780-89645780 on Build GRCh38
rs771900367 Chr.16: 89645472-89645472 on Build GRCh38
rs947260018 Chr.16: 89645437-89645437 on Build GRCh38
rs564231445 Chr.16: 89645430-89645430 on Build GRCh38
rs922670999 Chr.16: 89645588-89645588 on Build GRCh38
rs990846399 Chr.16: 89645814-89645814 on Build GRCh38
rs181181083 Chr.16: 89645396-89645396 on Build GRCh38
rs73265376 Chr.16: 89645674-89645674 on Build GRCh38
rs768215417 Chr.16: 89645561-89645561 on Build GRCh38
rs74679687 Chr.16: 89645571-89645571 on Build GRCh38
rs998599124 Chr.16: 89645736-89645736 on Build GRCh38
rs561154837 Chr.16: 89645605-89645605 on Build GRCh38
rs112967134 Chr.16: 89645661-89645661 on Build GRCh38
rs1021155727 Chr.16: 89645592-89645592 on Build GRCh38
rs938018127 Chr.16: 89645477-89645477 on Build GRCh38
rs944589844 Chr.16: 89645846-89645846 on Build GRCh38
rs533431580 Chr.16: 89645702-89645702 on Build GRCh38
rs988202473 Chr.16: 89645779-89645779 on Build GRCh38
rs115450693 Chr.16: 89645512-89645512 on Build GRCh38
rs927983268 Chr.16: 89645448-89645456 on Build GRCh38
rs115335995 Chr.16: 89645768-89645768 on Build GRCh38
rs892786551 Chr.16: 89645705-89645705 on Build GRCh38
rs913042694 Chr.16: 89645603-89645603 on Build GRCh38
rs1005823784 Chr.16: 89645699-89645699 on Build GRCh38
rs1042745054 Chr.16: 89645471-89645471 on Build GRCh38
rs1029638157 Chr.16: 89645758-89645758 on Build GRCh38
rs149328027 Chr.16: 89645403-89645403 on Build GRCh38
rs994292888 Chr.16: 89645537-89645537 on Build GRCh38
rs930585978 Chr.16: 89645607-89645607 on Build GRCh38
rs376598185 Chr.16: 89645698-89645698 on Build GRCh38
rs145010051 Chr.16: 89645534-89645536 on Build GRCh38
rs79171694 Chr.16: 89645572-89645572 on Build GRCh38
rs886256184 Chr.16: 89645540-89645540 on Build GRCh38
rs577721067 Chr.16: 89645821-89645821 on Build GRCh38
rs558102637 Chr.16: 89645391-89645391 on Build GRCh38
rs936995719 Chr.16: 89645645-89645645 on Build GRCh38
rs541217495 Chr.16: 89645453-89645453 on Build GRCh38
rs982198144 Chr.16: 89645446-89645446 on Build GRCh38
rs895571489 Chr.16: 89645660-89645660 on Build GRCh38
rs1003717411 Chr.16: 89645773-89645773 on Build GRCh38
rs766642356 Chr.16: 89645608-89645608 on Build GRCh38
rs976888499 Chr.16: 89645574-89645574 on Build GRCh38
rs959467311 Chr.16: 89645790-89645790 on Build GRCh38
rs952971443 Chr.16: 89645563-89645563 on Build GRCh38
rs866619128 Chr.16: 89645686-89645686 on Build GRCh38
rs935365607 Chr.16: 89645601-89645601 on Build GRCh38
rs927200309 Chr.16: 89645822-89645822 on Build GRCh38
rs937234561 Chr.16: 89645835-89645835 on Build GRCh38
rs537486249 Chr.16: 89645400-89645400 on Build GRCh38
rs983373737 Chr.16: 89645826-89645827 on Build GRCh38
rs977217172 Chr.16: 89645386-89645386 on Build GRCh38
rs1027641681 Chr.16: 89645818-89645818 on Build GRCh38
rs1035834776 Chr.16: 89645498-89645498 on Build GRCh38
rs866061288 Chr.16: 89645433-89645433 on Build GRCh38
rs1035564038 Chr.16: 89645785-89645785 on Build GRCh38
rs967213101 Chr.16: 89645444-89645444 on Build GRCh38
rs78208987 Chr.16: 89645630-89645630 on Build GRCh38

View Full Product Details