Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4149500-4150003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5D2
Entrez Gene ID
124936
Gene Name
cytochrome b5 domain containing 2
Gene Aliases
-
UniGene
Hs.513871
Gene Chromosome Location
Chr.17: 4143117-4157701 on Build GRCh38
Transcripts
NM_001254755.1 NM_001254756.1 NM_144611.3 XM_006721447.2 XM_011523649.2 XM_017024180.1 XM_017024181.1 XM_017024182.1 XM_017024183.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5926954 c.310G>A p.D104N Chr.17: 4149950-4149950 on Build GRCh38
5470679 c.313G>A p.V105I Chr.17: 4149953-4149953 on Build GRCh38
979482 c.336G>T p.E112D Chr.17: 4149976-4149976 on Build GRCh38
979481 c.259G>A p.A87T Chr.17: 4149899-4149899 on Build GRCh38
1202772 c.341T>C p.L114P Chr.17: 4149981-4149981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767101191 Chr.17: 4149692-4149692 on Build GRCh38
rs530190421 Chr.17: 4149660-4149660 on Build GRCh38
rs376171076 Chr.17: 4149919-4149919 on Build GRCh38
rs759501201 Chr.17: 4149877-4149877 on Build GRCh38
rs558507719 Chr.17: 4149685-4149685 on Build GRCh38
rs201923584 Chr.17: 4149956-4149956 on Build GRCh38
rs750718885 Chr.17: 4149970-4149970 on Build GRCh38
rs1026656665 Chr.17: 4149966-4149966 on Build GRCh38
rs148723615 Chr.17: 4149814-4149814 on Build GRCh38
rs573823542 Chr.17: 4149788-4149788 on Build GRCh38
rs905570761 Chr.17: 4149604-4149604 on Build GRCh38
rs947209401 Chr.17: 4149761-4149761 on Build GRCh38
rs1048016496 Chr.17: 4149840-4149840 on Build GRCh38
rs138353200 Chr.17: 4149798-4149799 on Build GRCh38
rs1035353144 Chr.17: 4149651-4149651 on Build GRCh38
rs952205534 Chr.17: 4149672-4149672 on Build GRCh38
rs568876969 Chr.17: 4149724-4149724 on Build GRCh38
rs192575758 Chr.17: 4149787-4149787 on Build GRCh38
rs1052907510 Chr.17: 4149531-4149531 on Build GRCh38
rs180873288 Chr.17: 4149960-4149960 on Build GRCh38
rs896844868 Chr.17: 4149811-4149811 on Build GRCh38
rs143079411 Chr.17: 4149892-4149892 on Build GRCh38
rs529621621 Chr.17: 4149830-4149830 on Build GRCh38
rs956047318 Chr.17: 4149913-4149913 on Build GRCh38
rs1026546110 Chr.17: 4149670-4149670 on Build GRCh38
rs1001872654 Chr.17: 4149681-4149681 on Build GRCh38
rs758750553 Chr.17: 4149971-4149971 on Build GRCh38
rs144222377 Chr.17: 4149740-4149740 on Build GRCh38
rs369854397 Chr.17: 4149861-4149861 on Build GRCh38
rs199813802 Chr.17: 4149894-4149894 on Build GRCh38
rs914110009 Chr.17: 4149835-4149835 on Build GRCh38
rs975397883 Chr.17: 4149799-4149803 on Build GRCh38
rs4790573 Chr.17: 4149675-4149675 on Build GRCh38
rs771159912 Chr.17: 4149908-4149908 on Build GRCh38
rs776479968 Chr.17: 4149751-4149751 on Build GRCh38
rs753557822 Chr.17: 4149961-4149961 on Build GRCh38
rs767153423 Chr.17: 4149878-4149878 on Build GRCh38
rs372881821 Chr.17: 4149953-4149953 on Build GRCh38
rs143847336 Chr.17: 4149808-4149809 on Build GRCh38
rs756809009 Chr.17: 4149964-4149964 on Build GRCh38
rs192560332 Chr.17: 4149580-4149580 on Build GRCh38
rs938492723 Chr.17: 4149963-4149963 on Build GRCh38
rs184347707 Chr.17: 4149667-4149667 on Build GRCh38
rs71366591 Chr.17: 4149823-4149823 on Build GRCh38
rs892989821 Chr.17: 4149546-4149546 on Build GRCh38
rs771739005 Chr.17: 4149547-4149547 on Build GRCh38
rs964822403 Chr.17: 4149688-4149688 on Build GRCh38
rs968655010 Chr.17: 4149943-4149943 on Build GRCh38
rs1018109775 Chr.17: 4149684-4149684 on Build GRCh38
rs962660360 Chr.17: 4149791-4149791 on Build GRCh38
rs566843776 Chr.17: 4149674-4149674 on Build GRCh38
rs1039891189 Chr.17: 4149648-4149648 on Build GRCh38
rs921543897 Chr.17: 4149659-4149659 on Build GRCh38
rs772508937 Chr.17: 4149914-4149914 on Build GRCh38
rs997629466 Chr.17: 4149909-4149909 on Build GRCh38
rs945536219 Chr.17: 4149875-4149875 on Build GRCh38
rs759026982 Chr.17: 4149863-4149863 on Build GRCh38
rs765648102 Chr.17: 4149969-4149969 on Build GRCh38
rs909918619 Chr.17: 4149597-4149597 on Build GRCh38
rs921501670 Chr.17: 4149802-4149803 on Build GRCh38
rs776953184 Chr.17: 4149922-4149922 on Build GRCh38
rs988701153 Chr.17: 4149755-4149755 on Build GRCh38
rs947192265 Chr.17: 4149579-4149579 on Build GRCh38
rs775550947 Chr.17: 4149958-4149958 on Build GRCh38
rs184653285 Chr.17: 4149910-4149910 on Build GRCh38
rs770784001 Chr.17: 4149944-4149944 on Build GRCh38
rs370012754 Chr.17: 4149920-4149920 on Build GRCh38
rs1017764281 Chr.17: 4149884-4149884 on Build GRCh38
rs1002989858 Chr.17: 4149629-4149629 on Build GRCh38
rs757679512 Chr.17: 4149898-4149898 on Build GRCh38
rs751549821 Chr.17: 4149871-4149871 on Build GRCh38
rs189292607 Chr.17: 4149930-4149930 on Build GRCh38
rs188839230 Chr.17: 4149734-4149734 on Build GRCh38
rs755676677 Chr.17: 4149893-4149893 on Build GRCh38
rs745342138 Chr.17: 4149543-4149543 on Build GRCh38
rs375782695 Chr.17: 4149655-4149655 on Build GRCh38
rs780587456 Chr.17: 4149972-4149972 on Build GRCh38
rs951525961 Chr.17: 4149571-4149571 on Build GRCh38
rs753407187 Chr.17: 4149895-4149895 on Build GRCh38
rs542445795 Chr.17: 4149848-4149848 on Build GRCh38
rs1044544328 Chr.17: 4149591-4149591 on Build GRCh38
rs774234123 Chr.17: 4149952-4149952 on Build GRCh38
rs879740482 Chr.17: 4149974-4149974 on Build GRCh38
rs1057227387 Chr.17: 4149793-4149793 on Build GRCh38
rs914455235 Chr.17: 4149757-4149757 on Build GRCh38
rs368344135 Chr.17: 4149959-4149959 on Build GRCh38
rs1044232130 Chr.17: 4149781-4149781 on Build GRCh38
rs887991074 Chr.17: 4149862-4149862 on Build GRCh38

View Full Product Details